Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 68545123-68545646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A1
Entrez Gene ID
407738
Gene Name
family with sequence similarity 19 member A1, C-C motif chemokine like
Gene Aliases
TAFA-1, TAFA1
UniGene
Hs.655061
Gene Chromosome Location
Chr.3: 67991592-68546056 on Build GRCh38
Transcripts
NM_001252216.1 NM_213609.3 XM_017006386.1 XM_017006387.1 XM_017006388.1 XM_017006389.1 XM_017006390.1

SNP Detail

SNP ID Chromosome Location
rs904083648 Chr.3: 68545480-68545481 on Build GRCh38
rs922629397 Chr.3: 68545426-68545426 on Build GRCh38
rs578152216 Chr.3: 68545297-68545297 on Build GRCh38
rs1000443800 Chr.3: 68545222-68545222 on Build GRCh38
rs749779441 Chr.3: 68545466-68545466 on Build GRCh38
rs1010232855 Chr.3: 68545240-68545240 on Build GRCh38
rs1049000345 Chr.3: 68545152-68545152 on Build GRCh38
rs3828367 Chr.3: 68545317-68545317 on Build GRCh38
rs1044384946 Chr.3: 68545448-68545448 on Build GRCh38
rs971088324 Chr.3: 68545284-68545284 on Build GRCh38
rs1041859931 Chr.3: 68545431-68545431 on Build GRCh38
rs943313797 Chr.3: 68545398-68545398 on Build GRCh38
rs149720279 Chr.3: 68545165-68545165 on Build GRCh38
rs542937167 Chr.3: 68545345-68545345 on Build GRCh38
rs543807327 Chr.3: 68545315-68545315 on Build GRCh38
rs980750164 Chr.3: 68545357-68545357 on Build GRCh38
rs888903795 Chr.3: 68545611-68545611 on Build GRCh38
rs991539148 Chr.3: 68545264-68545264 on Build GRCh38
rs894692517 Chr.3: 68545568-68545568 on Build GRCh38
rs1033710519 Chr.3: 68545228-68545228 on Build GRCh38
rs528629294 Chr.3: 68545523-68545523 on Build GRCh38
rs557416052 Chr.3: 68545316-68545316 on Build GRCh38
rs961864834 Chr.3: 68545326-68545326 on Build GRCh38
rs926670616 Chr.3: 68545179-68545179 on Build GRCh38
rs903392750 Chr.3: 68545205-68545205 on Build GRCh38
rs945533546 Chr.3: 68545436-68545436 on Build GRCh38
rs1020424247 Chr.3: 68545201-68545201 on Build GRCh38
rs796681227 Chr.3: 68545174-68545174 on Build GRCh38
rs984653404 Chr.3: 68545376-68545376 on Build GRCh38
rs1000389865 Chr.3: 68545535-68545535 on Build GRCh38
rs892754586 Chr.3: 68545237-68545237 on Build GRCh38
rs564601575 Chr.3: 68545588-68545588 on Build GRCh38
rs910434443 Chr.3: 68545382-68545382 on Build GRCh38
rs58766683 Chr.3: 68545223-68545224 on Build GRCh38
rs753017658 Chr.3: 68545192-68545192 on Build GRCh38
rs376873579 Chr.3: 68545286-68545286 on Build GRCh38
rs866436473 Chr.3: 68545158-68545158 on Build GRCh38
rs934061669 Chr.3: 68545428-68545428 on Build GRCh38
rs1040075586 Chr.3: 68545406-68545406 on Build GRCh38
rs116730999 Chr.3: 68545540-68545540 on Build GRCh38
rs866940940 Chr.3: 68545159-68545159 on Build GRCh38
rs9814022 Chr.3: 68545168-68545168 on Build GRCh38
rs76387714 Chr.3: 68545241-68545241 on Build GRCh38
rs1023988244 Chr.3: 68545274-68545274 on Build GRCh38
rs560039645 Chr.3: 68545364-68545364 on Build GRCh38
rs538874783 Chr.3: 68545193-68545193 on Build GRCh38
rs1050040614 Chr.3: 68545564-68545564 on Build GRCh38
rs1056633980 Chr.3: 68545211-68545211 on Build GRCh38
rs972398735 Chr.3: 68545332-68545332 on Build GRCh38
rs771677228 Chr.3: 68545597-68545597 on Build GRCh38
rs777148940 Chr.3: 68545173-68545173 on Build GRCh38
rs778297087 Chr.3: 68545361-68545361 on Build GRCh38
rs1028720876 Chr.3: 68545276-68545276 on Build GRCh38

View Full Product Details