Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 68732874-68733384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A4
Entrez Gene ID
151647
Gene Name
family with sequence similarity 19 member A4, C-C motif chemokine like
Gene Aliases
TAFA-4, TAFA4
UniGene
Hs.187873
Gene Chromosome Location
Chr.3: 68731764-68932610 on Build GRCh38
Transcripts
NM_001005527.2 NM_182522.4 XM_011533371.1 XM_011533372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7132248 c.417G>A p.T139T Chr.3: 68733148-68733148 on Build GRCh38
3357746 c.419G>A p.R140Q Chr.3: 68733146-68733146 on Build GRCh38
5025972 c.418C>T p.R140W Chr.3: 68733147-68733147 on Build GRCh38
2784315 c.416C>T p.T139M Chr.3: 68733149-68733149 on Build GRCh38
4547373 c.420G>A p.R140R Chr.3: 68733145-68733145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951312355 Chr.3: 68733271-68733271 on Build GRCh38
rs749618496 Chr.3: 68733098-68733098 on Build GRCh38
rs182314072 Chr.3: 68733089-68733089 on Build GRCh38
rs748497127 Chr.3: 68733149-68733149 on Build GRCh38
rs761494231 Chr.3: 68733237-68733237 on Build GRCh38
rs755019148 Chr.3: 68732967-68732967 on Build GRCh38
rs929839637 Chr.3: 68733109-68733109 on Build GRCh38
rs774035566 Chr.3: 68733167-68733167 on Build GRCh38
rs763637816 Chr.3: 68733212-68733212 on Build GRCh38
rs116094129 Chr.3: 68732915-68732915 on Build GRCh38
rs368709870 Chr.3: 68733128-68733128 on Build GRCh38
rs371664765 Chr.3: 68733220-68733220 on Build GRCh38
rs146641270 Chr.3: 68733148-68733148 on Build GRCh38
rs116027469 Chr.3: 68733127-68733127 on Build GRCh38
rs776768398 Chr.3: 68733164-68733164 on Build GRCh38
rs766308575 Chr.3: 68733342-68733342 on Build GRCh38
rs369649050 Chr.3: 68732966-68732966 on Build GRCh38
rs200362841 Chr.3: 68733189-68733189 on Build GRCh38
rs868695499 Chr.3: 68733102-68733102 on Build GRCh38
rs989883496 Chr.3: 68733273-68733273 on Build GRCh38
rs374501359 Chr.3: 68733198-68733198 on Build GRCh38
rs971337159 Chr.3: 68733047-68733047 on Build GRCh38
rs957844302 Chr.3: 68733044-68733044 on Build GRCh38
rs974685839 Chr.3: 68733282-68733282 on Build GRCh38
rs537297443 Chr.3: 68732928-68732928 on Build GRCh38
rs557872869 Chr.3: 68732919-68732919 on Build GRCh38
rs762697032 Chr.3: 68733310-68733310 on Build GRCh38
rs115807576 Chr.3: 68733351-68733351 on Build GRCh38
rs942079141 Chr.3: 68733349-68733349 on Build GRCh38
rs777267635 Chr.3: 68733101-68733102 on Build GRCh38
rs929305559 Chr.3: 68732911-68732911 on Build GRCh38
rs781287469 Chr.3: 68732976-68732976 on Build GRCh38
rs187683759 Chr.3: 68733297-68733297 on Build GRCh38
rs933402716 Chr.3: 68733345-68733345 on Build GRCh38
rs755550576 Chr.3: 68733145-68733145 on Build GRCh38
rs763860241 Chr.3: 68733139-68733139 on Build GRCh38
rs564741905 Chr.3: 68733199-68733199 on Build GRCh38
rs1003381302 Chr.3: 68733247-68733247 on Build GRCh38
rs1032420697 Chr.3: 68732985-68732985 on Build GRCh38
rs377521783 Chr.3: 68733133-68733133 on Build GRCh38
rs970433392 Chr.3: 68733242-68733242 on Build GRCh38
rs112376773 Chr.3: 68733279-68733279 on Build GRCh38
rs983512630 Chr.3: 68733114-68733114 on Build GRCh38
rs748647017 Chr.3: 68733094-68733094 on Build GRCh38
rs1047992189 Chr.3: 68733304-68733304 on Build GRCh38
rs909239965 Chr.3: 68733278-68733278 on Build GRCh38
rs560953014 Chr.3: 68733296-68733296 on Build GRCh38
rs748392879 Chr.3: 68733027-68733027 on Build GRCh38
rs372152260 Chr.3: 68733110-68733110 on Build GRCh38
rs369805556 Chr.3: 68733097-68733097 on Build GRCh38
rs760606511 Chr.3: 68733185-68733185 on Build GRCh38
rs386662040 Chr.3: 68733062-68733063 on Build GRCh38
rs767443371 Chr.3: 68733180-68733180 on Build GRCh38
rs866788868 Chr.3: 68733039-68733039 on Build GRCh38
rs751444722 Chr.3: 68733360-68733360 on Build GRCh38
rs984434261 Chr.3: 68733275-68733275 on Build GRCh38
rs1051839360 Chr.3: 68733172-68733172 on Build GRCh38
rs749962950 Chr.3: 68733142-68733142 on Build GRCh38
rs918078936 Chr.3: 68733025-68733025 on Build GRCh38
rs565928869 Chr.3: 68732909-68732909 on Build GRCh38
rs759007554 Chr.3: 68733154-68733154 on Build GRCh38
rs771192908 Chr.3: 68733162-68733162 on Build GRCh38
rs115476173 Chr.3: 68733063-68733063 on Build GRCh38
rs776213164 Chr.3: 68733186-68733186 on Build GRCh38
rs6549143 Chr.3: 68733062-68733062 on Build GRCh38
rs566121386 Chr.3: 68733244-68733244 on Build GRCh38
rs549065025 Chr.3: 68733240-68733240 on Build GRCh38
rs999201656 Chr.3: 68732978-68732978 on Build GRCh38
rs753369130 Chr.3: 68733194-68733194 on Build GRCh38
rs922021122 Chr.3: 68733341-68733341 on Build GRCh38
rs544125621 Chr.3: 68733218-68733218 on Build GRCh38
rs1025481364 Chr.3: 68733252-68733252 on Build GRCh38
rs970567695 Chr.3: 68733092-68733092 on Build GRCh38
rs981209627 Chr.3: 68733060-68733060 on Build GRCh38
rs369406426 Chr.3: 68733140-68733140 on Build GRCh38
rs1030551574 Chr.3: 68732971-68732971 on Build GRCh38
rs754629747 Chr.3: 68733147-68733147 on Build GRCh38
rs770033741 Chr.3: 68733166-68733166 on Build GRCh38
rs942108926 Chr.3: 68732921-68732921 on Build GRCh38
rs767390441 Chr.3: 68733141-68733141 on Build GRCh38
rs529876657 Chr.3: 68733221-68733221 on Build GRCh38
rs746528039 Chr.3: 68733146-68733146 on Build GRCh38
rs576588065 Chr.3: 68733217-68733217 on Build GRCh38
rs978629903 Chr.3: 68733200-68733200 on Build GRCh38
rs773194861 Chr.3: 68733182-68733182 on Build GRCh38
rs556685138 Chr.3: 68732902-68732902 on Build GRCh38
rs373615803 Chr.3: 68733159-68733159 on Build GRCh38
rs193301154 Chr.3: 68733314-68733314 on Build GRCh38
rs761626970 Chr.3: 68733179-68733179 on Build GRCh38
rs200862323 Chr.3: 68733126-68733126 on Build GRCh38
rs748812594 Chr.3: 68733113-68733114 on Build GRCh38
rs893444519 Chr.3: 68733211-68733211 on Build GRCh38
rs933189282 Chr.3: 68733130-68733131 on Build GRCh38
rs771283213 Chr.3: 68733119-68733119 on Build GRCh38

View Full Product Details