Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 68885093-68885596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A4
Entrez Gene ID
151647
Gene Name
family with sequence similarity 19 member A4, C-C motif chemokine like
Gene Aliases
TAFA-4, TAFA4
UniGene
Hs.187873
Gene Chromosome Location
Chr.3: 68731764-68932610 on Build GRCh38
Transcripts
NM_001005527.2 NM_182522.4 XM_011533371.1 XM_011533372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1670334 c.8C>T p.S3F Chr.3: 68885181-68885181 on Build GRCh38
6659295 c.7T>C p.S3P Chr.3: 68885182-68885182 on Build GRCh38
7220239 c.5G>A p.R2K Chr.3: 68885184-68885184 on Build GRCh38
5908284 c.12A>G p.P4P Chr.3: 68885177-68885177 on Build GRCh38
731664 c.5G>T p.R2M Chr.3: 68885184-68885184 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778198587 Chr.3: 68885370-68885370 on Build GRCh38
rs774328652 Chr.3: 68885146-68885146 on Build GRCh38
rs749613557 Chr.3: 68885194-68885194 on Build GRCh38
rs375863275 Chr.3: 68885129-68885129 on Build GRCh38
rs910856156 Chr.3: 68885323-68885323 on Build GRCh38
rs529139894 Chr.3: 68885515-68885515 on Build GRCh38
rs78056889 Chr.3: 68885184-68885184 on Build GRCh38
rs565079928 Chr.3: 68885357-68885357 on Build GRCh38
rs1034771623 Chr.3: 68885491-68885491 on Build GRCh38
rs202083123 Chr.3: 68885163-68885163 on Build GRCh38
rs542063194 Chr.3: 68885280-68885280 on Build GRCh38
rs756697619 Chr.3: 68885177-68885177 on Build GRCh38
rs372601148 Chr.3: 68885160-68885160 on Build GRCh38
rs908464007 Chr.3: 68885136-68885136 on Build GRCh38
rs962985257 Chr.3: 68885466-68885466 on Build GRCh38
rs888527061 Chr.3: 68885467-68885467 on Build GRCh38
rs967565389 Chr.3: 68885542-68885542 on Build GRCh38
rs773321669 Chr.3: 68885397-68885397 on Build GRCh38
rs1024543007 Chr.3: 68885540-68885540 on Build GRCh38
rs569898732 Chr.3: 68885497-68885497 on Build GRCh38
rs972468 Chr.3: 68885552-68885552 on Build GRCh38
rs772440946 Chr.3: 68885152-68885152 on Build GRCh38
rs770993848 Chr.3: 68885456-68885456 on Build GRCh38
rs973888817 Chr.3: 68885473-68885473 on Build GRCh38
rs890437357 Chr.3: 68885226-68885226 on Build GRCh38
rs540698646 Chr.3: 68885290-68885290 on Build GRCh38
rs750139127 Chr.3: 68885183-68885183 on Build GRCh38
rs748343609 Chr.3: 68885204-68885205 on Build GRCh38
rs1050471632 Chr.3: 68885555-68885555 on Build GRCh38
rs774799173 Chr.3: 68885563-68885563 on Build GRCh38
rs370168795 Chr.3: 68885269-68885269 on Build GRCh38
rs755447126 Chr.3: 68885195-68885195 on Build GRCh38
rs1053857701 Chr.3: 68885303-68885303 on Build GRCh38
rs4855535 Chr.3: 68885283-68885283 on Build GRCh38
rs76914752 Chr.3: 68885281-68885281 on Build GRCh38
rs868602633 Chr.3: 68885211-68885211 on Build GRCh38
rs901038434 Chr.3: 68885380-68885380 on Build GRCh38
rs761920358 Chr.3: 68885151-68885151 on Build GRCh38
rs532800013 Chr.3: 68885414-68885414 on Build GRCh38
rs563817435 Chr.3: 68885296-68885296 on Build GRCh38
rs780328683 Chr.3: 68885188-68885188 on Build GRCh38
rs779507574 Chr.3: 68885203-68885203 on Build GRCh38
rs143252845 Chr.3: 68885503-68885504 on Build GRCh38
rs944684385 Chr.3: 68885568-68885568 on Build GRCh38
rs17048099 Chr.3: 68885132-68885132 on Build GRCh38
rs917434198 Chr.3: 68885253-68885253 on Build GRCh38
rs764980998 Chr.3: 68885156-68885156 on Build GRCh38
rs762823201 Chr.3: 68885159-68885159 on Build GRCh38
rs1011070542 Chr.3: 68885297-68885297 on Build GRCh38
rs548914437 Chr.3: 68885532-68885532 on Build GRCh38
rs941165038 Chr.3: 68885142-68885142 on Build GRCh38
rs908454447 Chr.3: 68885464-68885464 on Build GRCh38
rs969094599 Chr.3: 68885367-68885367 on Build GRCh38
rs62256110 Chr.3: 68885432-68885432 on Build GRCh38
rs747284652 Chr.3: 68885236-68885236 on Build GRCh38
rs769180494 Chr.3: 68885143-68885143 on Build GRCh38
rs539749253 Chr.3: 68885223-68885223 on Build GRCh38
rs748281975 Chr.3: 68885198-68885198 on Build GRCh38
rs766398428 Chr.3: 68885499-68885499 on Build GRCh38
rs748655789 Chr.3: 68885205-68885205 on Build GRCh38
rs766999994 Chr.3: 68885181-68885181 on Build GRCh38
rs111761904 Chr.3: 68885326-68885327 on Build GRCh38
rs760975639 Chr.3: 68885155-68885155 on Build GRCh38
rs1022451254 Chr.3: 68885352-68885352 on Build GRCh38
rs780457990 Chr.3: 68885131-68885131 on Build GRCh38
rs775335276 Chr.3: 68885158-68885158 on Build GRCh38
rs1034747575 Chr.3: 68885374-68885374 on Build GRCh38
rs772345813 Chr.3: 68885222-68885222 on Build GRCh38

View Full Product Details