Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 68977592-68978135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EOGT
Entrez Gene ID
285203
Gene Name
EGF domain specific O-linked N-acetylglucosamine transferase
Gene Aliases
AER61, AOS4, C3orf64, EOGT1
UniGene
Hs.518059
Gene Chromosome Location
Chr.3: 68975212-69013961 on Build GRCh38
Transcripts
NM_001278689.1 NM_173654.2 NR_103826.1 XM_005264743.2 XM_011533599.1 XM_011533600.1 XM_011533602.1 XM_011533603.1 XM_017006204.1 XM_017006205.1 XM_017006206.1 XM_017006207.1 XM_017006209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4119962 c.1518T>A p.L506L Chr.3: 68977684-68977684 on Build GRCh38
1495619 c.1203G>A p.L401L Chr.3: 68977747-68977747 on Build GRCh38
6319500 c.1540C>A p.Q514K Chr.3: 68977662-68977662 on Build GRCh38
5928473 c.1475C>T p.T492I Chr.3: 68977727-68977727 on Build GRCh38
4984583 c.1533C>T p.H511H Chr.3: 68977669-68977669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185050622 Chr.3: 68978038-68978038 on Build GRCh38
rs773261304 Chr.3: 68977812-68977812 on Build GRCh38
rs1029860870 Chr.3: 68977870-68977870 on Build GRCh38
rs921385493 Chr.3: 68977902-68977902 on Build GRCh38
rs779311824 Chr.3: 68977903-68977903 on Build GRCh38
rs115047826 Chr.3: 68977786-68977786 on Build GRCh38
rs759007611 Chr.3: 68977761-68977761 on Build GRCh38
rs769384962 Chr.3: 68978067-68978067 on Build GRCh38
rs762829326 Chr.3: 68977808-68977821 on Build GRCh38
rs568378677 Chr.3: 68977966-68977966 on Build GRCh38
rs921772629 Chr.3: 68978082-68978082 on Build GRCh38
rs946416604 Chr.3: 68978046-68978046 on Build GRCh38
rs533004961 Chr.3: 68977828-68977828 on Build GRCh38
rs991946182 Chr.3: 68977916-68977916 on Build GRCh38
rs746138157 Chr.3: 68977790-68977790 on Build GRCh38
rs962744705 Chr.3: 68977907-68977907 on Build GRCh38
rs933763043 Chr.3: 68978106-68978106 on Build GRCh38
rs569367413 Chr.3: 68977862-68977862 on Build GRCh38
rs779419702 Chr.3: 68977632-68977632 on Build GRCh38
rs778355816 Chr.3: 68977762-68977762 on Build GRCh38
rs199832969 Chr.3: 68977736-68977736 on Build GRCh38
rs760176145 Chr.3: 68977732-68977732 on Build GRCh38
rs758952500 Chr.3: 68977650-68977650 on Build GRCh38
rs1002607498 Chr.3: 68978004-68978004 on Build GRCh38
rs768506105 Chr.3: 68977697-68977697 on Build GRCh38
rs761784124 Chr.3: 68977698-68977698 on Build GRCh38
rs755431954 Chr.3: 68977629-68977629 on Build GRCh38
rs988023090 Chr.3: 68978019-68978019 on Build GRCh38
rs747072942 Chr.3: 68977653-68977653 on Build GRCh38
rs887342814 Chr.3: 68977895-68977895 on Build GRCh38
rs192455220 Chr.3: 68977971-68977971 on Build GRCh38
rs1035372965 Chr.3: 68978011-68978011 on Build GRCh38
rs778699457 Chr.3: 68977796-68977796 on Build GRCh38
rs140481649 Chr.3: 68977627-68977627 on Build GRCh38
rs537930744 Chr.3: 68977897-68977897 on Build GRCh38
rs191230655 Chr.3: 68977744-68977744 on Build GRCh38
rs186884712 Chr.3: 68977937-68977937 on Build GRCh38
rs771945573 Chr.3: 68977714-68977714 on Build GRCh38
rs200700226 Chr.3: 68977795-68977795 on Build GRCh38
rs963320837 Chr.3: 68978045-68978045 on Build GRCh38
rs947918120 Chr.3: 68977704-68977704 on Build GRCh38
rs950334659 Chr.3: 68978037-68978037 on Build GRCh38
rs778022693 Chr.3: 68977652-68977652 on Build GRCh38
rs540081981 Chr.3: 68978091-68978091 on Build GRCh38
rs935249284 Chr.3: 68978016-68978016 on Build GRCh38
rs751602094 Chr.3: 68977765-68977765 on Build GRCh38
rs1031822207 Chr.3: 68977713-68977713 on Build GRCh38
rs899971286 Chr.3: 68977840-68977840 on Build GRCh38
rs905638662 Chr.3: 68978003-68978003 on Build GRCh38
rs562487945 Chr.3: 68977984-68977984 on Build GRCh38
rs974186036 Chr.3: 68977874-68977874 on Build GRCh38
rs188086670 Chr.3: 68977735-68977735 on Build GRCh38
rs531297946 Chr.3: 68978014-68978014 on Build GRCh38
rs534143984 Chr.3: 68977993-68977993 on Build GRCh38
rs891105142 Chr.3: 68977961-68977961 on Build GRCh38
rs893309831 Chr.3: 68977967-68977967 on Build GRCh38
rs781127806 Chr.3: 68977787-68977787 on Build GRCh38
rs187491348 Chr.3: 68978043-68978043 on Build GRCh38
rs1011335550 Chr.3: 68977742-68977742 on Build GRCh38
rs374926368 Chr.3: 68977836-68977836 on Build GRCh38
rs776853017 Chr.3: 68977683-68977683 on Build GRCh38
rs759083115 Chr.3: 68977743-68977743 on Build GRCh38
rs562651560 Chr.3: 68977668-68977668 on Build GRCh38
rs969601795 Chr.3: 68977957-68977957 on Build GRCh38
rs1004500491 Chr.3: 68977898-68977898 on Build GRCh38
rs916358461 Chr.3: 68977669-68977669 on Build GRCh38
rs766628622 Chr.3: 68978085-68978085 on Build GRCh38
rs753596888 Chr.3: 68977739-68977739 on Build GRCh38
rs934928632 Chr.3: 68977964-68977964 on Build GRCh38
rs999047805 Chr.3: 68977654-68977654 on Build GRCh38
rs377589190 Chr.3: 68977799-68977799 on Build GRCh38
rs748705086 Chr.3: 68977633-68977633 on Build GRCh38
rs527256891 Chr.3: 68977760-68977760 on Build GRCh38
rs769913561 Chr.3: 68977784-68977784 on Build GRCh38
rs891409527 Chr.3: 68977644-68977644 on Build GRCh38
rs531299546 Chr.3: 68977711-68977711 on Build GRCh38
rs370927829 Chr.3: 68977809-68977809 on Build GRCh38
rs991627397 Chr.3: 68977666-68977666 on Build GRCh38
rs549700308 Chr.3: 68977856-68977856 on Build GRCh38
rs969832931 Chr.3: 68977754-68977754 on Build GRCh38
rs899563649 Chr.3: 68978083-68978083 on Build GRCh38
rs772737749 Chr.3: 68977724-68977724 on Build GRCh38
rs746063649 Chr.3: 68977689-68977689 on Build GRCh38
rs1023126139 Chr.3: 68977998-68977998 on Build GRCh38

View Full Product Details