Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69104794-69105291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL6IP5
Entrez Gene ID
10550
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 5
Gene Aliases
DERP11, GTRAP3-18, HSPC127, JWA, PRAF3, Yip6b, addicsin, hp22, jmx
UniGene
Hs.518060
Gene Chromosome Location
Chr.3: 69084939-69106088 on Build GRCh38
Transcripts
NM_006407.3

SNP Detail

SNP ID Chromosome Location
rs1055778764 Chr.3: 69105121-69105121 on Build GRCh38
rs540702915 Chr.3: 69105134-69105134 on Build GRCh38
rs935014842 Chr.3: 69104856-69104856 on Build GRCh38
rs1023469916 Chr.3: 69105066-69105066 on Build GRCh38
rs373189064 Chr.3: 69104911-69104911 on Build GRCh38
rs765883756 Chr.3: 69105043-69105043 on Build GRCh38
rs1007327336 Chr.3: 69105217-69105217 on Build GRCh38
rs913778204 Chr.3: 69105045-69105045 on Build GRCh38
rs748978153 Chr.3: 69105263-69105263 on Build GRCh38
rs186177589 Chr.3: 69104907-69104907 on Build GRCh38
rs374041788 Chr.3: 69105169-69105169 on Build GRCh38
rs751259299 Chr.3: 69105132-69105132 on Build GRCh38
rs973564389 Chr.3: 69104956-69104956 on Build GRCh38
rs898592309 Chr.3: 69105028-69105028 on Build GRCh38
rs904572938 Chr.3: 69104849-69104849 on Build GRCh38
rs953156166 Chr.3: 69104965-69104965 on Build GRCh38
rs915017375 Chr.3: 69104883-69104883 on Build GRCh38
rs745898795 Chr.3: 69105008-69105008 on Build GRCh38
rs1043557972 Chr.3: 69105078-69105078 on Build GRCh38
rs1013383640 Chr.3: 69105033-69105033 on Build GRCh38
rs1802403 Chr.3: 69105265-69105265 on Build GRCh38
rs1017323926 Chr.3: 69105009-69105009 on Build GRCh38
rs1043536154 Chr.3: 69104842-69104842 on Build GRCh38
rs114156698 Chr.3: 69104898-69104898 on Build GRCh38
rs927858014 Chr.3: 69104839-69104839 on Build GRCh38
rs946548533 Chr.3: 69105077-69105077 on Build GRCh38
rs985815450 Chr.3: 69104990-69104990 on Build GRCh38
rs751066787 Chr.3: 69104931-69104931 on Build GRCh38
rs1036582465 Chr.3: 69105156-69105156 on Build GRCh38
rs911544592 Chr.3: 69104993-69104993 on Build GRCh38
rs762255958 Chr.3: 69104971-69104971 on Build GRCh38
rs904629229 Chr.3: 69105080-69105080 on Build GRCh38
rs754399448 Chr.3: 69105166-69105166 on Build GRCh38
rs3821564 Chr.3: 69105235-69105235 on Build GRCh38
rs1052084484 Chr.3: 69104863-69104863 on Build GRCh38
rs543359534 Chr.3: 69105196-69105196 on Build GRCh38
rs987820186 Chr.3: 69105145-69105145 on Build GRCh38
rs759769308 Chr.3: 69104926-69104930 on Build GRCh38
rs556573912 Chr.3: 69104821-69104821 on Build GRCh38
rs1027853780 Chr.3: 69104957-69104957 on Build GRCh38
rs895840571 Chr.3: 69105130-69105130 on Build GRCh38
rs1038379808 Chr.3: 69105254-69105254 on Build GRCh38
rs937429290 Chr.3: 69105104-69105104 on Build GRCh38
rs776971786 Chr.3: 69104925-69104925 on Build GRCh38
rs530184038 Chr.3: 69104971-69104971 on Build GRCh38
rs946547345 Chr.3: 69104834-69104834 on Build GRCh38
rs7039 Chr.3: 69105192-69105192 on Build GRCh38
rs756962806 Chr.3: 69104952-69104952 on Build GRCh38
rs1802402 Chr.3: 69105006-69105006 on Build GRCh38
rs969245999 Chr.3: 69105070-69105070 on Build GRCh38
rs796189353 Chr.3: 69104926-69104926 on Build GRCh38
rs1802401 Chr.3: 69105135-69105135 on Build GRCh38
rs1051185951 Chr.3: 69105221-69105221 on Build GRCh38
rs1034960076 Chr.3: 69105099-69105101 on Build GRCh38
rs777915242 Chr.3: 69105245-69105245 on Build GRCh38
rs373944686 Chr.3: 69104970-69104971 on Build GRCh38
rs115181949 Chr.3: 69104917-69104917 on Build GRCh38
rs953276671 Chr.3: 69105209-69105209 on Build GRCh38

View Full Product Details