Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69171421-69171926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4B
Entrez Gene ID
23150
Gene Name
FERM domain containing 4B
Gene Aliases
6030440G05Rik, GRSP1
UniGene
Hs.371681
Gene Chromosome Location
Chr.3: 69168782-69541340 on Build GRCh38
Transcripts
NM_015123.2 XM_005264722.1 XM_005264723.1 XM_017005989.1 XM_017005990.1 XM_017005991.1 XM_017005992.1 XM_017005993.1 XM_017005994.1 XM_017005995.1 XM_017005996.1 XM_017005997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5653471 c.3100G>C p.V1034L Chr.3: 69171866-69171866 on Build GRCh38
4405508 c.3080A>G p.D1027G Chr.3: 69171886-69171886 on Build GRCh38
4584618 c.3041A>G p.N1014S Chr.3: 69171925-69171925 on Build GRCh38
6886086 c.3064C>T p.L1022F Chr.3: 69171902-69171902 on Build GRCh38
6886087 c.2902C>T p.L968F Chr.3: 69171902-69171902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573865880 Chr.3: 69171830-69171830 on Build GRCh38
rs773187186 Chr.3: 69171832-69171832 on Build GRCh38
rs571281065 Chr.3: 69171849-69171849 on Build GRCh38
rs115600417 Chr.3: 69171466-69171466 on Build GRCh38
rs565663900 Chr.3: 69171707-69171707 on Build GRCh38
rs936936865 Chr.3: 69171670-69171670 on Build GRCh38
rs138676876 Chr.3: 69171485-69171485 on Build GRCh38
rs73835744 Chr.3: 69171495-69171495 on Build GRCh38
rs868673420 Chr.3: 69171799-69171799 on Build GRCh38
rs758262041 Chr.3: 69171754-69171754 on Build GRCh38
rs556139143 Chr.3: 69171529-69171529 on Build GRCh38
rs371111532 Chr.3: 69171829-69171829 on Build GRCh38
rs773739478 Chr.3: 69171839-69171839 on Build GRCh38
rs28529735 Chr.3: 69171533-69171533 on Build GRCh38
rs914769005 Chr.3: 69171801-69171801 on Build GRCh38
rs771781564 Chr.3: 69171836-69171836 on Build GRCh38
rs529094319 Chr.3: 69171687-69171687 on Build GRCh38
rs990479116 Chr.3: 69171633-69171633 on Build GRCh38
rs528050141 Chr.3: 69171780-69171780 on Build GRCh38
rs760082050 Chr.3: 69171873-69171873 on Build GRCh38
rs984611694 Chr.3: 69171858-69171858 on Build GRCh38
rs1019685037 Chr.3: 69171497-69171497 on Build GRCh38
rs752717550 Chr.3: 69171885-69171885 on Build GRCh38
rs147379378 Chr.3: 69171793-69171793 on Build GRCh38
rs61104752 Chr.3: 69171635-69171635 on Build GRCh38
rs541457062 Chr.3: 69171552-69171552 on Build GRCh38
rs376988405 Chr.3: 69171818-69171818 on Build GRCh38
rs965431438 Chr.3: 69171499-69171499 on Build GRCh38
rs769754579 Chr.3: 69171813-69171813 on Build GRCh38
rs934833514 Chr.3: 69171483-69171483 on Build GRCh38
rs954541168 Chr.3: 69171776-69171776 on Build GRCh38
rs1032317905 Chr.3: 69171476-69171476 on Build GRCh38
rs761568833 Chr.3: 69171834-69171834 on Build GRCh38
rs779811456 Chr.3: 69171816-69171816 on Build GRCh38
rs754342263 Chr.3: 69171866-69171866 on Build GRCh38
rs950706247 Chr.3: 69171534-69171534 on Build GRCh38
rs879131865 Chr.3: 69171809-69171809 on Build GRCh38
rs987206179 Chr.3: 69171777-69171777 on Build GRCh38
rs921576341 Chr.3: 69171770-69171770 on Build GRCh38
rs776062673 Chr.3: 69171697-69171697 on Build GRCh38
rs79609628 Chr.3: 69171700-69171700 on Build GRCh38
rs909112810 Chr.3: 69171592-69171592 on Build GRCh38
rs142730108 Chr.3: 69171622-69171622 on Build GRCh38
rs1019534786 Chr.3: 69171789-69171789 on Build GRCh38
rs543555068 Chr.3: 69171644-69171644 on Build GRCh38
rs1044360590 Chr.3: 69171563-69171563 on Build GRCh38
rs992975503 Chr.3: 69171586-69171586 on Build GRCh38
rs937692133 Chr.3: 69171607-69171607 on Build GRCh38
rs912703252 Chr.3: 69171471-69171471 on Build GRCh38
rs890022664 Chr.3: 69171732-69171732 on Build GRCh38
rs761068147 Chr.3: 69171843-69171843 on Build GRCh38
rs563334628 Chr.3: 69171666-69171666 on Build GRCh38
rs556490061 Chr.3: 69171470-69171470 on Build GRCh38
rs984703477 Chr.3: 69171559-69171559 on Build GRCh38
rs765289548 Chr.3: 69171875-69171875 on Build GRCh38
rs976324231 Chr.3: 69171531-69171531 on Build GRCh38
rs1005125822 Chr.3: 69171667-69171667 on Build GRCh38
rs780109217 Chr.3: 69171788-69171788 on Build GRCh38
rs778638589 Chr.3: 69171645-69171645 on Build GRCh38
rs758625950 Chr.3: 69171892-69171892 on Build GRCh38

View Full Product Details