Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69181573-69182074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4B
Entrez Gene ID
23150
Gene Name
FERM domain containing 4B
Gene Aliases
6030440G05Rik, GRSP1
UniGene
Hs.371681
Gene Chromosome Location
Chr.3: 69168782-69541340 on Build GRCh38
Transcripts
NM_015123.2 XM_005264722.1 XM_005264723.1 XM_017005989.1 XM_017005990.1 XM_017005991.1 XM_017005992.1 XM_017005993.1 XM_017005994.1 XM_017005995.1 XM_017005996.1 XM_017005997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3916640 c.2138C>T p.S713F Chr.3: 69181612-69181612 on Build GRCh38
2784616 c.1878A>G p.E626E Chr.3: 69181710-69181710 on Build GRCh38
6308199 c.2043C>A p.P681P Chr.3: 69181707-69181707 on Build GRCh38
3597267 c.2176C>T p.L726F Chr.3: 69181574-69181574 on Build GRCh38
4603499 c.2168_2169insGCA p.T723_E724insQ Chr.3: 69181581-69181582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772253988 Chr.3: 69181706-69181706 on Build GRCh38
rs538012715 Chr.3: 69181679-69181679 on Build GRCh38
rs957301773 Chr.3: 69181859-69181859 on Build GRCh38
rs992736419 Chr.3: 69182016-69182027 on Build GRCh38
rs889389241 Chr.3: 69181824-69181824 on Build GRCh38
rs750145740 Chr.3: 69181754-69181754 on Build GRCh38
rs981222551 Chr.3: 69181941-69181941 on Build GRCh38
rs9836305 Chr.3: 69181650-69181650 on Build GRCh38
rs1038311725 Chr.3: 69182023-69182023 on Build GRCh38
rs1009177483 Chr.3: 69181825-69181825 on Build GRCh38
rs748402987 Chr.3: 69181645-69181645 on Build GRCh38
rs1022655712 Chr.3: 69181902-69181902 on Build GRCh38
rs542382926 Chr.3: 69182032-69182032 on Build GRCh38
rs945694153 Chr.3: 69181733-69181733 on Build GRCh38
rs1034124504 Chr.3: 69182003-69182003 on Build GRCh38
rs763674289 Chr.3: 69181668-69181668 on Build GRCh38
rs747063842 Chr.3: 69181919-69181919 on Build GRCh38
rs139294486 Chr.3: 69181808-69181808 on Build GRCh38
rs777144004 Chr.3: 69181620-69181620 on Build GRCh38
rs562922918 Chr.3: 69181889-69181889 on Build GRCh38
rs747229195 Chr.3: 69181708-69181708 on Build GRCh38
rs958517284 Chr.3: 69182016-69182021 on Build GRCh38
rs201372509 Chr.3: 69181678-69181678 on Build GRCh38
rs960903632 Chr.3: 69181945-69181945 on Build GRCh38
rs969765684 Chr.3: 69181935-69181935 on Build GRCh38
rs763899131 Chr.3: 69181720-69181720 on Build GRCh38
rs145470885 Chr.3: 69182047-69182047 on Build GRCh38
rs775634806 Chr.3: 69181653-69181653 on Build GRCh38
rs766985565 Chr.3: 69181745-69181745 on Build GRCh38
rs546241707 Chr.3: 69181858-69181858 on Build GRCh38
rs542407367 Chr.3: 69181827-69181827 on Build GRCh38
rs374993549 Chr.3: 69182022-69182022 on Build GRCh38
rs867581914 Chr.3: 69181962-69181962 on Build GRCh38
rs755369529 Chr.3: 69181692-69181692 on Build GRCh38
rs202099937 Chr.3: 69181696-69181696 on Build GRCh38
rs527581690 Chr.3: 69181640-69181640 on Build GRCh38
rs201787281 Chr.3: 69181649-69181649 on Build GRCh38
rs141409273 Chr.3: 69181764-69181764 on Build GRCh38
rs908782076 Chr.3: 69181964-69181964 on Build GRCh38
rs996905823 Chr.3: 69181779-69181779 on Build GRCh38
rs561729822 Chr.3: 69182045-69182045 on Build GRCh38
rs992684398 Chr.3: 69181709-69181709 on Build GRCh38
rs766029125 Chr.3: 69181716-69181717 on Build GRCh38
rs200135116 Chr.3: 69181641-69181641 on Build GRCh38
rs928083070 Chr.3: 69181943-69181943 on Build GRCh38
rs201389931 Chr.3: 69181628-69181628 on Build GRCh38
rs756802830 Chr.3: 69181629-69181629 on Build GRCh38
rs762953594 Chr.3: 69181719-69181719 on Build GRCh38
rs749868604 Chr.3: 69181689-69181689 on Build GRCh38
rs993626689 Chr.3: 69181690-69181690 on Build GRCh38
rs952195776 Chr.3: 69181934-69181934 on Build GRCh38
rs774232697 Chr.3: 69181644-69181644 on Build GRCh38
rs780396995 Chr.3: 69181685-69181685 on Build GRCh38
rs1041506130 Chr.3: 69181778-69181778 on Build GRCh38
rs921278475 Chr.3: 69182043-69182043 on Build GRCh38
rs915648769 Chr.3: 69181597-69181597 on Build GRCh38
rs150833703 Chr.3: 69181804-69181804 on Build GRCh38
rs770310520 Chr.3: 69181635-69181635 on Build GRCh38
rs993903697 Chr.3: 69181884-69181884 on Build GRCh38
rs780589579 Chr.3: 69181638-69181638 on Build GRCh38
rs371581720 Chr.3: 69181632-69181632 on Build GRCh38
rs1005356718 Chr.3: 69181988-69181988 on Build GRCh38
rs553642909 Chr.3: 69182015-69182015 on Build GRCh38
rs748125813 Chr.3: 69181702-69181702 on Build GRCh38
rs775089962 Chr.3: 69181717-69181717 on Build GRCh38
rs200224295 Chr.3: 69181684-69181684 on Build GRCh38
rs752658505 Chr.3: 69181599-69181599 on Build GRCh38
rs762786190 Chr.3: 69181654-69181654 on Build GRCh38
rs746193724 Chr.3: 69181634-69181634 on Build GRCh38
rs772376147 Chr.3: 69181646-69181646 on Build GRCh38
rs757965783 Chr.3: 69181609-69181609 on Build GRCh38
rs375953069 Chr.3: 69181598-69181598 on Build GRCh38
rs570328726 Chr.3: 69181647-69181647 on Build GRCh38
rs773936502 Chr.3: 69181722-69181722 on Build GRCh38
rs576694870 Chr.3: 69181707-69181707 on Build GRCh38
rs761280389 Chr.3: 69181744-69181744 on Build GRCh38
rs769769404 Chr.3: 69181714-69181714 on Build GRCh38

View Full Product Details