Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 71683225-71683727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E3
Entrez Gene ID
317649
Gene Name
eukaryotic translation initiation factor 4E family member 3
Gene Aliases
eIF-4E3, eIF4E-3
UniGene
Hs.581355
Gene Chromosome Location
Chr.3: 71679289-71754831 on Build GRCh38
Transcripts
NM_001134649.2 NM_001134650.1 NM_001134651.1 NM_001282886.1 NM_173359.4 XM_011533651.2 XM_017006278.1 XM_017006279.1 XM_017006280.1 XM_017006281.1 XM_017006282.1

SNP Detail

SNP ID Chromosome Location
rs139416223 Chr.3: 71683617-71683617 on Build GRCh38
rs1049340620 Chr.3: 71683452-71683452 on Build GRCh38
rs1003296908 Chr.3: 71683650-71683650 on Build GRCh38
rs563104076 Chr.3: 71683606-71683606 on Build GRCh38
rs922793720 Chr.3: 71683459-71683459 on Build GRCh38
rs560423081 Chr.3: 71683621-71683621 on Build GRCh38
rs1011410567 Chr.3: 71683611-71683611 on Build GRCh38
rs967285980 Chr.3: 71683656-71683656 on Build GRCh38
rs944769421 Chr.3: 71683371-71683371 on Build GRCh38
rs956791341 Chr.3: 71683395-71683395 on Build GRCh38
rs747807909 Chr.3: 71683324-71683324 on Build GRCh38
rs976238505 Chr.3: 71683385-71683385 on Build GRCh38
rs544467869 Chr.3: 71683589-71683589 on Build GRCh38
rs1021532330 Chr.3: 71683294-71683294 on Build GRCh38
rs966539665 Chr.3: 71683685-71683685 on Build GRCh38
rs982157461 Chr.3: 71683388-71683388 on Build GRCh38
rs976095838 Chr.3: 71683447-71683447 on Build GRCh38
rs899056322 Chr.3: 71683270-71683270 on Build GRCh38
rs924028440 Chr.3: 71683370-71683370 on Build GRCh38
rs1023453233 Chr.3: 71683319-71683319 on Build GRCh38
rs773615896 Chr.3: 71683471-71683471 on Build GRCh38
rs888027807 Chr.3: 71683491-71683491 on Build GRCh38
rs79502136 Chr.3: 71683410-71683410 on Build GRCh38
rs1031079203 Chr.3: 71683393-71683393 on Build GRCh38
rs906358738 Chr.3: 71683627-71683627 on Build GRCh38
rs770127182 Chr.3: 71683423-71683423 on Build GRCh38
rs1030811557 Chr.3: 71683697-71683697 on Build GRCh38
rs968490074 Chr.3: 71683305-71683305 on Build GRCh38
rs915256359 Chr.3: 71683412-71683412 on Build GRCh38
rs544126983 Chr.3: 71683658-71683658 on Build GRCh38
rs906967732 Chr.3: 71683569-71683569 on Build GRCh38
rs940358600 Chr.3: 71683492-71683492 on Build GRCh38
rs763246840 Chr.3: 71683480-71683480 on Build GRCh38
rs574430499 Chr.3: 71683583-71683583 on Build GRCh38
rs189755359 Chr.3: 71683436-71683436 on Build GRCh38
rs1030978010 Chr.3: 71683678-71683678 on Build GRCh38
rs114335669 Chr.3: 71683529-71683529 on Build GRCh38
rs1002599304 Chr.3: 71683578-71683578 on Build GRCh38
rs970990814 Chr.3: 71683350-71683350 on Build GRCh38
rs181344301 Chr.3: 71683510-71683510 on Build GRCh38
rs577159247 Chr.3: 71683670-71683670 on Build GRCh38

View Full Product Details