Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 71684370-71684909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E3
Entrez Gene ID
317649
Gene Name
eukaryotic translation initiation factor 4E family member 3
Gene Aliases
eIF-4E3, eIF4E-3
UniGene
Hs.581355
Gene Chromosome Location
Chr.3: 71679289-71754831 on Build GRCh38
Transcripts
NM_001134649.2 NM_001134650.1 NM_001134651.1 NM_001282886.1 NM_173359.4 XM_011533651.2 XM_017006278.1 XM_017006279.1 XM_017006280.1 XM_017006281.1 XM_017006282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4150333 c.629-1G>C p.? Chr.3: 71684729-71684729 on Build GRCh38
4680647 c.344G>A p.R115H Chr.3: 71684695-71684695 on Build GRCh38
5399994 c.630C>T p.P210P Chr.3: 71684727-71684727 on Build GRCh38
4680646 c.662G>A p.R221H Chr.3: 71684695-71684695 on Build GRCh38
4680649 c.311C>T p.P104L Chr.3: 71684728-71684728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149660263 Chr.3: 71684856-71684856 on Build GRCh38
rs947021503 Chr.3: 71684568-71684568 on Build GRCh38
rs932543412 Chr.3: 71684617-71684617 on Build GRCh38
rs763353443 Chr.3: 71684659-71684659 on Build GRCh38
rs777224016 Chr.3: 71684796-71684796 on Build GRCh38
rs932708622 Chr.3: 71684656-71684656 on Build GRCh38
rs548414807 Chr.3: 71684731-71684731 on Build GRCh38
rs375802441 Chr.3: 71684762-71684763 on Build GRCh38
rs563819259 Chr.3: 71684517-71684517 on Build GRCh38
rs186368414 Chr.3: 71684492-71684492 on Build GRCh38
rs551647009 Chr.3: 71684775-71684775 on Build GRCh38
rs763967491 Chr.3: 71684851-71684854 on Build GRCh38
rs939055653 Chr.3: 71684545-71684545 on Build GRCh38
rs762169267 Chr.3: 71684679-71684679 on Build GRCh38
rs1015605865 Chr.3: 71684865-71684866 on Build GRCh38
rs879362238 Chr.3: 71684408-71684409 on Build GRCh38
rs544357074 Chr.3: 71684850-71684851 on Build GRCh38
rs904863210 Chr.3: 71684518-71684518 on Build GRCh38
rs995802905 Chr.3: 71684873-71684873 on Build GRCh38
rs534295694 Chr.3: 71684869-71684869 on Build GRCh38
rs544889111 Chr.3: 71684448-71684448 on Build GRCh38
rs199847532 Chr.3: 71684751-71684751 on Build GRCh38
rs773659075 Chr.3: 71684740-71684740 on Build GRCh38
rs899894392 Chr.3: 71684426-71684426 on Build GRCh38
rs901116902 Chr.3: 71684616-71684616 on Build GRCh38
rs972632073 Chr.3: 71684516-71684516 on Build GRCh38
rs60485726 Chr.3: 71684674-71684674 on Build GRCh38
rs181352857 Chr.3: 71684473-71684473 on Build GRCh38
rs75378269 Chr.3: 71684407-71684408 on Build GRCh38
rs774180480 Chr.3: 71684877-71684877 on Build GRCh38
rs749832311 Chr.3: 71684756-71684756 on Build GRCh38
rs991171063 Chr.3: 71684540-71684540 on Build GRCh38
rs978558202 Chr.3: 71684570-71684570 on Build GRCh38
rs770263319 Chr.3: 71684739-71684740 on Build GRCh38
rs200576271 Chr.3: 71684676-71684676 on Build GRCh38
rs200683614 Chr.3: 71684523-71684523 on Build GRCh38
rs866136283 Chr.3: 71684705-71684705 on Build GRCh38
rs749685562 Chr.3: 71684805-71684805 on Build GRCh38
rs752533191 Chr.3: 71684696-71684696 on Build GRCh38
rs750498700 Chr.3: 71684682-71684682 on Build GRCh38
rs769564093 Chr.3: 71684632-71684632 on Build GRCh38
rs889140370 Chr.3: 71684680-71684680 on Build GRCh38
rs1006633446 Chr.3: 71684724-71684724 on Build GRCh38
rs1016744321 Chr.3: 71684482-71684482 on Build GRCh38
rs562915527 Chr.3: 71684458-71684458 on Build GRCh38
rs374822068 Chr.3: 71684681-71684681 on Build GRCh38
rs546180609 Chr.3: 71684534-71684534 on Build GRCh38
rs776053611 Chr.3: 71684633-71684633 on Build GRCh38
rs747816857 Chr.3: 71684576-71684576 on Build GRCh38
rs897868641 Chr.3: 71684794-71684794 on Build GRCh38
rs751527021 Chr.3: 71684757-71684757 on Build GRCh38
rs1010662745 Chr.3: 71684414-71684414 on Build GRCh38
rs533128061 Chr.3: 71684772-71684772 on Build GRCh38
rs1006337411 Chr.3: 71684475-71684475 on Build GRCh38
rs994498522 Chr.3: 71684415-71684415 on Build GRCh38
rs758241545 Chr.3: 71684746-71684746 on Build GRCh38
rs528220937 Chr.3: 71684519-71684519 on Build GRCh38
rs1049622355 Chr.3: 71684670-71684670 on Build GRCh38
rs997475433 Chr.3: 71684456-71684456 on Build GRCh38
rs1043572414 Chr.3: 71684424-71684424 on Build GRCh38
rs934521179 Chr.3: 71684588-71684588 on Build GRCh38
rs201647025 Chr.3: 71684677-71684677 on Build GRCh38
rs913783882 Chr.3: 71684573-71684573 on Build GRCh38
rs955380972 Chr.3: 71684464-71684465 on Build GRCh38
rs996952570 Chr.3: 71684457-71684460 on Build GRCh38
rs1041237673 Chr.3: 71684598-71684598 on Build GRCh38
rs1043919372 Chr.3: 71684596-71684596 on Build GRCh38
rs1044794238 Chr.3: 71684511-71684511 on Build GRCh38
rs1023203972 Chr.3: 71684875-71684875 on Build GRCh38
rs1035835458 Chr.3: 71684827-71684827 on Build GRCh38
rs757072702 Chr.3: 71684765-71684765 on Build GRCh38
rs561357317 Chr.3: 71684544-71684544 on Build GRCh38
rs760948130 Chr.3: 71684695-71684695 on Build GRCh38
rs201373516 Chr.3: 71684747-71684747 on Build GRCh38
rs192207779 Chr.3: 71684597-71684597 on Build GRCh38
rs578137884 Chr.3: 71684491-71684491 on Build GRCh38
rs745356457 Chr.3: 71684773-71684773 on Build GRCh38
rs147569869 Chr.3: 71684725-71684725 on Build GRCh38
rs201952234 Chr.3: 71684742-71684742 on Build GRCh38
rs1051226864 Chr.3: 71684825-71684825 on Build GRCh38

View Full Product Details