Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 84958860-84959359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM2
Entrez Gene ID
253559
Gene Name
cell adhesion molecule 2
Gene Aliases
IGSF4D, NECL3, Necl-3, SynCAM 2, synCAM2
UniGene
Hs.164578
Gene Chromosome Location
Chr.3: 84958982-86074429 on Build GRCh38
Transcripts
NM_001167674.1 NM_001167675.1 XM_006713081.3 XM_006713082.3 XM_017006062.1

SNP Detail

SNP ID Chromosome Location
rs1011693607 Chr.3: 84959122-84959122 on Build GRCh38
rs560153650 Chr.3: 84959078-84959095 on Build GRCh38
rs115294369 Chr.3: 84958962-84958962 on Build GRCh38
rs941810726 Chr.3: 84959005-84959006 on Build GRCh38
rs1052754070 Chr.3: 84959132-84959132 on Build GRCh38
rs921750749 Chr.3: 84959228-84959228 on Build GRCh38
rs928267163 Chr.3: 84958989-84958989 on Build GRCh38
rs935679840 Chr.3: 84959081-84959081 on Build GRCh38
rs970704892 Chr.3: 84958916-84958916 on Build GRCh38
rs970274615 Chr.3: 84959138-84959138 on Build GRCh38
rs1039271566 Chr.3: 84959021-84959021 on Build GRCh38
rs931482076 Chr.3: 84959055-84959066 on Build GRCh38
rs999848627 Chr.3: 84958955-84958955 on Build GRCh38
rs775751042 Chr.3: 84959309-84959309 on Build GRCh38
rs544069573 Chr.3: 84958998-84958999 on Build GRCh38
rs1033749331 Chr.3: 84958990-84958990 on Build GRCh38
rs891100030 Chr.3: 84959038-84959038 on Build GRCh38
rs902439365 Chr.3: 84959302-84959302 on Build GRCh38
rs190135150 Chr.3: 84959060-84959060 on Build GRCh38
rs1002727880 Chr.3: 84958984-84958984 on Build GRCh38
rs573056862 Chr.3: 84959142-84959142 on Build GRCh38
rs571371046 Chr.3: 84958975-84958975 on Build GRCh38
rs533756630 Chr.3: 84959328-84959328 on Build GRCh38
rs1048654447 Chr.3: 84959293-84959304 on Build GRCh38
rs554590383 Chr.3: 84959128-84959128 on Build GRCh38
rs987678842 Chr.3: 84959031-84959031 on Build GRCh38
rs1031730944 Chr.3: 84958882-84958882 on Build GRCh38
rs1032027104 Chr.3: 84958957-84958957 on Build GRCh38
rs905718936 Chr.3: 84959149-84959149 on Build GRCh38
rs908911935 Chr.3: 84959210-84959210 on Build GRCh38
rs867312717 Chr.3: 84959155-84959155 on Build GRCh38
rs1001350738 Chr.3: 84959150-84959150 on Build GRCh38
rs995247656 Chr.3: 84959275-84959275 on Build GRCh38
rs548633223 Chr.3: 84958917-84958917 on Build GRCh38
rs4528960 Chr.3: 84959226-84959226 on Build GRCh38
rs1039220274 Chr.3: 84959056-84959056 on Build GRCh38
rs943529625 Chr.3: 84959041-84959041 on Build GRCh38
rs756984416 Chr.3: 84958926-84958926 on Build GRCh38
rs550687090 Chr.3: 84959000-84959000 on Build GRCh38
rs963459606 Chr.3: 84959273-84959273 on Build GRCh38
rs893228116 Chr.3: 84959117-84959117 on Build GRCh38
rs950710571 Chr.3: 84959301-84959301 on Build GRCh38
rs1038777727 Chr.3: 84958886-84958886 on Build GRCh38
rs1045663368 Chr.3: 84959147-84959147 on Build GRCh38
rs891719580 Chr.3: 84959141-84959141 on Build GRCh38
rs950928121 Chr.3: 84959014-84959014 on Build GRCh38
rs904218531 Chr.3: 84958891-84958891 on Build GRCh38
rs965266575 Chr.3: 84959336-84959336 on Build GRCh38
rs560816156 Chr.3: 84958947-84958947 on Build GRCh38
rs1023180222 Chr.3: 84958909-84958909 on Build GRCh38
rs536222634 Chr.3: 84959094-84959094 on Build GRCh38
rs900373688 Chr.3: 84959026-84959026 on Build GRCh38
rs1014105337 Chr.3: 84958970-84958970 on Build GRCh38
rs987245635 Chr.3: 84958961-84958961 on Build GRCh38
rs969876946 Chr.3: 84958973-84958973 on Build GRCh38
rs1024106014 Chr.3: 84958971-84958971 on Build GRCh38
rs1248861 Chr.3: 84958897-84958897 on Build GRCh38
rs912130460 Chr.3: 84959322-84959322 on Build GRCh38
rs919508712 Chr.3: 84959006-84959006 on Build GRCh38
rs912097940 Chr.3: 84959035-84959035 on Build GRCh38
rs1026309908 Chr.3: 84959291-84959291 on Build GRCh38
rs957510209 Chr.3: 84958895-84958895 on Build GRCh38
rs954605108 Chr.3: 84958941-84958941 on Build GRCh38
rs187187703 Chr.3: 84958992-84958992 on Build GRCh38
rs908965971 Chr.3: 84959001-84959002 on Build GRCh38
rs907996937 Chr.3: 84959085-84959085 on Build GRCh38
rs552766400 Chr.3: 84958972-84958972 on Build GRCh38
rs1028696894 Chr.3: 84958939-84958939 on Build GRCh38

View Full Product Details