Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 85912286-85912794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM2
Entrez Gene ID
253559
Gene Name
cell adhesion molecule 2
Gene Aliases
IGSF4D, NECL3, Necl-3, SynCAM 2, synCAM2
UniGene
Hs.164578
Gene Chromosome Location
Chr.3: 84958982-86074429 on Build GRCh38
Transcripts
NM_001167674.1 NM_001167675.1 NM_001256502.1 NM_001256503.1 NM_001256504.1 NM_001256505.1 NM_153184.3 XM_006713081.3 XM_006713082.3 XM_017006062.1 XM_017006063.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4120412 c.610A>T p.S204C Chr.3: 85912453-85912453 on Build GRCh38
1693020 c.653C>T p.S218F Chr.3: 85912496-85912496 on Build GRCh38
4930883 c.534A>T p.V178V Chr.3: 85912377-85912377 on Build GRCh38
7193109 c.569delC p.T190fs*25 Chr.3: 85912412-85912412 on Build GRCh38
5474650 c.640G>A p.A214T Chr.3: 85912504-85912504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775736759 Chr.3: 85912540-85912540 on Build GRCh38
rs766072450 Chr.3: 85912451-85912451 on Build GRCh38
rs755386010 Chr.3: 85912358-85912358 on Build GRCh38
rs772828446 Chr.3: 85912555-85912555 on Build GRCh38
rs771876014 Chr.3: 85912549-85912549 on Build GRCh38
rs759679851 Chr.3: 85912323-85912327 on Build GRCh38
rs1008680142 Chr.3: 85912642-85912642 on Build GRCh38
rs534833211 Chr.3: 85912633-85912633 on Build GRCh38
rs577958419 Chr.3: 85912762-85912762 on Build GRCh38
rs189737615 Chr.3: 85912427-85912427 on Build GRCh38
rs867682262 Chr.3: 85912470-85912470 on Build GRCh38
rs150681488 Chr.3: 85912442-85912442 on Build GRCh38
rs987266390 Chr.3: 85912323-85912323 on Build GRCh38
rs747412372 Chr.3: 85912521-85912521 on Build GRCh38
rs528505472 Chr.3: 85912609-85912609 on Build GRCh38
rs780344385 Chr.3: 85912436-85912436 on Build GRCh38
rs201652066 Chr.3: 85912492-85912492 on Build GRCh38
rs777931263 Chr.3: 85912515-85912515 on Build GRCh38
rs546980830 Chr.3: 85912627-85912627 on Build GRCh38
rs759648918 Chr.3: 85912405-85912405 on Build GRCh38
rs879162445 Chr.3: 85912562-85912562 on Build GRCh38
rs112272003 Chr.3: 85912600-85912600 on Build GRCh38
rs776515929 Chr.3: 85912401-85912401 on Build GRCh38
rs769973812 Chr.3: 85912537-85912537 on Build GRCh38
rs182526463 Chr.3: 85912654-85912654 on Build GRCh38
rs898808338 Chr.3: 85912598-85912598 on Build GRCh38
rs934666245 Chr.3: 85912469-85912469 on Build GRCh38
rs754449690 Chr.3: 85912472-85912472 on Build GRCh38
rs758932762 Chr.3: 85912505-85912505 on Build GRCh38
rs758844527 Chr.3: 85912593-85912593 on Build GRCh38
rs145952783 Chr.3: 85912469-85912470 on Build GRCh38
rs773587405 Chr.3: 85912392-85912392 on Build GRCh38
rs750266974 Chr.3: 85912446-85912446 on Build GRCh38
rs765481031 Chr.3: 85912419-85912419 on Build GRCh38
rs772143506 Chr.3: 85912554-85912554 on Build GRCh38
rs761893686 Chr.3: 85912431-85912431 on Build GRCh38
rs201403660 Chr.3: 85912582-85912582 on Build GRCh38
rs748691442 Chr.3: 85912373-85912373 on Build GRCh38
rs567814326 Chr.3: 85912606-85912606 on Build GRCh38
rs143748547 Chr.3: 85912561-85912561 on Build GRCh38
rs941279482 Chr.3: 85912430-85912430 on Build GRCh38
rs771188436 Chr.3: 85912399-85912399 on Build GRCh38
rs755659572 Chr.3: 85912450-85912450 on Build GRCh38
rs766123091 Chr.3: 85912568-85912568 on Build GRCh38
rs928012596 Chr.3: 85912666-85912666 on Build GRCh38
rs866759081 Chr.3: 85912615-85912615 on Build GRCh38
rs964975210 Chr.3: 85912346-85912346 on Build GRCh38
rs772400982 Chr.3: 85912387-85912387 on Build GRCh38
rs752206329 Chr.3: 85912586-85912586 on Build GRCh38
rs778440155 Chr.3: 85912484-85912484 on Build GRCh38
rs557483687 Chr.3: 85912738-85912738 on Build GRCh38
rs771257590 Chr.3: 85912522-85912522 on Build GRCh38
rs886991618 Chr.3: 85912422-85912422 on Build GRCh38
rs776908085 Chr.3: 85912523-85912523 on Build GRCh38
rs777769542 Chr.3: 85912655-85912655 on Build GRCh38
rs753503679 Chr.3: 85912579-85912579 on Build GRCh38
rs939163514 Chr.3: 85912318-85912318 on Build GRCh38
rs145436462 Chr.3: 85912491-85912491 on Build GRCh38
rs1025982909 Chr.3: 85912620-85912620 on Build GRCh38
rs147870348 Chr.3: 85912455-85912455 on Build GRCh38
rs779393808 Chr.3: 85912335-85912335 on Build GRCh38
rs557724747 Chr.3: 85912639-85912639 on Build GRCh38
rs890647605 Chr.3: 85912528-85912528 on Build GRCh38
rs749572236 Chr.3: 85912598-85912599 on Build GRCh38
rs950153269 Chr.3: 85912553-85912553 on Build GRCh38
rs375253682 Chr.3: 85912527-85912527 on Build GRCh38
rs910424229 Chr.3: 85912339-85912339 on Build GRCh38
rs374780088 Chr.3: 85912363-85912363 on Build GRCh38
rs1046967821 Chr.3: 85912356-85912356 on Build GRCh38
rs748838533 Chr.3: 85912340-85912340 on Build GRCh38
rs1029387859 Chr.3: 85912524-85912524 on Build GRCh38
rs917346005 Chr.3: 85912502-85912502 on Build GRCh38
rs979061989 Chr.3: 85912672-85912672 on Build GRCh38
rs565823521 Chr.3: 85912610-85912610 on Build GRCh38
rs113408089 Chr.3: 85912329-85912329 on Build GRCh38
rs747157258 Chr.3: 85912395-85912395 on Build GRCh38
rs761804143 Chr.3: 85912550-85912550 on Build GRCh38

View Full Product Details