Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 88055672-88056183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGGBP1
Entrez Gene ID
8545
Gene Name
CGG triplet repeat binding protein 1
Gene Aliases
CGGBP, p20-CGGBP
UniGene
Hs.444818
Gene Chromosome Location
Chr.3: 88051950-88149866 on Build GRCh38
Transcripts
NM_001008390.1 NM_001195308.1 NM_003663.3 XM_017007355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2828169 c.147T>G p.V49V Chr.3: 88055830-88055830 on Build GRCh38
1732286 c.116G>A p.G39E Chr.3: 88055861-88055861 on Build GRCh38
1200911 c.253C>T p.P85S Chr.3: 88055724-88055724 on Build GRCh38
3916724 c.148C>T p.L50L Chr.3: 88055829-88055829 on Build GRCh38
1732287 c.116G>A p.G39E Chr.3: 88055861-88055861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772202096 Chr.3: 88055996-88055996 on Build GRCh38
rs747262023 Chr.3: 88055866-88055866 on Build GRCh38
rs779164321 Chr.3: 88055969-88055969 on Build GRCh38
rs368186575 Chr.3: 88056077-88056077 on Build GRCh38
rs769657444 Chr.3: 88056009-88056009 on Build GRCh38
rs748293372 Chr.3: 88055749-88055749 on Build GRCh38
rs778934633 Chr.3: 88055745-88055745 on Build GRCh38
rs904972005 Chr.3: 88056128-88056128 on Build GRCh38
rs930245447 Chr.3: 88056053-88056053 on Build GRCh38
rs768362067 Chr.3: 88056028-88056028 on Build GRCh38
rs750695344 Chr.3: 88055717-88055717 on Build GRCh38
rs759189344 Chr.3: 88055813-88055813 on Build GRCh38
rs760128034 Chr.3: 88056005-88056006 on Build GRCh38
rs369645552 Chr.3: 88055984-88055984 on Build GRCh38
rs767844041 Chr.3: 88055875-88055875 on Build GRCh38
rs879077330 Chr.3: 88056124-88056124 on Build GRCh38
rs544742450 Chr.3: 88056082-88056082 on Build GRCh38
rs757831800 Chr.3: 88055710-88055710 on Build GRCh38
rs763902627 Chr.3: 88055854-88055854 on Build GRCh38
rs896144569 Chr.3: 88056083-88056083 on Build GRCh38
rs762659986 Chr.3: 88055853-88055853 on Build GRCh38
rs770764837 Chr.3: 88055775-88055775 on Build GRCh38
rs771293927 Chr.3: 88055985-88055988 on Build GRCh38
rs373418108 Chr.3: 88056014-88056014 on Build GRCh38
rs376176852 Chr.3: 88055917-88055917 on Build GRCh38
rs745843693 Chr.3: 88056001-88056001 on Build GRCh38
rs771064069 Chr.3: 88055902-88055902 on Build GRCh38
rs755189178 Chr.3: 88055938-88055938 on Build GRCh38
rs768917893 Chr.3: 88055731-88055731 on Build GRCh38
rs775418057 Chr.3: 88056010-88056010 on Build GRCh38
rs1001005464 Chr.3: 88056144-88056144 on Build GRCh38
rs781749484 Chr.3: 88055715-88055715 on Build GRCh38
rs775196022 Chr.3: 88055837-88055837 on Build GRCh38
rs776647535 Chr.3: 88055788-88055788 on Build GRCh38
rs763308303 Chr.3: 88055959-88055961 on Build GRCh38
rs556488291 Chr.3: 88056105-88056105 on Build GRCh38
rs765838209 Chr.3: 88056157-88056157 on Build GRCh38
rs752303792 Chr.3: 88056003-88056004 on Build GRCh38
rs956631930 Chr.3: 88055770-88055770 on Build GRCh38
rs750968952 Chr.3: 88055881-88055881 on Build GRCh38
rs561056802 Chr.3: 88056095-88056095 on Build GRCh38
rs1045121257 Chr.3: 88056116-88056118 on Build GRCh38
rs780954372 Chr.3: 88055999-88055999 on Build GRCh38
rs759946028 Chr.3: 88056155-88056155 on Build GRCh38
rs764053592 Chr.3: 88056018-88056019 on Build GRCh38
rs756504487 Chr.3: 88055719-88055719 on Build GRCh38
rs756382459 Chr.3: 88055910-88055910 on Build GRCh38
rs988034136 Chr.3: 88055809-88055809 on Build GRCh38
rs757944337 Chr.3: 88055701-88055701 on Build GRCh38
rs780098553 Chr.3: 88055724-88055724 on Build GRCh38
rs763673047 Chr.3: 88055702-88055702 on Build GRCh38
rs1047272783 Chr.3: 88056061-88056061 on Build GRCh38
rs769664578 Chr.3: 88055815-88055815 on Build GRCh38
rs780445875 Chr.3: 88055914-88055914 on Build GRCh38
rs920073309 Chr.3: 88056037-88056037 on Build GRCh38
rs751152101 Chr.3: 88055703-88055703 on Build GRCh38
rs3732776 Chr.3: 88056114-88056114 on Build GRCh38
rs955367173 Chr.3: 88055942-88055942 on Build GRCh38
rs373052025 Chr.3: 88055833-88055833 on Build GRCh38
rs766987404 Chr.3: 88056002-88056003 on Build GRCh38
rs773976003 Chr.3: 88056000-88056001 on Build GRCh38

View Full Product Details