Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 88157676-88158192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf38
Entrez Gene ID
285237
Gene Name
chromosome 3 open reading frame 38
Gene Aliases
-
UniGene
Hs.476944
Gene Chromosome Location
Chr.3: 88149713-88157965 on Build GRCh38
Transcripts
NM_173824.3 XM_005264745.4

SNP Detail

SNP ID Chromosome Location
rs1001542184 Chr.3: 88157884-88157884 on Build GRCh38
rs182500670 Chr.3: 88157784-88157784 on Build GRCh38
rs146271889 Chr.3: 88157702-88157702 on Build GRCh38
rs116049896 Chr.3: 88157903-88157903 on Build GRCh38
rs917768751 Chr.3: 88157791-88157791 on Build GRCh38
rs117216688 Chr.3: 88157757-88157757 on Build GRCh38
rs982970179 Chr.3: 88158062-88158062 on Build GRCh38
rs550174500 Chr.3: 88157894-88157894 on Build GRCh38
rs949237207 Chr.3: 88157792-88157792 on Build GRCh38
rs567283358 Chr.3: 88157782-88157782 on Build GRCh38
rs566580001 Chr.3: 88158115-88158116 on Build GRCh38
rs936548055 Chr.3: 88158129-88158129 on Build GRCh38
rs774133356 Chr.3: 88158135-88158135 on Build GRCh38
rs771154966 Chr.3: 88157819-88157819 on Build GRCh38
rs752813211 Chr.3: 88158069-88158069 on Build GRCh38
rs189638093 Chr.3: 88158085-88158085 on Build GRCh38
rs776111946 Chr.3: 88157992-88157992 on Build GRCh38
rs964232130 Chr.3: 88157994-88157994 on Build GRCh38
rs879662229 Chr.3: 88157975-88157975 on Build GRCh38
rs955763696 Chr.3: 88157977-88157977 on Build GRCh38
rs539649046 Chr.3: 88157988-88157988 on Build GRCh38
rs929700879 Chr.3: 88157764-88157764 on Build GRCh38
rs187156027 Chr.3: 88157864-88157864 on Build GRCh38
rs770153536 Chr.3: 88157978-88157978 on Build GRCh38
rs1053684593 Chr.3: 88158134-88158134 on Build GRCh38
rs905068460 Chr.3: 88157877-88157877 on Build GRCh38
rs1044869257 Chr.3: 88158084-88158084 on Build GRCh38
rs557391491 Chr.3: 88158007-88158007 on Build GRCh38
rs1046900924 Chr.3: 88157781-88157781 on Build GRCh38
rs763194112 Chr.3: 88157993-88157993 on Build GRCh38
rs926545305 Chr.3: 88158109-88158109 on Build GRCh38
rs536182921 Chr.3: 88157904-88157904 on Build GRCh38
rs560077016 Chr.3: 88157728-88157732 on Build GRCh38
rs547667726 Chr.3: 88157916-88157916 on Build GRCh38
rs563741056 Chr.3: 88157944-88157944 on Build GRCh38
rs949243171 Chr.3: 88158077-88158080 on Build GRCh38
rs936310619 Chr.3: 88158028-88158028 on Build GRCh38

View Full Product Details