Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 96814855-96815361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA6
Entrez Gene ID
285220
Gene Name
EPH receptor A6
Gene Aliases
EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066
UniGene
Hs.272208
Gene Chromosome Location
Chr.3: 96814581-97761532 on Build GRCh38
Transcripts
NM_001080448.2 NM_001278301.1 XM_006713592.3 XM_011512705.2 XM_011512706.2 XM_011512707.2 XM_017006210.1 XM_017006211.1 XM_017006214.1 XM_017006219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6836116 c.362G>A p.C121Y Chr.3: 96814985-96814985 on Build GRCh38
1176540 c.310C>A p.L104I Chr.3: 96814933-96814933 on Build GRCh38
6469149 c.86A>T p.H29L Chr.3: 96814991-96814991 on Build GRCh38
6836115 c.64G>A p.A22T Chr.3: 96814969-96814969 on Build GRCh38
480630 c.49T>A p.L17M Chr.3: 96814954-96814954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972467303 Chr.3: 96815109-96815109 on Build GRCh38
rs961730637 Chr.3: 96815068-96815068 on Build GRCh38
rs372676830 Chr.3: 96814877-96814877 on Build GRCh38
rs577710370 Chr.3: 96815245-96815245 on Build GRCh38
rs184935114 Chr.3: 96815024-96815024 on Build GRCh38
rs1009135247 Chr.3: 96815225-96815225 on Build GRCh38
rs1011554488 Chr.3: 96815258-96815258 on Build GRCh38
rs1022059815 Chr.3: 96815263-96815263 on Build GRCh38
rs772574696 Chr.3: 96814906-96814906 on Build GRCh38
rs1011425829 Chr.3: 96815184-96815184 on Build GRCh38
rs865808468 Chr.3: 96815053-96815053 on Build GRCh38
rs201007981 Chr.3: 96815279-96815279 on Build GRCh38
rs568474301 Chr.3: 96814992-96814992 on Build GRCh38
rs953632033 Chr.3: 96815017-96815017 on Build GRCh38
rs938375547 Chr.3: 96815107-96815107 on Build GRCh38
rs1019418550 Chr.3: 96815014-96815014 on Build GRCh38
rs763075281 Chr.3: 96814971-96814971 on Build GRCh38
rs550618554 Chr.3: 96815285-96815285 on Build GRCh38
rs540992359 Chr.3: 96815209-96815209 on Build GRCh38
rs746664331 Chr.3: 96815038-96815038 on Build GRCh38
rs757309852 Chr.3: 96815001-96815001 on Build GRCh38
rs972918232 Chr.3: 96815329-96815329 on Build GRCh38
rs761648098 Chr.3: 96814958-96814958 on Build GRCh38
rs113461882 Chr.3: 96814975-96814975 on Build GRCh38
rs539533620 Chr.3: 96815083-96815083 on Build GRCh38
rs906538457 Chr.3: 96815133-96815133 on Build GRCh38
rs537227700 Chr.3: 96814960-96814960 on Build GRCh38
rs555596612 Chr.3: 96815139-96815139 on Build GRCh38
rs961699745 Chr.3: 96815260-96815260 on Build GRCh38
rs1035987937 Chr.3: 96815317-96815317 on Build GRCh38
rs971805445 Chr.3: 96815068-96815074 on Build GRCh38
rs747749547 Chr.3: 96814898-96814898 on Build GRCh38
rs768379917 Chr.3: 96814884-96814884 on Build GRCh38
rs958437947 Chr.3: 96815000-96815000 on Build GRCh38
rs990585703 Chr.3: 96815073-96815073 on Build GRCh38
rs771813763 Chr.3: 96814904-96814904 on Build GRCh38
rs760135539 Chr.3: 96814941-96814941 on Build GRCh38
rs1052392693 Chr.3: 96815156-96815156 on Build GRCh38
rs960180197 Chr.3: 96815320-96815320 on Build GRCh38
rs563913260 Chr.3: 96815222-96815222 on Build GRCh38
rs543542086 Chr.3: 96815302-96815302 on Build GRCh38
rs867146210 Chr.3: 96814918-96814918 on Build GRCh38
rs765631591 Chr.3: 96814950-96814950 on Build GRCh38
rs938426577 Chr.3: 96814889-96814889 on Build GRCh38
rs901900822 Chr.3: 96815022-96815022 on Build GRCh38
rs535746701 Chr.3: 96815130-96815130 on Build GRCh38
rs570952271 Chr.3: 96815055-96815055 on Build GRCh38
rs1021860395 Chr.3: 96815088-96815088 on Build GRCh38
rs780924354 Chr.3: 96815046-96815046 on Build GRCh38
rs766018382 Chr.3: 96814910-96814910 on Build GRCh38
rs766617578 Chr.3: 96815177-96815177 on Build GRCh38
rs754972423 Chr.3: 96815027-96815027 on Build GRCh38
rs999349670 Chr.3: 96815181-96815181 on Build GRCh38
rs980355689 Chr.3: 96815298-96815298 on Build GRCh38
rs138975686 Chr.3: 96815112-96815112 on Build GRCh38
rs930974141 Chr.3: 96815119-96815119 on Build GRCh38
rs115065583 Chr.3: 96815030-96815030 on Build GRCh38
rs971690945 Chr.3: 96815262-96815262 on Build GRCh38
rs931005774 Chr.3: 96814927-96814927 on Build GRCh38
rs928798364 Chr.3: 96815106-96815106 on Build GRCh38
rs563479508 Chr.3: 96815327-96815327 on Build GRCh38
rs368151653 Chr.3: 96815031-96815031 on Build GRCh38
rs111306544 Chr.3: 96815092-96815092 on Build GRCh38
rs777228686 Chr.3: 96814917-96814917 on Build GRCh38
rs749242356 Chr.3: 96814878-96814878 on Build GRCh38
rs774133014 Chr.3: 96814893-96814893 on Build GRCh38
rs956058325 Chr.3: 96815032-96815032 on Build GRCh38
rs889680883 Chr.3: 96815149-96815149 on Build GRCh38
rs200313366 Chr.3: 96815002-96815002 on Build GRCh38
rs751748107 Chr.3: 96815277-96815277 on Build GRCh38
rs761881532 Chr.3: 96814889-96814889 on Build GRCh38
rs549254497 Chr.3: 96814895-96814895 on Build GRCh38
rs999544585 Chr.3: 96814993-96814993 on Build GRCh38
rs377032298 Chr.3: 96814997-96814997 on Build GRCh38
rs777442407 Chr.3: 96815037-96815037 on Build GRCh38
rs980238636 Chr.3: 96815094-96815094 on Build GRCh38
rs188841097 Chr.3: 96815208-96815208 on Build GRCh38
rs942628377 Chr.3: 96815161-96815161 on Build GRCh38
rs891900162 Chr.3: 96815248-96815248 on Build GRCh38
rs1040816628 Chr.3: 96815162-96815162 on Build GRCh38
rs935367772 Chr.3: 96815136-96815136 on Build GRCh38
rs762969527 Chr.3: 96815140-96815140 on Build GRCh38
rs1036667671 Chr.3: 96814891-96814891 on Build GRCh38
rs991663373 Chr.3: 96815321-96815322 on Build GRCh38
rs760271886 Chr.3: 96814909-96814909 on Build GRCh38

View Full Product Details