Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 96866453-96866962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA6
Entrez Gene ID
285220
Gene Name
EPH receptor A6
Gene Aliases
EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066
UniGene
Hs.272208
Gene Chromosome Location
Chr.3: 96814581-97761532 on Build GRCh38
Transcripts
NM_001080448.2 NM_001278301.1 XM_006713592.3 XM_011512705.2 XM_011512706.2 XM_011512707.2 XM_017006210.1 XM_017006211.1 XM_017006212.1 XM_017006214.1 XM_017006219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4924342 c.416G>A p.G139E Chr.3: 96866855-96866855 on Build GRCh38
4571134 c.105T>G p.V35V Chr.3: 96866826-96866826 on Build GRCh38
5530551 c.425G>A p.G142E Chr.3: 96866864-96866864 on Build GRCh38
5657876 c.107T>G p.V36G Chr.3: 96866828-96866828 on Build GRCh38
5811009 c.388G>C p.V130L Chr.3: 96866827-96866827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001009571 Chr.3: 96866557-96866557 on Build GRCh38
rs543157358 Chr.3: 96866748-96866748 on Build GRCh38
rs1011993560 Chr.3: 96866603-96866603 on Build GRCh38
rs917664284 Chr.3: 96866841-96866841 on Build GRCh38
rs775699553 Chr.3: 96866801-96866801 on Build GRCh38
rs1028501482 Chr.3: 96866711-96866712 on Build GRCh38
rs1012431627 Chr.3: 96866914-96866914 on Build GRCh38
rs35847126 Chr.3: 96866911-96866911 on Build GRCh38
rs372867987 Chr.3: 96866936-96866936 on Build GRCh38
rs919692134 Chr.3: 96866581-96866581 on Build GRCh38
rs376152081 Chr.3: 96866797-96866797 on Build GRCh38
rs973324623 Chr.3: 96866741-96866741 on Build GRCh38
rs761942565 Chr.3: 96866903-96866903 on Build GRCh38
rs565610344 Chr.3: 96866845-96866845 on Build GRCh38
rs376073893 Chr.3: 96866927-96866927 on Build GRCh38
rs955609330 Chr.3: 96866505-96866505 on Build GRCh38
rs61076237 Chr.3: 96866552-96866552 on Build GRCh38
rs961623641 Chr.3: 96866745-96866745 on Build GRCh38
rs1038029629 Chr.3: 96866566-96866566 on Build GRCh38
rs745907900 Chr.3: 96866855-96866855 on Build GRCh38
rs776287096 Chr.3: 96866777-96866777 on Build GRCh38
rs563651731 Chr.3: 96866638-96866638 on Build GRCh38
rs1054073059 Chr.3: 96866879-96866879 on Build GRCh38
rs902410110 Chr.3: 96866639-96866642 on Build GRCh38
rs757439554 Chr.3: 96866718-96866721 on Build GRCh38
rs777060332 Chr.3: 96866848-96866848 on Build GRCh38
rs955555943 Chr.3: 96866717-96866717 on Build GRCh38
rs1014821579 Chr.3: 96866655-96866655 on Build GRCh38
rs931856281 Chr.3: 96866794-96866794 on Build GRCh38
rs754887340 Chr.3: 96866923-96866923 on Build GRCh38
rs949266872 Chr.3: 96866911-96866911 on Build GRCh38
rs1046121116 Chr.3: 96866917-96866917 on Build GRCh38
rs987423240 Chr.3: 96866547-96866547 on Build GRCh38
rs535966907 Chr.3: 96866487-96866487 on Build GRCh38
rs142909067 Chr.3: 96866827-96866827 on Build GRCh38
rs921761982 Chr.3: 96866769-96866769 on Build GRCh38
rs963248873 Chr.3: 96866699-96866699 on Build GRCh38
rs1032983718 Chr.3: 96866687-96866687 on Build GRCh38
rs912645678 Chr.3: 96866813-96866813 on Build GRCh38
rs543871501 Chr.3: 96866621-96866621 on Build GRCh38
rs908819166 Chr.3: 96866745-96866747 on Build GRCh38
rs575689625 Chr.3: 96866874-96866874 on Build GRCh38
rs375514399 Chr.3: 96866606-96866606 on Build GRCh38
rs767578177 Chr.3: 96866912-96866912 on Build GRCh38
rs56339330 Chr.3: 96866821-96866821 on Build GRCh38
rs937740833 Chr.3: 96866592-96866592 on Build GRCh38
rs919555864 Chr.3: 96866792-96866792 on Build GRCh38
rs975591347 Chr.3: 96866492-96866492 on Build GRCh38
rs939707584 Chr.3: 96866554-96866554 on Build GRCh38
rs1042609837 Chr.3: 96866532-96866532 on Build GRCh38
rs750557948 Chr.3: 96866921-96866921 on Build GRCh38
rs574052999 Chr.3: 96866688-96866688 on Build GRCh38
rs957428775 Chr.3: 96866490-96866490 on Build GRCh38
rs558859606 Chr.3: 96866600-96866600 on Build GRCh38
rs538025193 Chr.3: 96866501-96866501 on Build GRCh38
rs983538168 Chr.3: 96866650-96866650 on Build GRCh38
rs760117332 Chr.3: 96866779-96866779 on Build GRCh38
rs986982365 Chr.3: 96866731-96866731 on Build GRCh38
rs1032491983 Chr.3: 96866579-96866579 on Build GRCh38
rs13315814 Chr.3: 96866599-96866599 on Build GRCh38

View Full Product Details