Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 97780274-97780779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL6
Entrez Gene ID
84100
Gene Name
ADP ribosylation factor like GTPase 6
Gene Aliases
BBS3, RP55
UniGene
Hs.373801
Gene Chromosome Location
Chr.3: 97762875-97812585 on Build GRCh38
Transcripts
NR_103511.1 XM_006713783.3 XM_017007311.1 XM_017007312.1 XM_017007313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1736357 c.189G>C p.L63F Chr.3: 97780618-97780618 on Build GRCh38
4150374 c.254+1G>T p.? Chr.3: 97780684-97780684 on Build GRCh38
731373 c.227A>G p.Y76C Chr.3: 97780656-97780656 on Build GRCh38
4569801 c.204T>C p.F68F Chr.3: 97780633-97780633 on Build GRCh38
1741631 c.221G>A p.G74E Chr.3: 97780650-97780650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773293069 Chr.3: 97780692-97780692 on Build GRCh38
rs973302448 Chr.3: 97780368-97780368 on Build GRCh38
rs780739267 Chr.3: 97780637-97780637 on Build GRCh38
rs140114162 Chr.3: 97780621-97780621 on Build GRCh38
rs953394619 Chr.3: 97780338-97780338 on Build GRCh38
rs554109839 Chr.3: 97780524-97780524 on Build GRCh38
rs752447497 Chr.3: 97780572-97780572 on Build GRCh38
rs550534162 Chr.3: 97780588-97780588 on Build GRCh38
rs933406429 Chr.3: 97780392-97780392 on Build GRCh38
rs937131629 Chr.3: 97780539-97780539 on Build GRCh38
rs568945242 Chr.3: 97780311-97780311 on Build GRCh38
rs77906443 Chr.3: 97780597-97780597 on Build GRCh38
rs893962096 Chr.3: 97780753-97780753 on Build GRCh38
rs879023102 Chr.3: 97780693-97780693 on Build GRCh38
rs770377692 Chr.3: 97780631-97780631 on Build GRCh38
rs1029024719 Chr.3: 97780315-97780315 on Build GRCh38
rs991353279 Chr.3: 97780586-97780586 on Build GRCh38
rs772021931 Chr.3: 97780691-97780691 on Build GRCh38
rs913261851 Chr.3: 97780432-97780432 on Build GRCh38
rs751279881 Chr.3: 97780601-97780601 on Build GRCh38
rs764012416 Chr.3: 97780732-97780732 on Build GRCh38
rs192414222 Chr.3: 97780502-97780502 on Build GRCh38
rs766922535 Chr.3: 97780726-97780728 on Build GRCh38
rs147929512 Chr.3: 97780488-97780488 on Build GRCh38
rs988788791 Chr.3: 97780403-97780403 on Build GRCh38
rs987851955 Chr.3: 97780372-97780372 on Build GRCh38
rs547786796 Chr.3: 97780588-97780590 on Build GRCh38
rs541850084 Chr.3: 97780674-97780674 on Build GRCh38
rs965278479 Chr.3: 97780299-97780299 on Build GRCh38
rs905694891 Chr.3: 97780491-97780491 on Build GRCh38
rs749599192 Chr.3: 97780655-97780655 on Build GRCh38
rs974851045 Chr.3: 97780300-97780300 on Build GRCh38
rs537864598 Chr.3: 97780465-97780465 on Build GRCh38
rs550048078 Chr.3: 97780728-97780728 on Build GRCh38
rs187991133 Chr.3: 97780492-97780492 on Build GRCh38
rs1025244924 Chr.3: 97780636-97780636 on Build GRCh38
rs1006463087 Chr.3: 97780738-97780738 on Build GRCh38
rs375431319 Chr.3: 97780679-97780679 on Build GRCh38
rs930584341 Chr.3: 97780486-97780486 on Build GRCh38
rs765090459 Chr.3: 97780700-97780700 on Build GRCh38
rs75485312 Chr.3: 97780305-97780305 on Build GRCh38
rs774422821 Chr.3: 97780664-97780664 on Build GRCh38
rs775416513 Chr.3: 97780713-97780713 on Build GRCh38
rs576180978 Chr.3: 97780662-97780662 on Build GRCh38
rs947275255 Chr.3: 97780440-97780440 on Build GRCh38
rs370656150 Chr.3: 97780604-97780604 on Build GRCh38
rs150667690 Chr.3: 97780626-97780626 on Build GRCh38
rs762719644 Chr.3: 97780721-97780721 on Build GRCh38
rs376047177 Chr.3: 97780733-97780733 on Build GRCh38
rs367959480 Chr.3: 97780695-97780695 on Build GRCh38
rs888140182 Chr.3: 97780678-97780678 on Build GRCh38
rs758191224 Chr.3: 97780592-97780592 on Build GRCh38
rs911981509 Chr.3: 97780396-97780396 on Build GRCh38

View Full Product Details