Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 97872969-97873480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBG3
Entrez Gene ID
131544
Gene Name
crystallin beta-gamma domain containing 3
Gene Aliases
DKFZp667G2110, vlAKAP
UniGene
Hs.733378
Gene Chromosome Location
Chr.3: 97821930-97944986 on Build GRCh38
Transcripts
NM_153605.3 XM_005247117.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5974929 c.1852A>G p.S618G Chr.3: 97873046-97873046 on Build GRCh38
4150375 c.2117A>G p.K706R Chr.3: 97873311-97873311 on Build GRCh38
5642257 c.1931G>T p.S644I Chr.3: 97873125-97873125 on Build GRCh38
5974931 c.2261A>G p.H754R Chr.3: 97873455-97873455 on Build GRCh38
5974930 c.2028T>G p.N676K Chr.3: 97873222-97873222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915831198 Chr.3: 97873451-97873451 on Build GRCh38
rs895626624 Chr.3: 97873117-97873117 on Build GRCh38
rs772117338 Chr.3: 97873262-97873262 on Build GRCh38
rs748407669 Chr.3: 97873209-97873209 on Build GRCh38
rs988921740 Chr.3: 97873343-97873343 on Build GRCh38
rs150876199 Chr.3: 97873164-97873164 on Build GRCh38
rs971603719 Chr.3: 97873455-97873455 on Build GRCh38
rs538259394 Chr.3: 97873166-97873166 on Build GRCh38
rs937218802 Chr.3: 97873081-97873081 on Build GRCh38
rs889245241 Chr.3: 97873342-97873344 on Build GRCh38
rs775300925 Chr.3: 97873316-97873316 on Build GRCh38
rs575186549 Chr.3: 97873344-97873344 on Build GRCh38
rs1054046094 Chr.3: 97873241-97873241 on Build GRCh38
rs934445140 Chr.3: 97873173-97873173 on Build GRCh38
rs747119358 Chr.3: 97873282-97873282 on Build GRCh38
rs920635580 Chr.3: 97872999-97872999 on Build GRCh38
rs567741110 Chr.3: 97873032-97873032 on Build GRCh38
rs954905141 Chr.3: 97873342-97873342 on Build GRCh38
rs536372300 Chr.3: 97873108-97873108 on Build GRCh38
rs1027642001 Chr.3: 97873430-97873430 on Build GRCh38
rs927244476 Chr.3: 97873379-97873379 on Build GRCh38
rs937290375 Chr.3: 97873385-97873385 on Build GRCh38
rs139338714 Chr.3: 97873434-97873434 on Build GRCh38
rs927269291 Chr.3: 97873071-97873071 on Build GRCh38
rs372658686 Chr.3: 97873046-97873046 on Build GRCh38
rs930529970 Chr.3: 97873033-97873033 on Build GRCh38
rs780457369 Chr.3: 97873015-97873015 on Build GRCh38
rs1005257389 Chr.3: 97873175-97873175 on Build GRCh38
rs1030471332 Chr.3: 97873289-97873289 on Build GRCh38
rs962120529 Chr.3: 97873372-97873372 on Build GRCh38
rs991431272 Chr.3: 97873043-97873043 on Build GRCh38
rs544067758 Chr.3: 97873346-97873346 on Build GRCh38
rs991378751 Chr.3: 97873444-97873444 on Build GRCh38
rs777780843 Chr.3: 97873280-97873280 on Build GRCh38
rs754524099 Chr.3: 97873359-97873359 on Build GRCh38
rs964993085 Chr.3: 97873265-97873265 on Build GRCh38
rs545280501 Chr.3: 97873152-97873152 on Build GRCh38
rs993607706 Chr.3: 97873382-97873382 on Build GRCh38
rs558086868 Chr.3: 97873317-97873317 on Build GRCh38
rs769701562 Chr.3: 97873304-97873304 on Build GRCh38
rs180695872 Chr.3: 97873146-97873146 on Build GRCh38
rs765645713 Chr.3: 97873102-97873102 on Build GRCh38
rs978790689 Chr.3: 97873366-97873366 on Build GRCh38
rs1050718616 Chr.3: 97873306-97873306 on Build GRCh38
rs551209800 Chr.3: 97873022-97873022 on Build GRCh38
rs554785329 Chr.3: 97873433-97873433 on Build GRCh38

View Full Product Details