Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 97886547-97887045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBG3
Entrez Gene ID
131544
Gene Name
crystallin beta-gamma domain containing 3
Gene Aliases
DKFZp667G2110, vlAKAP
UniGene
Hs.733378
Gene Chromosome Location
Chr.3: 97821930-97944986 on Build GRCh38
Transcripts
NM_153605.3 XM_005247117.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4328021 c.1348G>A p.D450N Chr.3: 97886670-97886670 on Build GRCh38
2949832 c.1316G>A p.S439N Chr.3: 97886638-97886638 on Build GRCh38
6883024 c.1310A>G p.D437G Chr.3: 97886632-97886632 on Build GRCh38
7257549 c.1313_1314delGC p.C438fs*1 Chr.3: 97886635-97886636 on Build GRCh38
6635878 c.1343A>G p.Q448R Chr.3: 97886665-97886665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999624332 Chr.3: 97886578-97886578 on Build GRCh38
rs74493950 Chr.3: 97886627-97886627 on Build GRCh38
rs745686570 Chr.3: 97886612-97886612 on Build GRCh38
rs900542342 Chr.3: 97886943-97886943 on Build GRCh38
rs947127048 Chr.3: 97886824-97886824 on Build GRCh38
rs775867105 Chr.3: 97886616-97886616 on Build GRCh38
rs780715374 Chr.3: 97886847-97886847 on Build GRCh38
rs191932500 Chr.3: 97886618-97886618 on Build GRCh38
rs752637385 Chr.3: 97886890-97886890 on Build GRCh38
rs758568827 Chr.3: 97886800-97886800 on Build GRCh38
rs768964100 Chr.3: 97886616-97886617 on Build GRCh38
rs754227089 Chr.3: 97886781-97886781 on Build GRCh38
rs752703868 Chr.3: 97886794-97886794 on Build GRCh38
rs1016744340 Chr.3: 97886931-97886931 on Build GRCh38
rs757082650 Chr.3: 97886694-97886694 on Build GRCh38
rs776344546 Chr.3: 97886617-97886618 on Build GRCh38
rs996575801 Chr.3: 97886955-97886955 on Build GRCh38
rs370924230 Chr.3: 97886756-97886756 on Build GRCh38
rs886487799 Chr.3: 97886699-97886699 on Build GRCh38
rs752822963 Chr.3: 97886690-97886690 on Build GRCh38
rs988654230 Chr.3: 97887017-97887017 on Build GRCh38
rs560032380 Chr.3: 97886659-97886659 on Build GRCh38
rs1030252738 Chr.3: 97886774-97886774 on Build GRCh38
rs774020761 Chr.3: 97886622-97886622 on Build GRCh38
rs745644960 Chr.3: 97886703-97886703 on Build GRCh38
rs146893133 Chr.3: 97886865-97886865 on Build GRCh38
rs796473332 Chr.3: 97886737-97886737 on Build GRCh38
rs751998952 Chr.3: 97886818-97886818 on Build GRCh38
rs543877399 Chr.3: 97886962-97886962 on Build GRCh38
rs1050475511 Chr.3: 97886934-97886934 on Build GRCh38
rs1016238826 Chr.3: 97886720-97886720 on Build GRCh38
rs772446465 Chr.3: 97886765-97886765 on Build GRCh38
rs1013698963 Chr.3: 97886667-97886667 on Build GRCh38
rs373991975 Chr.3: 97886695-97886695 on Build GRCh38
rs776219115 Chr.3: 97886608-97886608 on Build GRCh38
rs1052531948 Chr.3: 97886895-97886895 on Build GRCh38
rs1006337424 Chr.3: 97886917-97886917 on Build GRCh38
rs1003176095 Chr.3: 97886861-97886873 on Build GRCh38
rs777678638 Chr.3: 97886922-97886922 on Build GRCh38
rs537132161 Chr.3: 97886808-97886808 on Build GRCh38
rs375726435 Chr.3: 97886619-97886619 on Build GRCh38
rs965061016 Chr.3: 97886749-97886749 on Build GRCh38
rs831902 Chr.3: 97886833-97886833 on Build GRCh38
rs768454359 Chr.3: 97886725-97886725 on Build GRCh38
rs184136088 Chr.3: 97886969-97886969 on Build GRCh38
rs779830857 Chr.3: 97886713-97886713 on Build GRCh38
rs761402130 Chr.3: 97886747-97886747 on Build GRCh38
rs954585476 Chr.3: 97887008-97887008 on Build GRCh38
rs550330846 Chr.3: 97886823-97886823 on Build GRCh38
rs988601758 Chr.3: 97886802-97886802 on Build GRCh38
rs775348476 Chr.3: 97886616-97886616 on Build GRCh38
rs948359119 Chr.3: 97886845-97886845 on Build GRCh38
rs759684617 Chr.3: 97886788-97886788 on Build GRCh38
rs765987394 Chr.3: 97886772-97886772 on Build GRCh38
rs766586689 Chr.3: 97886662-97886662 on Build GRCh38
rs762187735 Chr.3: 97886635-97886635 on Build GRCh38
rs773668380 Chr.3: 97886728-97886728 on Build GRCh38
rs913001951 Chr.3: 97886818-97886818 on Build GRCh38
rs889001144 Chr.3: 97886935-97886935 on Build GRCh38
rs755031746 Chr.3: 97886681-97886681 on Build GRCh38
rs573210007 Chr.3: 97886868-97886868 on Build GRCh38
rs528708109 Chr.3: 97886691-97886691 on Build GRCh38
rs1047863252 Chr.3: 97886911-97886911 on Build GRCh38
rs139351624 Chr.3: 97886904-97886904 on Build GRCh38
rs748899301 Chr.3: 97886714-97886714 on Build GRCh38
rs188199462 Chr.3: 97886614-97886614 on Build GRCh38
rs552858798 Chr.3: 97886848-97886848 on Build GRCh38
rs768074277 Chr.3: 97886638-97886638 on Build GRCh38
rs751200308 Chr.3: 97886791-97886791 on Build GRCh38
rs558684507 Chr.3: 97886894-97886894 on Build GRCh38
rs571267735 Chr.3: 97886786-97886786 on Build GRCh38
rs566919193 Chr.3: 97886832-97886832 on Build GRCh38
rs542089352 Chr.3: 97886959-97886960 on Build GRCh38
rs371001126 Chr.3: 97886711-97886711 on Build GRCh38
rs770928933 Chr.3: 97886599-97886599 on Build GRCh38
rs778920415 Chr.3: 97886687-97886687 on Build GRCh38
rs760919914 Chr.3: 97886775-97886775 on Build GRCh38
rs77747100 Chr.3: 97886617-97886617 on Build GRCh38
rs761046610 Chr.3: 97886617-97886617 on Build GRCh38
rs781245403 Chr.3: 97886701-97886701 on Build GRCh38
rs574075256 Chr.3: 97886978-97886978 on Build GRCh38
rs755988239 Chr.3: 97886913-97886913 on Build GRCh38
rs370599431 Chr.3: 97886666-97886666 on Build GRCh38
rs766782973 Chr.3: 97886778-97886778 on Build GRCh38
rs750697238 Chr.3: 97886655-97886655 on Build GRCh38
rs779953878 Chr.3: 97886811-97886811 on Build GRCh38

View Full Product Details