Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98588622-98589025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPOX
Entrez Gene ID
1371
Gene Name
coproporphyrinogen oxidase
Gene Aliases
CPO, CPX, HCP
UniGene
Hs.476982
Gene Chromosome Location
Chr.3: 98568448-98593684 on Build GRCh38
Transcripts
NM_000097.5 XM_005247125.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3598402 c.905A>G p.E302G Chr.3: 98588761-98588761 on Build GRCh38
4674420 c.824G>T p.W275L Chr.3: 98588842-98588842 on Build GRCh38
6098298 c.945T>G p.F315L Chr.3: 98588721-98588721 on Build GRCh38
4002855 c.880G>T p.V294F Chr.3: 98588786-98588786 on Build GRCh38
3760286 c.880G>A p.V294I Chr.3: 98588786-98588786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761281919 Chr.3: 98588839-98588839 on Build GRCh38
rs1024614583 Chr.3: 98588713-98588713 on Build GRCh38
rs1032748531 Chr.3: 98588871-98588871 on Build GRCh38
rs544238353 Chr.3: 98588992-98588992 on Build GRCh38
rs747040461 Chr.3: 98588883-98588883 on Build GRCh38
rs573797955 Chr.3: 98588821-98588821 on Build GRCh38
rs372047148 Chr.3: 98588749-98588749 on Build GRCh38
rs917176443 Chr.3: 98588990-98588990 on Build GRCh38
rs991091530 Chr.3: 98588885-98588885 on Build GRCh38
rs936496857 Chr.3: 98588800-98588800 on Build GRCh38
rs121917870 Chr.3: 98588783-98588783 on Build GRCh38
rs140052404 Chr.3: 98588769-98588769 on Build GRCh38
rs758690019 Chr.3: 98588872-98588872 on Build GRCh38
rs121917874 Chr.3: 98588831-98588831 on Build GRCh38
rs762827865 Chr.3: 98588789-98588789 on Build GRCh38
rs772225283 Chr.3: 98588748-98588748 on Build GRCh38
rs748245146 Chr.3: 98588730-98588730 on Build GRCh38
rs755085431 Chr.3: 98588756-98588756 on Build GRCh38
rs940889396 Chr.3: 98588969-98588969 on Build GRCh38
rs779295994 Chr.3: 98588854-98588854 on Build GRCh38
rs1051746625 Chr.3: 98588956-98588956 on Build GRCh38
rs753030473 Chr.3: 98588862-98588862 on Build GRCh38
rs756562257 Chr.3: 98588689-98588689 on Build GRCh38
rs773924747 Chr.3: 98588805-98588805 on Build GRCh38
rs766771353 Chr.3: 98588844-98588844 on Build GRCh38
rs767913223 Chr.3: 98588670-98588670 on Build GRCh38
rs773155019 Chr.3: 98588758-98588758 on Build GRCh38
rs183898870 Chr.3: 98588941-98588941 on Build GRCh38
rs77651204 Chr.3: 98588656-98588656 on Build GRCh38
rs756715937 Chr.3: 98588665-98588665 on Build GRCh38
rs755296583 Chr.3: 98588705-98588705 on Build GRCh38
rs557074694 Chr.3: 98588804-98588804 on Build GRCh38
rs189121366 Chr.3: 98588975-98588975 on Build GRCh38
rs780531734 Chr.3: 98588693-98588693 on Build GRCh38
rs140475004 Chr.3: 98588788-98588788 on Build GRCh38
rs777948742 Chr.3: 98588882-98588882 on Build GRCh38
rs769794490 Chr.3: 98588905-98588905 on Build GRCh38
rs542726479 Chr.3: 98588849-98588849 on Build GRCh38
rs545312520 Chr.3: 98588955-98588955 on Build GRCh38
rs372927967 Chr.3: 98588790-98588790 on Build GRCh38
rs941482094 Chr.3: 98588649-98588649 on Build GRCh38
rs373085466 Chr.3: 98588710-98588710 on Build GRCh38
rs886452836 Chr.3: 98588969-98588969 on Build GRCh38
rs750882934 Chr.3: 98588682-98588682 on Build GRCh38
rs933417800 Chr.3: 98588951-98588951 on Build GRCh38
rs565173214 Chr.3: 98588970-98588970 on Build GRCh38
rs761326041 Chr.3: 98588811-98588811 on Build GRCh38
rs1131857 Chr.3: 98588852-98588852 on Build GRCh38
rs1038446540 Chr.3: 98588773-98588773 on Build GRCh38
rs777386286 Chr.3: 98588977-98588977 on Build GRCh38
rs2228056 Chr.3: 98588786-98588786 on Build GRCh38
rs1038363210 Chr.3: 98588980-98588980 on Build GRCh38
rs758030024 Chr.3: 98588933-98588933 on Build GRCh38
rs749892280 Chr.3: 98588827-98588827 on Build GRCh38
rs749582392 Chr.3: 98588696-98588696 on Build GRCh38
rs921999005 Chr.3: 98588912-98588913 on Build GRCh38

View Full Product Details