Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98590879-98591402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPOX
Entrez Gene ID
1371
Gene Name
coproporphyrinogen oxidase
Gene Aliases
CPO, CPX, HCP
UniGene
Hs.476982
Gene Chromosome Location
Chr.3: 98568448-98593684 on Build GRCh38
Transcripts
NM_000097.5 XM_005247125.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
732202 c.697G>T p.D233Y Chr.3: 98591015-98591015 on Build GRCh38
6634745 c.616A>C p.S206R Chr.3: 98591096-98591096 on Build GRCh38
7250832 c.640C>A p.L214I Chr.3: 98591072-98591072 on Build GRCh38
115674 c.677G>T p.G226V Chr.3: 98591035-98591035 on Build GRCh38
4674421 c.672C>T p.S224S Chr.3: 98591040-98591040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902400767 Chr.3: 98590903-98590903 on Build GRCh38
rs552752095 Chr.3: 98591080-98591080 on Build GRCh38
rs777472751 Chr.3: 98591049-98591049 on Build GRCh38
rs533876699 Chr.3: 98591333-98591333 on Build GRCh38
rs149638048 Chr.3: 98591371-98591371 on Build GRCh38
rs1729978 Chr.3: 98591346-98591346 on Build GRCh38
rs762184805 Chr.3: 98591095-98591095 on Build GRCh38
rs760260164 Chr.3: 98591322-98591322 on Build GRCh38
rs953564251 Chr.3: 98591157-98591157 on Build GRCh38
rs972233462 Chr.3: 98591288-98591288 on Build GRCh38
rs772309560 Chr.3: 98591099-98591099 on Build GRCh38
rs556309775 Chr.3: 98591175-98591175 on Build GRCh38
rs1045141887 Chr.3: 98591312-98591312 on Build GRCh38
rs750156215 Chr.3: 98591158-98591158 on Build GRCh38
rs777025757 Chr.3: 98591283-98591287 on Build GRCh38
rs149384011 Chr.3: 98591100-98591100 on Build GRCh38
rs752636733 Chr.3: 98591046-98591046 on Build GRCh38
rs763754912 Chr.3: 98591362-98591362 on Build GRCh38
rs749826763 Chr.3: 98591081-98591081 on Build GRCh38
rs999375185 Chr.3: 98590913-98590913 on Build GRCh38
rs1040699841 Chr.3: 98591219-98591219 on Build GRCh38
rs766515451 Chr.3: 98591112-98591112 on Build GRCh38
rs753927678 Chr.3: 98591114-98591114 on Build GRCh38
rs774623952 Chr.3: 98591084-98591084 on Build GRCh38
rs781006907 Chr.3: 98591148-98591148 on Build GRCh38
rs35909472 Chr.3: 98591241-98591241 on Build GRCh38
rs568211802 Chr.3: 98591320-98591320 on Build GRCh38
rs763388089 Chr.3: 98590976-98590976 on Build GRCh38
rs906597094 Chr.3: 98591318-98591318 on Build GRCh38
rs554081993 Chr.3: 98591356-98591356 on Build GRCh38
rs138479596 Chr.3: 98591061-98591061 on Build GRCh38
rs986269338 Chr.3: 98591359-98591359 on Build GRCh38
rs751343617 Chr.3: 98591137-98591137 on Build GRCh38
rs1025941294 Chr.3: 98591317-98591317 on Build GRCh38
rs1012013845 Chr.3: 98591311-98591311 on Build GRCh38
rs764422808 Chr.3: 98590980-98590980 on Build GRCh38
rs965086306 Chr.3: 98591374-98591374 on Build GRCh38
rs962573015 Chr.3: 98591097-98591097 on Build GRCh38
rs370219453 Chr.3: 98591181-98591181 on Build GRCh38
rs199774737 Chr.3: 98591027-98591027 on Build GRCh38
rs377407802 Chr.3: 98590966-98590966 on Build GRCh38
rs146049447 Chr.3: 98591068-98591068 on Build GRCh38
rs1053993506 Chr.3: 98590920-98590920 on Build GRCh38
rs121917872 Chr.3: 98591089-98591089 on Build GRCh38
rs775612289 Chr.3: 98591285-98591285 on Build GRCh38
rs748458175 Chr.3: 98590910-98590910 on Build GRCh38
rs759347283 Chr.3: 98591147-98591147 on Build GRCh38
rs747337203 Chr.3: 98590983-98590983 on Build GRCh38
rs774772873 Chr.3: 98590983-98590983 on Build GRCh38
rs746798334 Chr.3: 98591051-98591051 on Build GRCh38
rs1008380716 Chr.3: 98591207-98591207 on Build GRCh38
rs765176336 Chr.3: 98591030-98591030 on Build GRCh38
rs1053703616 Chr.3: 98591277-98591277 on Build GRCh38
rs548706583 Chr.3: 98590909-98590909 on Build GRCh38
rs201663136 Chr.3: 98591000-98591000 on Build GRCh38
rs761981172 Chr.3: 98590987-98590987 on Build GRCh38
rs753484065 Chr.3: 98591350-98591350 on Build GRCh38
rs934761324 Chr.3: 98591014-98591014 on Build GRCh38
rs567313907 Chr.3: 98590914-98590914 on Build GRCh38
rs748809525 Chr.3: 98591179-98591179 on Build GRCh38
rs373637695 Chr.3: 98591186-98591186 on Build GRCh38
rs748513805 Chr.3: 98591195-98591195 on Build GRCh38
rs748220649 Chr.3: 98590962-98590963 on Build GRCh38
rs778423283 Chr.3: 98591145-98591147 on Build GRCh38
rs779469661 Chr.3: 98591178-98591178 on Build GRCh38
rs897821860 Chr.3: 98591326-98591326 on Build GRCh38
rs373551666 Chr.3: 98591163-98591163 on Build GRCh38
rs756992469 Chr.3: 98591054-98591054 on Build GRCh38
rs910950366 Chr.3: 98591183-98591183 on Build GRCh38
rs548088221 Chr.3: 98591242-98591242 on Build GRCh38
rs758356130 Chr.3: 98591048-98591048 on Build GRCh38
rs113195017 Chr.3: 98591025-98591025 on Build GRCh38
rs756305768 Chr.3: 98590980-98590981 on Build GRCh38

View Full Product Details