Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101724565-101725060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP97
Entrez Gene ID
79598
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, LRRIQ2
UniGene
Hs.444135
Gene Chromosome Location
Chr.3: 101724098-101770562 on Build GRCh38
Transcripts
NM_001303401.1 NM_024548.3 XM_006713743.3 XM_011513127.1 XM_017007178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3584666 c.38G>A p.G13E Chr.3: 101724714-101724714 on Build GRCh38
3584667 c.38G>A p.G13E Chr.3: 101724714-101724714 on Build GRCh38
6095443 c.36C>T p.P12P Chr.3: 101724712-101724712 on Build GRCh38
5863116 c.31C>T p.P11S Chr.3: 101724707-101724707 on Build GRCh38
1035828 c.40G>A p.E14K Chr.3: 101724716-101724716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040201547 Chr.3: 101724624-101724624 on Build GRCh38
rs549591031 Chr.3: 101724983-101724983 on Build GRCh38
rs748809597 Chr.3: 101724714-101724714 on Build GRCh38
rs544147174 Chr.3: 101724899-101724899 on Build GRCh38
rs769300227 Chr.3: 101724645-101724645 on Build GRCh38
rs924754267 Chr.3: 101724608-101724608 on Build GRCh38
rs181297781 Chr.3: 101724850-101724850 on Build GRCh38
rs6790109 Chr.3: 101724656-101724656 on Build GRCh38
rs757072345 Chr.3: 101724694-101724694 on Build GRCh38
rs115499192 Chr.3: 101724613-101724613 on Build GRCh38
rs1056884839 Chr.3: 101724976-101724976 on Build GRCh38
rs377040183 Chr.3: 101724730-101724730 on Build GRCh38
rs573895244 Chr.3: 101724888-101724888 on Build GRCh38
rs1048766027 Chr.3: 101724965-101724965 on Build GRCh38
rs572567778 Chr.3: 101724777-101724777 on Build GRCh38
rs567726737 Chr.3: 101725012-101725012 on Build GRCh38
rs979409201 Chr.3: 101724834-101724834 on Build GRCh38
rs113730099 Chr.3: 101724651-101724651 on Build GRCh38
rs578050989 Chr.3: 101724649-101724649 on Build GRCh38
rs377554540 Chr.3: 101724761-101724761 on Build GRCh38
rs934871801 Chr.3: 101724609-101724609 on Build GRCh38
rs73863393 Chr.3: 101724847-101724847 on Build GRCh38
rs958892813 Chr.3: 101724791-101724791 on Build GRCh38
rs1035665271 Chr.3: 101724861-101724861 on Build GRCh38
rs773569838 Chr.3: 101724667-101724667 on Build GRCh38
rs1034472140 Chr.3: 101724993-101724993 on Build GRCh38
rs745540482 Chr.3: 101724989-101724989 on Build GRCh38
rs777781069 Chr.3: 101724833-101724833 on Build GRCh38
rs753184901 Chr.3: 101724845-101724845 on Build GRCh38
rs968769796 Chr.3: 101724785-101724785 on Build GRCh38
rs753924919 Chr.3: 101724687-101724687 on Build GRCh38
rs527376529 Chr.3: 101724977-101724977 on Build GRCh38
rs764007295 Chr.3: 101724768-101724768 on Build GRCh38
rs201771736 Chr.3: 101724690-101724690 on Build GRCh38
rs780812135 Chr.3: 101724639-101724639 on Build GRCh38
rs956307732 Chr.3: 101724882-101724882 on Build GRCh38
rs780014804 Chr.3: 101724820-101724820 on Build GRCh38
rs778402282 Chr.3: 101724883-101724883 on Build GRCh38
rs191271728 Chr.3: 101724759-101724759 on Build GRCh38
rs745451146 Chr.3: 101724641-101724641 on Build GRCh38
rs761345831 Chr.3: 101724639-101724641 on Build GRCh38
rs921680182 Chr.3: 101724927-101724927 on Build GRCh38
rs772575367 Chr.3: 101724752-101724752 on Build GRCh38
rs944356682 Chr.3: 101724893-101724893 on Build GRCh38
rs769175929 Chr.3: 101724650-101724650 on Build GRCh38
rs370313105 Chr.3: 101724829-101724829 on Build GRCh38
rs758311009 Chr.3: 101724689-101724689 on Build GRCh38
rs373337025 Chr.3: 101724704-101724704 on Build GRCh38
rs1012988556 Chr.3: 101724776-101724776 on Build GRCh38
rs368851013 Chr.3: 101724736-101724736 on Build GRCh38
rs748534690 Chr.3: 101724744-101724744 on Build GRCh38
rs375142336 Chr.3: 101724635-101724635 on Build GRCh38
rs979073820 Chr.3: 101724583-101724583 on Build GRCh38
rs1034418154 Chr.3: 101724781-101724781 on Build GRCh38
rs377037537 Chr.3: 101724680-101724680 on Build GRCh38
rs769100893 Chr.3: 101724726-101724726 on Build GRCh38
rs755715767 Chr.3: 101724706-101724706 on Build GRCh38
rs768028749 Chr.3: 101724746-101724746 on Build GRCh38
rs779572200 Chr.3: 101724710-101724710 on Build GRCh38
rs886913005 Chr.3: 101724719-101724719 on Build GRCh38
rs559863836 Chr.3: 101724968-101724968 on Build GRCh38
rs774925234 Chr.3: 101724648-101724648 on Build GRCh38
rs1010543606 Chr.3: 101724975-101724975 on Build GRCh38
rs111828531 Chr.3: 101724653-101724653 on Build GRCh38
rs369522245 Chr.3: 101724612-101724612 on Build GRCh38
rs758148380 Chr.3: 101724992-101724992 on Build GRCh38
rs1022918710 Chr.3: 101724782-101724782 on Build GRCh38
rs756803112 Chr.3: 101724905-101724905 on Build GRCh38
rs201090433 Chr.3: 101724684-101724684 on Build GRCh38
rs773659559 Chr.3: 101724755-101724755 on Build GRCh38
rs995953225 Chr.3: 101724637-101724638 on Build GRCh38
rs112269712 Chr.3: 101724952-101724952 on Build GRCh38
rs370058080 Chr.3: 101724697-101724697 on Build GRCh38
rs562145707 Chr.3: 101724909-101724909 on Build GRCh38
rs550951357 Chr.3: 101724960-101724960 on Build GRCh38
rs759651144 Chr.3: 101724766-101724766 on Build GRCh38
rs777464678 Chr.3: 101724626-101724626 on Build GRCh38
rs931008202 Chr.3: 101724961-101724961 on Build GRCh38
rs943768075 Chr.3: 101724889-101724889 on Build GRCh38
rs995679920 Chr.3: 101725022-101725022 on Build GRCh38
rs994573 Chr.3: 101724617-101724617 on Build GRCh38
rs939815454 Chr.3: 101724974-101724974 on Build GRCh38
rs958736704 Chr.3: 101725003-101725003 on Build GRCh38
rs746792187 Chr.3: 101724633-101724633 on Build GRCh38
rs931206640 Chr.3: 101724664-101724664 on Build GRCh38

View Full Product Details