Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101779751-101780246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE3
Entrez Gene ID
91775
Gene Name
neurexophilin and PC-esterase domain family member 3
Gene Aliases
FAM55C, MST115, MSTP115
UniGene
Hs.595933
Gene Chromosome Location
Chr.3: 101779185-101828231 on Build GRCh38
Transcripts
NM_001134456.1 NM_145037.2

SNP Detail

SNP ID Chromosome Location
rs896665199 Chr.3: 101779870-101779870 on Build GRCh38
rs200262284 Chr.3: 101780074-101780074 on Build GRCh38
rs553830097 Chr.3: 101779864-101779864 on Build GRCh38
rs903751053 Chr.3: 101779818-101779818 on Build GRCh38
rs1013755632 Chr.3: 101779874-101779874 on Build GRCh38
rs1043397313 Chr.3: 101780185-101780185 on Build GRCh38
rs1017029580 Chr.3: 101780073-101780073 on Build GRCh38
rs80031994 Chr.3: 101780124-101780124 on Build GRCh38
rs567243873 Chr.3: 101780169-101780169 on Build GRCh38
rs935422855 Chr.3: 101779845-101779845 on Build GRCh38
rs537890308 Chr.3: 101780204-101780204 on Build GRCh38
rs955247436 Chr.3: 101780140-101780140 on Build GRCh38
rs781248426 Chr.3: 101780113-101780113 on Build GRCh38
rs545306530 Chr.3: 101779819-101779819 on Build GRCh38
rs116001607 Chr.3: 101779879-101779879 on Build GRCh38
rs1044104925 Chr.3: 101779776-101779776 on Build GRCh38
rs1043811912 Chr.3: 101779798-101779798 on Build GRCh38
rs769266215 Chr.3: 101780172-101780172 on Build GRCh38
rs758791580 Chr.3: 101779862-101779862 on Build GRCh38
rs867107446 Chr.3: 101779861-101779861 on Build GRCh38
rs1006443362 Chr.3: 101780038-101780038 on Build GRCh38
rs894789805 Chr.3: 101779769-101779769 on Build GRCh38
rs999889987 Chr.3: 101779854-101779854 on Build GRCh38
rs974091090 Chr.3: 101780007-101780007 on Build GRCh38
rs190227829 Chr.3: 101780139-101780139 on Build GRCh38
rs548839238 Chr.3: 101779784-101779784 on Build GRCh38
rs1011881094 Chr.3: 101779795-101779795 on Build GRCh38
rs938870523 Chr.3: 101780136-101780136 on Build GRCh38
rs928766304 Chr.3: 101780125-101780125 on Build GRCh38
rs964014070 Chr.3: 101779991-101779991 on Build GRCh38
rs1018391607 Chr.3: 101779965-101779965 on Build GRCh38
rs1027438154 Chr.3: 101780070-101780070 on Build GRCh38

View Full Product Details