Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101782573-101783041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE3
Entrez Gene ID
91775
Gene Name
neurexophilin and PC-esterase domain family member 3
Gene Aliases
FAM55C, MST115, MSTP115
UniGene
Hs.595933
Gene Chromosome Location
Chr.3: 101779185-101828231 on Build GRCh38
Transcripts
NM_001134456.1 NM_145037.2

SNP Detail

SNP ID Chromosome Location
rs932254488 Chr.3: 101782828-101782828 on Build GRCh38
rs937884274 Chr.3: 101782763-101782763 on Build GRCh38
rs896826399 Chr.3: 101782644-101782644 on Build GRCh38
rs540710616 Chr.3: 101782848-101782848 on Build GRCh38
rs996246632 Chr.3: 101782826-101782826 on Build GRCh38
rs567205603 Chr.3: 101782912-101782912 on Build GRCh38
rs773247670 Chr.3: 101782993-101782993 on Build GRCh38
rs920402274 Chr.3: 101782784-101782784 on Build GRCh38
rs989038934 Chr.3: 101782630-101782634 on Build GRCh38
rs896804975 Chr.3: 101782903-101782903 on Build GRCh38
rs549059793 Chr.3: 101782765-101782765 on Build GRCh38
rs187322332 Chr.3: 101782658-101782658 on Build GRCh38
rs575710196 Chr.3: 101782817-101782817 on Build GRCh38
rs1036746795 Chr.3: 101782877-101782877 on Build GRCh38
rs775230984 Chr.3: 101782857-101782857 on Build GRCh38
rs574972771 Chr.3: 101782759-101782759 on Build GRCh38
rs1010500597 Chr.3: 101782883-101782883 on Build GRCh38
rs1020635443 Chr.3: 101782643-101782643 on Build GRCh38
rs192933496 Chr.3: 101782906-101782906 on Build GRCh38
rs957475528 Chr.3: 101782791-101782791 on Build GRCh38
rs905773643 Chr.3: 101782919-101782919 on Build GRCh38
rs913202884 Chr.3: 101782816-101782816 on Build GRCh38
rs113343197 Chr.3: 101782907-101782907 on Build GRCh38
rs1020542058 Chr.3: 101782895-101782895 on Build GRCh38
rs1033293326 Chr.3: 101782780-101782780 on Build GRCh38
rs576978402 Chr.3: 101782927-101782927 on Build GRCh38
rs2466367 Chr.3: 101782983-101782983 on Build GRCh38
rs778649548 Chr.3: 101782733-101782733 on Build GRCh38
rs192218364 Chr.3: 101782794-101782794 on Build GRCh38
rs554397500 Chr.3: 101782649-101782649 on Build GRCh38
rs114177307 Chr.3: 101782820-101782820 on Build GRCh38
rs1001363940 Chr.3: 101782737-101782737 on Build GRCh38
rs527534429 Chr.3: 101782760-101782760 on Build GRCh38
rs183979293 Chr.3: 101782815-101782815 on Build GRCh38
rs72937817 Chr.3: 101783000-101783000 on Build GRCh38
rs756947530 Chr.3: 101782723-101782723 on Build GRCh38
rs189513612 Chr.3: 101782958-101782958 on Build GRCh38
rs144005260 Chr.3: 101782822-101782822 on Build GRCh38
rs991048963 Chr.3: 101782767-101782767 on Build GRCh38
rs771901743 Chr.3: 101782838-101782838 on Build GRCh38
rs970404588 Chr.3: 101782685-101782685 on Build GRCh38
rs1020398307 Chr.3: 101783004-101783004 on Build GRCh38
rs377526128 Chr.3: 101782785-101782785 on Build GRCh38
rs927702095 Chr.3: 101782717-101782717 on Build GRCh38
rs188144678 Chr.3: 101782840-101782840 on Build GRCh38
rs893269956 Chr.3: 101782852-101782852 on Build GRCh38

View Full Product Details