Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101801568-101802074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE3
Entrez Gene ID
91775
Gene Name
neurexophilin and PC-esterase domain family member 3
Gene Aliases
FAM55C, MST115, MSTP115
UniGene
Hs.595933
Gene Chromosome Location
Chr.3: 101779185-101828231 on Build GRCh38
Transcripts
NM_001134456.1 NM_145037.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5863124 c.773G>A p.S258N Chr.3: 101801914-101801914 on Build GRCh38
5863123 c.568C>T p.P190S Chr.3: 101801709-101801709 on Build GRCh38
4495316 c.450C>T p.S150S Chr.3: 101801591-101801591 on Build GRCh38
1035852 c.487G>A p.D163N Chr.3: 101801628-101801628 on Build GRCh38
4683086 c.721C>A p.L241I Chr.3: 101801862-101801862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038350374 Chr.3: 101801682-101801682 on Build GRCh38
rs771052889 Chr.3: 101801764-101801764 on Build GRCh38
rs148108023 Chr.3: 101801925-101801925 on Build GRCh38
rs146775844 Chr.3: 101801783-101801783 on Build GRCh38
rs764131420 Chr.3: 101801661-101801661 on Build GRCh38
rs950049078 Chr.3: 101801602-101801602 on Build GRCh38
rs765677856 Chr.3: 101801922-101801922 on Build GRCh38
rs771097346 Chr.3: 101802007-101802007 on Build GRCh38
rs567933453 Chr.3: 101801834-101801834 on Build GRCh38
rs773968419 Chr.3: 101801623-101801623 on Build GRCh38
rs751579719 Chr.3: 101801776-101801776 on Build GRCh38
rs770675524 Chr.3: 101801861-101801861 on Build GRCh38
rs759701678 Chr.3: 101801771-101801771 on Build GRCh38
rs573446827 Chr.3: 101801814-101801814 on Build GRCh38
rs776844767 Chr.3: 101801770-101801770 on Build GRCh38
rs1053560430 Chr.3: 101801992-101801992 on Build GRCh38
rs140500702 Chr.3: 101801723-101801723 on Build GRCh38
rs769814229 Chr.3: 101801869-101801869 on Build GRCh38
rs909642388 Chr.3: 101801972-101801972 on Build GRCh38
rs865798800 Chr.3: 101801926-101801926 on Build GRCh38
rs766669488 Chr.3: 101801911-101801911 on Build GRCh38
rs746081450 Chr.3: 101801785-101801785 on Build GRCh38
rs781017668 Chr.3: 101801632-101801632 on Build GRCh38
rs908927032 Chr.3: 101801830-101801830 on Build GRCh38
rs549028239 Chr.3: 101801805-101801805 on Build GRCh38
rs755702543 Chr.3: 101801704-101801704 on Build GRCh38
rs1045669817 Chr.3: 101801605-101801605 on Build GRCh38
rs370550526 Chr.3: 101801749-101801749 on Build GRCh38
rs773405177 Chr.3: 101801902-101801902 on Build GRCh38
rs140976351 Chr.3: 101801944-101801944 on Build GRCh38
rs201844634 Chr.3: 101801963-101801963 on Build GRCh38
rs952851561 Chr.3: 101801819-101801819 on Build GRCh38
rs78410316 Chr.3: 101801951-101801951 on Build GRCh38
rs776855499 Chr.3: 101802010-101802010 on Build GRCh38
rs759173578 Chr.3: 101801751-101801755 on Build GRCh38
rs755360990 Chr.3: 101801919-101801919 on Build GRCh38
rs775499698 Chr.3: 101801970-101801970 on Build GRCh38
rs765583850 Chr.3: 101802035-101802035 on Build GRCh38
rs2305990 Chr.3: 101801693-101801693 on Build GRCh38
rs143729248 Chr.3: 101801737-101801737 on Build GRCh38
rs373676788 Chr.3: 101801800-101801800 on Build GRCh38
rs863223380 Chr.3: 101801593-101801600 on Build GRCh38
rs200918733 Chr.3: 101801745-101801745 on Build GRCh38
rs775589660 Chr.3: 101801885-101801885 on Build GRCh38
rs763347741 Chr.3: 101802041-101802041 on Build GRCh38
rs745697319 Chr.3: 101801868-101801868 on Build GRCh38
rs753119978 Chr.3: 101802036-101802036 on Build GRCh38
rs775971828 Chr.3: 101801901-101801901 on Build GRCh38
rs758815650 Chr.3: 101801838-101801838 on Build GRCh38
rs746638156 Chr.3: 101801850-101801850 on Build GRCh38
rs781022980 Chr.3: 101801962-101801962 on Build GRCh38
rs368075599 Chr.3: 101801822-101801822 on Build GRCh38
rs761013796 Chr.3: 101801759-101801759 on Build GRCh38
rs201056771 Chr.3: 101801933-101801933 on Build GRCh38
rs768091800 Chr.3: 101801736-101801736 on Build GRCh38
rs150145477 Chr.3: 101801975-101801975 on Build GRCh38
rs760843517 Chr.3: 101802004-101802004 on Build GRCh38
rs746579624 Chr.3: 101801937-101801937 on Build GRCh38
rs765359813 Chr.3: 101801775-101801775 on Build GRCh38
rs766950081 Chr.3: 101801879-101801884 on Build GRCh38
rs747396776 Chr.3: 101801913-101801913 on Build GRCh38
rs773138681 Chr.3: 101802000-101802000 on Build GRCh38
rs868341034 Chr.3: 101801927-101801927 on Build GRCh38
rs201611335 Chr.3: 101801694-101801694 on Build GRCh38
rs766526706 Chr.3: 101801611-101801611 on Build GRCh38
rs863223381 Chr.3: 101801603-101801603 on Build GRCh38
rs778265388 Chr.3: 101801840-101801840 on Build GRCh38
rs574888676 Chr.3: 101801690-101801690 on Build GRCh38
rs975676156 Chr.3: 101801767-101801767 on Build GRCh38
rs367754464 Chr.3: 101801742-101801742 on Build GRCh38
rs772461123 Chr.3: 101801898-101801898 on Build GRCh38
rs768693650 Chr.3: 101801993-101801993 on Build GRCh38
rs190780909 Chr.3: 101801705-101801705 on Build GRCh38
rs751485137 Chr.3: 101801665-101801665 on Build GRCh38
rs767395513 Chr.3: 101801781-101801781 on Build GRCh38
rs1032389873 Chr.3: 101801650-101801650 on Build GRCh38
rs199892236 Chr.3: 101801818-101801818 on Build GRCh38
rs375395786 Chr.3: 101801673-101801673 on Build GRCh38
rs759830675 Chr.3: 101802028-101802028 on Build GRCh38
rs554813235 Chr.3: 101801669-101801669 on Build GRCh38
rs760791122 Chr.3: 101801910-101801910 on Build GRCh38
rs1028612356 Chr.3: 101801817-101801817 on Build GRCh38
rs375557339 Chr.3: 101801964-101801964 on Build GRCh38
rs761812195 Chr.3: 101801778-101801778 on Build GRCh38
rs776545742 Chr.3: 101801862-101801862 on Build GRCh38
rs773424888 Chr.3: 101801757-101801757 on Build GRCh38
rs972115673 Chr.3: 101801881-101801881 on Build GRCh38
rs1000688250 Chr.3: 101801601-101801601 on Build GRCh38
rs763050092 Chr.3: 101801655-101801655 on Build GRCh38
rs759012798 Chr.3: 101801931-101801931 on Build GRCh38
rs754510979 Chr.3: 101801820-101801820 on Build GRCh38

View Full Product Details