Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101821244-101821782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE3
Entrez Gene ID
91775
Gene Name
neurexophilin and PC-esterase domain family member 3
Gene Aliases
FAM55C, MST115, MSTP115
UniGene
Hs.595933
Gene Chromosome Location
Chr.3: 101779185-101828231 on Build GRCh38
Transcripts
NM_001134456.1 NM_145037.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4943253 c.1223A>G p.H408R Chr.3: 101821497-101821497 on Build GRCh38
6731952 c.1444C>T p.R482W Chr.3: 101821718-101821718 on Build GRCh38
2915884 c.1391G>A p.R464H Chr.3: 101821665-101821665 on Build GRCh38
6731954 c.1406G>A p.R469Q Chr.3: 101821680-101821680 on Build GRCh38
2915881 c.1224T>C p.H408H Chr.3: 101821498-101821498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778173816 Chr.3: 101821688-101821688 on Build GRCh38
rs570084480 Chr.3: 101821645-101821645 on Build GRCh38
rs761784698 Chr.3: 101821566-101821566 on Build GRCh38
rs758665528 Chr.3: 101821687-101821687 on Build GRCh38
rs764839422 Chr.3: 101821445-101821445 on Build GRCh38
rs200832757 Chr.3: 101821697-101821697 on Build GRCh38
rs376234770 Chr.3: 101821473-101821473 on Build GRCh38
rs1002484017 Chr.3: 101821388-101821388 on Build GRCh38
rs397735798 Chr.3: 101821314-101821314 on Build GRCh38
rs755032069 Chr.3: 101821591-101821591 on Build GRCh38
rs374457110 Chr.3: 101821650-101821650 on Build GRCh38
rs752875492 Chr.3: 101821684-101821684 on Build GRCh38
rs918148307 Chr.3: 101821290-101821290 on Build GRCh38
rs760736754 Chr.3: 101821679-101821679 on Build GRCh38
rs370722332 Chr.3: 101821696-101821696 on Build GRCh38
rs779031147 Chr.3: 101821512-101821512 on Build GRCh38
rs751502260 Chr.3: 101821481-101821481 on Build GRCh38
rs917930136 Chr.3: 101821537-101821537 on Build GRCh38
rs781676538 Chr.3: 101821699-101821699 on Build GRCh38
rs369282230 Chr.3: 101821426-101821426 on Build GRCh38
rs776065353 Chr.3: 101821418-101821418 on Build GRCh38
rs201325874 Chr.3: 101821719-101821719 on Build GRCh38
rs149452985 Chr.3: 101821491-101821491 on Build GRCh38
rs779515254 Chr.3: 101821706-101821706 on Build GRCh38
rs373062505 Chr.3: 101821278-101821278 on Build GRCh38
rs1009171318 Chr.3: 101821368-101821368 on Build GRCh38
rs375029998 Chr.3: 101821363-101821363 on Build GRCh38
rs759567920 Chr.3: 101821700-101821700 on Build GRCh38
rs143967067 Chr.3: 101821511-101821511 on Build GRCh38
rs548864461 Chr.3: 101821592-101821592 on Build GRCh38
rs747507330 Chr.3: 101821528-101821528 on Build GRCh38
rs374892410 Chr.3: 101821386-101821386 on Build GRCh38
rs1009882535 Chr.3: 101821439-101821439 on Build GRCh38
rs35598292 Chr.3: 101821543-101821543 on Build GRCh38
rs141185345 Chr.3: 101821563-101821563 on Build GRCh38
rs139135706 Chr.3: 101821551-101821551 on Build GRCh38
rs758213453 Chr.3: 101821449-101821449 on Build GRCh38
rs905358846 Chr.3: 101821305-101821305 on Build GRCh38
rs778353603 Chr.3: 101821522-101821522 on Build GRCh38
rs775706577 Chr.3: 101821757-101821757 on Build GRCh38
rs775048030 Chr.3: 101821661-101821661 on Build GRCh38
rs753911382 Chr.3: 101821586-101821586 on Build GRCh38
rs200023533 Chr.3: 101821538-101821538 on Build GRCh38
rs756603058 Chr.3: 101821412-101821413 on Build GRCh38
rs34762683 Chr.3: 101821723-101821723 on Build GRCh38
rs763871177 Chr.3: 101821561-101821561 on Build GRCh38
rs1039200022 Chr.3: 101821305-101821306 on Build GRCh38
rs949627043 Chr.3: 101821294-101821294 on Build GRCh38
rs745327151 Chr.3: 101821647-101821647 on Build GRCh38
rs766462727 Chr.3: 101821728-101821728 on Build GRCh38
rs530088393 Chr.3: 101821307-101821307 on Build GRCh38
rs760510105 Chr.3: 101821724-101821724 on Build GRCh38
rs781572583 Chr.3: 101821646-101821646 on Build GRCh38
rs547768037 Chr.3: 101821490-101821490 on Build GRCh38
rs184213876 Chr.3: 101821335-101821335 on Build GRCh38
rs1053273398 Chr.3: 101821347-101821347 on Build GRCh38
rs398082309 Chr.3: 101821313-101821313 on Build GRCh38
rs762578763 Chr.3: 101821664-101821664 on Build GRCh38
rs758753450 Chr.3: 101821596-101821596 on Build GRCh38
rs377569267 Chr.3: 101821533-101821533 on Build GRCh38
rs914301439 Chr.3: 101821649-101821649 on Build GRCh38
rs775137955 Chr.3: 101821545-101821545 on Build GRCh38
rs778263811 Chr.3: 101821621-101821621 on Build GRCh38
rs140128269 Chr.3: 101821722-101821722 on Build GRCh38
rs747915827 Chr.3: 101821721-101821721 on Build GRCh38
rs747375688 Chr.3: 101821637-101821637 on Build GRCh38
rs35027466 Chr.3: 101821306-101821306 on Build GRCh38
rs886354615 Chr.3: 101821737-101821737 on Build GRCh38
rs1028786921 Chr.3: 101821501-101821501 on Build GRCh38
rs73143886 Chr.3: 101821552-101821552 on Build GRCh38
rs752967344 Chr.3: 101821593-101821593 on Build GRCh38
rs765912363 Chr.3: 101821390-101821390 on Build GRCh38
rs751734957 Chr.3: 101821739-101821739 on Build GRCh38
rs559152261 Chr.3: 101821306-101821307 on Build GRCh38
rs892152088 Chr.3: 101821359-101821359 on Build GRCh38
rs1045199933 Chr.3: 101821299-101821299 on Build GRCh38
rs778261482 Chr.3: 101821663-101821663 on Build GRCh38
rs774098111 Chr.3: 101821667-101821667 on Build GRCh38
rs200120136 Chr.3: 101821519-101821519 on Build GRCh38
rs756671139 Chr.3: 101821705-101821705 on Build GRCh38
rs1033518588 Chr.3: 101821413-101821413 on Build GRCh38
rs532227522 Chr.3: 101821450-101821450 on Build GRCh38
rs767759644 Chr.3: 101821754-101821754 on Build GRCh38
rs574253161 Chr.3: 101821711-101821711 on Build GRCh38
rs372755782 Chr.3: 101821462-101821462 on Build GRCh38
rs552589678 Chr.3: 101821718-101821718 on Build GRCh38
rs752449633 Chr.3: 101821447-101821447 on Build GRCh38
rs750337247 Chr.3: 101821678-101821678 on Build GRCh38
rs765398777 Chr.3: 101821680-101821680 on Build GRCh38
rs200277463 Chr.3: 101821665-101821665 on Build GRCh38
rs373122882 Chr.3: 101821529-101821529 on Build GRCh38
rs150669329 Chr.3: 101821668-101821668 on Build GRCh38
rs757565230 Chr.3: 101821741-101821741 on Build GRCh38
rs757207291 Chr.3: 101821485-101821485 on Build GRCh38
rs763150196 Chr.3: 101821729-101821729 on Build GRCh38
rs1046158356 Chr.3: 101821379-101821379 on Build GRCh38
rs201698560 Chr.3: 101821315-101821317 on Build GRCh38
rs368322476 Chr.3: 101821421-101821421 on Build GRCh38
rs901519570 Chr.3: 101821691-101821691 on Build GRCh38
rs767436113 Chr.3: 101821675-101821675 on Build GRCh38
rs1019927195 Chr.3: 101821443-101821443 on Build GRCh38
rs764354286 Chr.3: 101821734-101821734 on Build GRCh38
rs757576746 Chr.3: 101821692-101821692 on Build GRCh38
rs748471762 Chr.3: 101821516-101821516 on Build GRCh38
rs1053325782 Chr.3: 101821644-101821644 on Build GRCh38

View Full Product Details