Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 105366628-105367124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALCAM
Entrez Gene ID
214
Gene Name
activated leukocyte cell adhesion molecule
Gene Aliases
CD166, MEMD
UniGene
Hs.591293
Gene Chromosome Location
Chr.3: 105366713-105576913 on Build GRCh38
Transcripts
NM_001243280.1 NM_001243281.1 NM_001243283.1 NM_001627.3

SNP Detail

SNP ID Chromosome Location
rs556721698 Chr.3: 105367004-105367006 on Build GRCh38
rs1001060973 Chr.3: 105366829-105366829 on Build GRCh38
rs111709132 Chr.3: 105367032-105367032 on Build GRCh38
rs766169810 Chr.3: 105366866-105366866 on Build GRCh38
rs944862225 Chr.3: 105366744-105366744 on Build GRCh38
rs1005007669 Chr.3: 105366811-105366811 on Build GRCh38
rs925581738 Chr.3: 105366707-105366707 on Build GRCh38
rs146071978 Chr.3: 105366675-105366675 on Build GRCh38
rs951266826 Chr.3: 105366945-105366945 on Build GRCh38
rs918451235 Chr.3: 105366731-105366731 on Build GRCh38
rs368613526 Chr.3: 105366979-105366979 on Build GRCh38
rs955705237 Chr.3: 105366906-105366906 on Build GRCh38
rs1039418412 Chr.3: 105366823-105366823 on Build GRCh38
rs911622609 Chr.3: 105366925-105366925 on Build GRCh38
rs138206557 Chr.3: 105367015-105367015 on Build GRCh38
rs531509943 Chr.3: 105366850-105366850 on Build GRCh38
rs969321092 Chr.3: 105366690-105366690 on Build GRCh38
rs13082435 Chr.3: 105366910-105366910 on Build GRCh38
rs929834264 Chr.3: 105366747-105366747 on Build GRCh38
rs142971151 Chr.3: 105366750-105366750 on Build GRCh38
rs551331956 Chr.3: 105366692-105366692 on Build GRCh38
rs978471179 Chr.3: 105366697-105366697 on Build GRCh38
rs527374458 Chr.3: 105366710-105366710 on Build GRCh38
rs553942605 Chr.3: 105366978-105366978 on Build GRCh38
rs892592401 Chr.3: 105366888-105366888 on Build GRCh38
rs922057939 Chr.3: 105366943-105366943 on Build GRCh38
rs186956057 Chr.3: 105366922-105366922 on Build GRCh38
rs1046576133 Chr.3: 105367028-105367028 on Build GRCh38
rs1012310655 Chr.3: 105366989-105366989 on Build GRCh38
rs114199796 Chr.3: 105367052-105367052 on Build GRCh38
rs1056587198 Chr.3: 105366982-105366982 on Build GRCh38
rs1049065097 Chr.3: 105366783-105366783 on Build GRCh38
rs907902630 Chr.3: 105366970-105366970 on Build GRCh38
rs13082461 Chr.3: 105366968-105366968 on Build GRCh38
rs556397658 Chr.3: 105366781-105366781 on Build GRCh38
rs1023199277 Chr.3: 105366858-105366858 on Build GRCh38
rs182830881 Chr.3: 105366718-105366718 on Build GRCh38
rs1031609942 Chr.3: 105366658-105366658 on Build GRCh38
rs900771290 Chr.3: 105366763-105366763 on Build GRCh38
rs1016241522 Chr.3: 105366924-105366932 on Build GRCh38
rs956972943 Chr.3: 105366659-105366659 on Build GRCh38
rs538753587 Chr.3: 105366935-105366935 on Build GRCh38
rs1044245245 Chr.3: 105367003-105367003 on Build GRCh38
rs1049774448 Chr.3: 105366969-105366969 on Build GRCh38
rs776477480 Chr.3: 105366792-105366792 on Build GRCh38
rs1025231243 Chr.3: 105366689-105366689 on Build GRCh38
rs1022878232 Chr.3: 105366688-105366688 on Build GRCh38
rs1001030296 Chr.3: 105366653-105366653 on Build GRCh38
rs773353438 Chr.3: 105366755-105366755 on Build GRCh38
rs974894473 Chr.3: 105366941-105366941 on Build GRCh38
rs1016479258 Chr.3: 105367074-105367074 on Build GRCh38
rs1000487584 Chr.3: 105366876-105366876 on Build GRCh38
rs1051414580 Chr.3: 105366748-105366748 on Build GRCh38
rs549204790 Chr.3: 105366729-105366729 on Build GRCh38
rs149622390 Chr.3: 105367095-105367095 on Build GRCh38
rs986667416 Chr.3: 105366961-105366961 on Build GRCh38
rs139037862 Chr.3: 105366708-105366708 on Build GRCh38
rs924972479 Chr.3: 105366977-105366977 on Build GRCh38
rs1014080762 Chr.3: 105366840-105366840 on Build GRCh38
rs896593096 Chr.3: 105366983-105366983 on Build GRCh38
rs777181950 Chr.3: 105366651-105366651 on Build GRCh38
rs537605466 Chr.3: 105366751-105366751 on Build GRCh38
rs1056556604 Chr.3: 105366824-105366824 on Build GRCh38
rs147066598 Chr.3: 105366825-105366825 on Build GRCh38
rs1032682961 Chr.3: 105366899-105366899 on Build GRCh38

View Full Product Details