Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 105533359-105533758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALCAM
Entrez Gene ID
214
Gene Name
activated leukocyte cell adhesion molecule
Gene Aliases
CD166, MEMD
UniGene
Hs.591293
Gene Chromosome Location
Chr.3: 105366713-105576913 on Build GRCh38
Transcripts
NM_001243280.1 NM_001243281.1 NM_001627.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
318646 c.536C>A p.P179H Chr.3: 105533679-105533679 on Build GRCh38
4111999 c.498C>T p.G166G Chr.3: 105533641-105533641 on Build GRCh38
138385 c.536C>T p.P179L Chr.3: 105533679-105533679 on Build GRCh38
4112000 c.533A>G p.H178R Chr.3: 105533676-105533676 on Build GRCh38
3372937 c.470G>A p.C157Y Chr.3: 105533613-105533613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552369951 Chr.3: 105533419-105533419 on Build GRCh38
rs763351683 Chr.3: 105533605-105533605 on Build GRCh38
rs769622438 Chr.3: 105533605-105533605 on Build GRCh38
rs925834406 Chr.3: 105533536-105533536 on Build GRCh38
rs771054944 Chr.3: 105533662-105533662 on Build GRCh38
rs747816561 Chr.3: 105533497-105533497 on Build GRCh38
rs1035212596 Chr.3: 105533435-105533435 on Build GRCh38
rs867478283 Chr.3: 105533460-105533460 on Build GRCh38
rs777181155 Chr.3: 105533561-105533561 on Build GRCh38
rs371886215 Chr.3: 105533447-105533447 on Build GRCh38
rs886701103 Chr.3: 105533413-105533413 on Build GRCh38
rs1023758063 Chr.3: 105533503-105533503 on Build GRCh38
rs765345746 Chr.3: 105533455-105533455 on Build GRCh38
rs564200245 Chr.3: 105533425-105533425 on Build GRCh38
rs990888172 Chr.3: 105533482-105533482 on Build GRCh38
rs369564690 Chr.3: 105533551-105533551 on Build GRCh38
rs555994583 Chr.3: 105533627-105533627 on Build GRCh38
rs2171146 Chr.3: 105533533-105533533 on Build GRCh38
rs968036322 Chr.3: 105533528-105533528 on Build GRCh38
rs753306688 Chr.3: 105533553-105533553 on Build GRCh38
rs759738526 Chr.3: 105533677-105533677 on Build GRCh38
rs760885989 Chr.3: 105533638-105533638 on Build GRCh38
rs777052818 Chr.3: 105533668-105533668 on Build GRCh38
rs1021670229 Chr.3: 105533557-105533557 on Build GRCh38
rs570585856 Chr.3: 105533542-105533542 on Build GRCh38
rs1027823327 Chr.3: 105533399-105533399 on Build GRCh38
rs1038757546 Chr.3: 105533617-105533617 on Build GRCh38
rs374454592 Chr.3: 105533565-105533565 on Build GRCh38
rs769482222 Chr.3: 105533530-105533530 on Build GRCh38
rs570204103 Chr.3: 105533394-105533394 on Build GRCh38
rs753218420 Chr.3: 105533700-105533700 on Build GRCh38
rs563710596 Chr.3: 105533390-105533390 on Build GRCh38
rs756844872 Chr.3: 105533729-105533729 on Build GRCh38
rs997767400 Chr.3: 105533531-105533531 on Build GRCh38
rs780960968 Chr.3: 105533594-105533594 on Build GRCh38
rs749252813 Chr.3: 105533620-105533620 on Build GRCh38
rs992071063 Chr.3: 105533726-105533726 on Build GRCh38
rs751024594 Chr.3: 105533723-105533723 on Build GRCh38
rs1044849933 Chr.3: 105533459-105533459 on Build GRCh38
rs763396627 Chr.3: 105533718-105533718 on Build GRCh38
rs76635565 Chr.3: 105533637-105533637 on Build GRCh38
rs763604825 Chr.3: 105533721-105533721 on Build GRCh38
rs942197283 Chr.3: 105533467-105533467 on Build GRCh38
rs921689530 Chr.3: 105533623-105533623 on Build GRCh38
rs947809579 Chr.3: 105533458-105533458 on Build GRCh38
rs900170221 Chr.3: 105533383-105533383 on Build GRCh38
rs775373645 Chr.3: 105533611-105533611 on Build GRCh38
rs368628373 Chr.3: 105533587-105533587 on Build GRCh38
rs956002936 Chr.3: 105533538-105533538 on Build GRCh38
rs537051127 Chr.3: 105533452-105533452 on Build GRCh38
rs569070957 Chr.3: 105533584-105533584 on Build GRCh38
rs148287455 Chr.3: 105533554-105533554 on Build GRCh38
rs765739761 Chr.3: 105533680-105533680 on Build GRCh38
rs112322448 Chr.3: 105533632-105533632 on Build GRCh38
rs745646740 Chr.3: 105533603-105533603 on Build GRCh38
rs917575176 Chr.3: 105533452-105533453 on Build GRCh38
rs956864853 Chr.3: 105533679-105533679 on Build GRCh38
rs759959513 Chr.3: 105533674-105533674 on Build GRCh38
rs377006541 Chr.3: 105533418-105533418 on Build GRCh38
rs546588595 Chr.3: 105533483-105533483 on Build GRCh38

View Full Product Details