Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 105534585-105535095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALCAM
Entrez Gene ID
214
Gene Name
activated leukocyte cell adhesion molecule
Gene Aliases
CD166, MEMD
UniGene
Hs.591293
Gene Chromosome Location
Chr.3: 105366713-105576913 on Build GRCh38
Transcripts
NM_001243280.1 NM_001243281.1 NM_001627.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1035936 c.633G>T p.K211N Chr.3: 105534748-105534748 on Build GRCh38
5059177 c.552G>A p.V184V Chr.3: 105534667-105534667 on Build GRCh38
1035937 c.649C>A p.P217T Chr.3: 105534764-105534764 on Build GRCh38
4112001 c.712G>A p.A238T Chr.3: 105534827-105534827 on Build GRCh38
3845722 c.654C>T p.F218F Chr.3: 105534769-105534769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369249025 Chr.3: 105534643-105534643 on Build GRCh38
rs754860502 Chr.3: 105534731-105534731 on Build GRCh38
rs548916544 Chr.3: 105534914-105534914 on Build GRCh38
rs754693995 Chr.3: 105534750-105534750 on Build GRCh38
rs6437601 Chr.3: 105534631-105534631 on Build GRCh38
rs372983523 Chr.3: 105534860-105534860 on Build GRCh38
rs878919173 Chr.3: 105534908-105534909 on Build GRCh38
rs377299560 Chr.3: 105534862-105534862 on Build GRCh38
rs372772944 Chr.3: 105534819-105534819 on Build GRCh38
rs914043141 Chr.3: 105534958-105534958 on Build GRCh38
rs200414406 Chr.3: 105534684-105534684 on Build GRCh38
rs753626110 Chr.3: 105534730-105534730 on Build GRCh38
rs988857726 Chr.3: 105534855-105534855 on Build GRCh38
rs772713149 Chr.3: 105534858-105534858 on Build GRCh38
rs761194368 Chr.3: 105534695-105534695 on Build GRCh38
rs929199765 Chr.3: 105535034-105535036 on Build GRCh38
rs753576438 Chr.3: 105534870-105534870 on Build GRCh38
rs377120473 Chr.3: 105534714-105534714 on Build GRCh38
rs112184836 Chr.3: 105534697-105534697 on Build GRCh38
rs762403213 Chr.3: 105534820-105534820 on Build GRCh38
rs958550479 Chr.3: 105534905-105534905 on Build GRCh38
rs746032698 Chr.3: 105534777-105534777 on Build GRCh38
rs988433699 Chr.3: 105534922-105534922 on Build GRCh38
rs927330304 Chr.3: 105534947-105534947 on Build GRCh38
rs752152863 Chr.3: 105534690-105534690 on Build GRCh38
rs961935392 Chr.3: 105534789-105534789 on Build GRCh38
rs184700099 Chr.3: 105535037-105535037 on Build GRCh38
rs527448102 Chr.3: 105534910-105534910 on Build GRCh38
rs764492238 Chr.3: 105534689-105534689 on Build GRCh38
rs1028825774 Chr.3: 105534722-105534722 on Build GRCh38
rs373637035 Chr.3: 105534633-105534633 on Build GRCh38
rs577619201 Chr.3: 105534763-105534763 on Build GRCh38
rs1013746699 Chr.3: 105534800-105534800 on Build GRCh38
rs144172449 Chr.3: 105534663-105534663 on Build GRCh38
rs747046838 Chr.3: 105534665-105534665 on Build GRCh38
rs750026519 Chr.3: 105534711-105534711 on Build GRCh38
rs879915272 Chr.3: 105534648-105534648 on Build GRCh38
rs79951414 Chr.3: 105534787-105534787 on Build GRCh38
rs10933819 Chr.3: 105534801-105534801 on Build GRCh38
rs779756906 Chr.3: 105534716-105534716 on Build GRCh38
rs374212439 Chr.3: 105534889-105534889 on Build GRCh38
rs770879279 Chr.3: 105534766-105534766 on Build GRCh38
rs111722123 Chr.3: 105534778-105534778 on Build GRCh38
rs769102419 Chr.3: 105534796-105534796 on Build GRCh38
rs1008582488 Chr.3: 105534748-105534748 on Build GRCh38
rs899113908 Chr.3: 105534969-105534969 on Build GRCh38
rs1037934078 Chr.3: 105534965-105534965 on Build GRCh38
rs921348777 Chr.3: 105535039-105535039 on Build GRCh38
rs371943794 Chr.3: 105534668-105534668 on Build GRCh38
rs748157879 Chr.3: 105534764-105534764 on Build GRCh38
rs781446347 Chr.3: 105534769-105534769 on Build GRCh38
rs755861717 Chr.3: 105534636-105534636 on Build GRCh38
rs944253258 Chr.3: 105534972-105534972 on Build GRCh38
rs996840691 Chr.3: 105534718-105534718 on Build GRCh38
rs766953782 Chr.3: 105534642-105534642 on Build GRCh38
rs759871973 Chr.3: 105534672-105534672 on Build GRCh38
rs368171432 Chr.3: 105534739-105534739 on Build GRCh38
rs766954758 Chr.3: 105534840-105534840 on Build GRCh38
rs567307615 Chr.3: 105535007-105535007 on Build GRCh38
rs887670635 Chr.3: 105535062-105535062 on Build GRCh38
rs780684310 Chr.3: 105534617-105534617 on Build GRCh38
rs776014875 Chr.3: 105534680-105534680 on Build GRCh38
rs755632205 Chr.3: 105535068-105535068 on Build GRCh38
rs368126568 Chr.3: 105534659-105534659 on Build GRCh38
rs148602469 Chr.3: 105534874-105534874 on Build GRCh38
rs775908598 Chr.3: 105534786-105534786 on Build GRCh38
rs751414998 Chr.3: 105534664-105534664 on Build GRCh38
rs770238326 Chr.3: 105534677-105534677 on Build GRCh38
rs73006174 Chr.3: 105535043-105535043 on Build GRCh38
rs1048393045 Chr.3: 105535049-105535049 on Build GRCh38
rs570947513 Chr.3: 105535060-105535060 on Build GRCh38

View Full Product Details