Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 105539856-105540393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALCAM
Entrez Gene ID
214
Gene Name
activated leukocyte cell adhesion molecule
Gene Aliases
CD166, MEMD
UniGene
Hs.591293
Gene Chromosome Location
Chr.3: 105366713-105576913 on Build GRCh38
Transcripts
NM_001243280.1 NM_001243281.1 NM_001627.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3759579 c.773A>G p.N258S Chr.3: 105540017-105540017 on Build GRCh38
1732799 c.768delA p.N258fs*14 Chr.3: 105540012-105540012 on Build GRCh38
5727416 c.731-1G>A p.? Chr.3: 105539974-105539974 on Build GRCh38
4429451 c.790G>C p.D264H Chr.3: 105540034-105540034 on Build GRCh38
6266239 c.846G>T p.L282F Chr.3: 105540090-105540090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758489960 Chr.3: 105540117-105540117 on Build GRCh38
rs774694563 Chr.3: 105540108-105540118 on Build GRCh38
rs373519238 Chr.3: 105540182-105540183 on Build GRCh38
rs760157001 Chr.3: 105539997-105539997 on Build GRCh38
rs376241637 Chr.3: 105540133-105540133 on Build GRCh38
rs755231406 Chr.3: 105540042-105540042 on Build GRCh38
rs773767295 Chr.3: 105539989-105539989 on Build GRCh38
rs980986388 Chr.3: 105540200-105540200 on Build GRCh38
rs919271898 Chr.3: 105540044-105540044 on Build GRCh38
rs11328261 Chr.3: 105540304-105540304 on Build GRCh38
rs925403884 Chr.3: 105540250-105540250 on Build GRCh38
rs879240854 Chr.3: 105540248-105540248 on Build GRCh38
rs764886320 Chr.3: 105540012-105540012 on Build GRCh38
rs397990706 Chr.3: 105540312-105540312 on Build GRCh38
rs767883557 Chr.3: 105540136-105540137 on Build GRCh38
rs372752372 Chr.3: 105540131-105540131 on Build GRCh38
rs75640036 Chr.3: 105540311-105540312 on Build GRCh38
rs755109930 Chr.3: 105540231-105540231 on Build GRCh38
rs759952514 Chr.3: 105540118-105540121 on Build GRCh38
rs752545144 Chr.3: 105540016-105540016 on Build GRCh38
rs776220454 Chr.3: 105540082-105540082 on Build GRCh38
rs770798533 Chr.3: 105540165-105540165 on Build GRCh38
rs545922334 Chr.3: 105540328-105540328 on Build GRCh38
rs374882622 Chr.3: 105540038-105540038 on Build GRCh38
rs750919004 Chr.3: 105540106-105540106 on Build GRCh38
rs776110226 Chr.3: 105540003-105540003 on Build GRCh38
rs991922265 Chr.3: 105539988-105539988 on Build GRCh38
rs962270653 Chr.3: 105540195-105540195 on Build GRCh38
rs908100077 Chr.3: 105540172-105540172 on Build GRCh38
rs903815128 Chr.3: 105540018-105540018 on Build GRCh38
rs996029276 Chr.3: 105540360-105540360 on Build GRCh38
rs150404737 Chr.3: 105540268-105540268 on Build GRCh38
rs749410741 Chr.3: 105540040-105540040 on Build GRCh38
rs35248527 Chr.3: 105540006-105540006 on Build GRCh38
rs184508448 Chr.3: 105539931-105539931 on Build GRCh38
rs201952077 Chr.3: 105539980-105539980 on Build GRCh38
rs951782756 Chr.3: 105539890-105539890 on Build GRCh38
rs113972673 Chr.3: 105539910-105539910 on Build GRCh38
rs778223241 Chr.3: 105540062-105540062 on Build GRCh38
rs180824660 Chr.3: 105539930-105539930 on Build GRCh38
rs751491217 Chr.3: 105540116-105540116 on Build GRCh38
rs372163777 Chr.3: 105540312-105540312 on Build GRCh38
rs775507165 Chr.3: 105540191-105540192 on Build GRCh38
rs768851829 Chr.3: 105539967-105539967 on Build GRCh38
rs749553113 Chr.3: 105539951-105539951 on Build GRCh38
rs918069817 Chr.3: 105540212-105540212 on Build GRCh38
rs936644890 Chr.3: 105540257-105540257 on Build GRCh38
rs113403424 Chr.3: 105539896-105539896 on Build GRCh38
rs769486800 Chr.3: 105540091-105540091 on Build GRCh38
rs756219834 Chr.3: 105540037-105540037 on Build GRCh38
rs542014731 Chr.3: 105540299-105540299 on Build GRCh38
rs373142677 Chr.3: 105539946-105539946 on Build GRCh38
rs770380290 Chr.3: 105540069-105540069 on Build GRCh38
rs909389669 Chr.3: 105540238-105540238 on Build GRCh38
rs948249051 Chr.3: 105540230-105540230 on Build GRCh38
rs963484771 Chr.3: 105540256-105540256 on Build GRCh38
rs563580937 Chr.3: 105540302-105540302 on Build GRCh38
rs752281909 Chr.3: 105540148-105540150 on Build GRCh38
rs779173959 Chr.3: 105540139-105540139 on Build GRCh38
rs201173033 Chr.3: 105540085-105540085 on Build GRCh38
rs182099822 Chr.3: 105540350-105540350 on Build GRCh38
rs1052370180 Chr.3: 105540278-105540278 on Build GRCh38
rs775067433 Chr.3: 105540100-105540100 on Build GRCh38
rs530897182 Chr.3: 105540327-105540327 on Build GRCh38
rs553588905 Chr.3: 105540171-105540171 on Build GRCh38
rs992438568 Chr.3: 105540199-105540199 on Build GRCh38
rs77157868 Chr.3: 105540127-105540127 on Build GRCh38
rs751584013 Chr.3: 105540035-105540035 on Build GRCh38
rs1044240 Chr.3: 105540017-105540017 on Build GRCh38
rs764096895 Chr.3: 105540023-105540023 on Build GRCh38
rs984467363 Chr.3: 105540167-105540167 on Build GRCh38
rs112128595 Chr.3: 105540246-105540246 on Build GRCh38
rs1040306173 Chr.3: 105540338-105540338 on Build GRCh38
rs779070106 Chr.3: 105540043-105540043 on Build GRCh38
rs141740492 Chr.3: 105540102-105540102 on Build GRCh38
rs756307491 Chr.3: 105539937-105539937 on Build GRCh38
rs772474173 Chr.3: 105540048-105540048 on Build GRCh38
rs761853010 Chr.3: 105540120-105540120 on Build GRCh38
rs774008127 Chr.3: 105540216-105540216 on Build GRCh38
rs747547045 Chr.3: 105540065-105540065 on Build GRCh38
rs772563885 Chr.3: 105539985-105539985 on Build GRCh38
rs754203338 Chr.3: 105540004-105540004 on Build GRCh38
rs933361586 Chr.3: 105540321-105540321 on Build GRCh38
rs369658061 Chr.3: 105540147-105540147 on Build GRCh38
rs151133594 Chr.3: 105540001-105540001 on Build GRCh38
rs752530380 Chr.3: 105539924-105539924 on Build GRCh38
rs765730707 Chr.3: 105540022-105540022 on Build GRCh38
rs200883766 Chr.3: 105540124-105540124 on Build GRCh38
rs147654520 Chr.3: 105540363-105540363 on Build GRCh38
rs774790957 Chr.3: 105539977-105539977 on Build GRCh38
rs908129528 Chr.3: 105539920-105539920 on Build GRCh38

View Full Product Details