Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108624252-108624776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZIP3
Entrez Gene ID
9666
Gene Name
DAZ interacting zinc finger protein 3
Gene Aliases
PPP1R66, UURF2, hRUL138
UniGene
Hs.409210
Gene Chromosome Location
Chr.3: 108589490-108694846 on Build GRCh38
Transcripts
NM_014648.3 XM_005247914.3 XM_005247915.3 XM_005247916.3 XM_005247917.3 XM_005247918.3 XM_005247919.3 XM_017007540.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1308365 c.439G>A p.E147K Chr.3: 108624507-108624507 on Build GRCh38
244176 c.377C>T p.A126V Chr.3: 108624445-108624445 on Build GRCh38
4408548 c.441A>G p.E147E Chr.3: 108624509-108624509 on Build GRCh38
5791746 c.453A>G p.K151K Chr.3: 108624521-108624521 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758569528 Chr.3: 108624420-108624420 on Build GRCh38
rs770449172 Chr.3: 108624544-108624544 on Build GRCh38
rs746340279 Chr.3: 108624543-108624543 on Build GRCh38
rs900376274 Chr.3: 108624671-108624694 on Build GRCh38
rs911734957 Chr.3: 108624751-108624751 on Build GRCh38
rs779561184 Chr.3: 108624546-108624546 on Build GRCh38
rs775821630 Chr.3: 108624511-108624511 on Build GRCh38
rs9288878 Chr.3: 108624617-108624617 on Build GRCh38
rs1039951175 Chr.3: 108624302-108624302 on Build GRCh38
rs775174364 Chr.3: 108624462-108624462 on Build GRCh38
rs934008831 Chr.3: 108624338-108624338 on Build GRCh38
rs771813038 Chr.3: 108624572-108624572 on Build GRCh38
rs529995975 Chr.3: 108624514-108624514 on Build GRCh38
rs369866805 Chr.3: 108624463-108624463 on Build GRCh38
rs200974372 Chr.3: 108624509-108624509 on Build GRCh38
rs751940620 Chr.3: 108624525-108624525 on Build GRCh38
rs953223616 Chr.3: 108624705-108624705 on Build GRCh38
rs921732840 Chr.3: 108624384-108624386 on Build GRCh38
rs769687462 Chr.3: 108624442-108624442 on Build GRCh38
rs878939434 Chr.3: 108624303-108624303 on Build GRCh38
rs761565166 Chr.3: 108624568-108624568 on Build GRCh38
rs772863286 Chr.3: 108624651-108624654 on Build GRCh38
rs562537079 Chr.3: 108624342-108624342 on Build GRCh38
rs768928695 Chr.3: 108624723-108624723 on Build GRCh38
rs371283195 Chr.3: 108624688-108624689 on Build GRCh38
rs547815580 Chr.3: 108624448-108624448 on Build GRCh38
rs887197089 Chr.3: 108624345-108624345 on Build GRCh38
rs545249886 Chr.3: 108624410-108624410 on Build GRCh38
rs577251490 Chr.3: 108624426-108624426 on Build GRCh38
rs921539242 Chr.3: 108624663-108624663 on Build GRCh38
rs777925110 Chr.3: 108624518-108624518 on Build GRCh38
rs202179235 Chr.3: 108624445-108624445 on Build GRCh38
rs375057588 Chr.3: 108624411-108624411 on Build GRCh38
rs915432352 Chr.3: 108624626-108624626 on Build GRCh38
rs147007758 Chr.3: 108624491-108624491 on Build GRCh38
rs774189036 Chr.3: 108624488-108624488 on Build GRCh38
rs983223188 Chr.3: 108624304-108624304 on Build GRCh38
rs181576494 Chr.3: 108624516-108624516 on Build GRCh38
rs184868092 Chr.3: 108624444-108624444 on Build GRCh38
rs969264314 Chr.3: 108624500-108624500 on Build GRCh38
rs78132162 Chr.3: 108624473-108624473 on Build GRCh38
rs757745957 Chr.3: 108624527-108624527 on Build GRCh38
rs573698924 Chr.3: 108624676-108624676 on Build GRCh38
rs748813024 Chr.3: 108624550-108624550 on Build GRCh38
rs760493447 Chr.3: 108624449-108624450 on Build GRCh38
rs768338179 Chr.3: 108624560-108624560 on Build GRCh38
rs767493090 Chr.3: 108624412-108624413 on Build GRCh38
rs1051024919 Chr.3: 108624339-108624339 on Build GRCh38
rs773919925 Chr.3: 108624566-108624566 on Build GRCh38
rs866929008 Chr.3: 108624610-108624610 on Build GRCh38
rs113039152 Chr.3: 108624299-108624299 on Build GRCh38
rs778144529 Chr.3: 108624425-108624425 on Build GRCh38
rs78119686 Chr.3: 108624670-108624670 on Build GRCh38
rs747340995 Chr.3: 108624430-108624430 on Build GRCh38
rs988716573 Chr.3: 108624277-108624277 on Build GRCh38
rs750400768 Chr.3: 108624497-108624497 on Build GRCh38
rs752693091 Chr.3: 108624429-108624429 on Build GRCh38
rs181844635 Chr.3: 108624645-108624645 on Build GRCh38
rs73210672 Chr.3: 108624641-108624641 on Build GRCh38
rs531964771 Chr.3: 108624372-108624372 on Build GRCh38
rs752923261 Chr.3: 108624412-108624412 on Build GRCh38
rs74527976 Chr.3: 108624534-108624534 on Build GRCh38
rs904207346 Chr.3: 108624483-108624483 on Build GRCh38
rs13090268 Chr.3: 108624300-108624300 on Build GRCh38
rs899875649 Chr.3: 108624323-108624323 on Build GRCh38
rs376413790 Chr.3: 108624285-108624285 on Build GRCh38
rs755723605 Chr.3: 108624433-108624433 on Build GRCh38
rs956666539 Chr.3: 108624595-108624595 on Build GRCh38
rs184179156 Chr.3: 108624633-108624633 on Build GRCh38
rs34808574 Chr.3: 108624300-108624302 on Build GRCh38
rs149336494 Chr.3: 108624476-108624476 on Build GRCh38

View Full Product Details