Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 109300369-109300879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPPA2
Entrez Gene ID
151871
Gene Name
developmental pluripotency associated 2
Gene Aliases
CT100, ECAT15-2, PESCRG1
UniGene
Hs.351113
Gene Chromosome Location
Chr.3: 109293788-109316553 on Build GRCh38
Transcripts
NM_138815.3 XM_011512443.1 XM_011512444.2 XM_011512445.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7248159 c.895T>A p.*299K Chr.3: 109300395-109300395 on Build GRCh38
4317634 c.864G>C p.K288N Chr.3: 109300426-109300426 on Build GRCh38
3585114 c.877T>A p.L293I Chr.3: 109300413-109300413 on Build GRCh38
3585115 c.855G>A p.R285R Chr.3: 109300435-109300435 on Build GRCh38
6162760 c.886G>A p.V296I Chr.3: 109300404-109300404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749376125 Chr.3: 109300450-109300450 on Build GRCh38
rs570352786 Chr.3: 109300650-109300650 on Build GRCh38
rs370418198 Chr.3: 109300843-109300843 on Build GRCh38
rs973845222 Chr.3: 109300705-109300705 on Build GRCh38
rs368690006 Chr.3: 109300461-109300461 on Build GRCh38
rs773695181 Chr.3: 109300823-109300824 on Build GRCh38
rs757263815 Chr.3: 109300443-109300443 on Build GRCh38
rs776180781 Chr.3: 109300486-109300486 on Build GRCh38
rs981509756 Chr.3: 109300500-109300500 on Build GRCh38
rs774453659 Chr.3: 109300822-109300822 on Build GRCh38
rs934875018 Chr.3: 109300515-109300515 on Build GRCh38
rs772366959 Chr.3: 109300476-109300476 on Build GRCh38
rs762977506 Chr.3: 109300523-109300523 on Build GRCh38
rs73852687 Chr.3: 109300543-109300543 on Build GRCh38
rs771526581 Chr.3: 109300822-109300824 on Build GRCh38
rs901513466 Chr.3: 109300576-109300576 on Build GRCh38
rs869226849 Chr.3: 109300821-109300823 on Build GRCh38
rs764845192 Chr.3: 109300394-109300394 on Build GRCh38
rs548466199 Chr.3: 109300625-109300625 on Build GRCh38
rs752936191 Chr.3: 109300438-109300438 on Build GRCh38
rs747477710 Chr.3: 109300484-109300484 on Build GRCh38
rs139530575 Chr.3: 109300425-109300425 on Build GRCh38
rs963029830 Chr.3: 109300427-109300427 on Build GRCh38
rs767043097 Chr.3: 109300824-109300824 on Build GRCh38
rs886253088 Chr.3: 109300666-109300666 on Build GRCh38
rs765421702 Chr.3: 109300415-109300415 on Build GRCh38
rs971383663 Chr.3: 109300729-109300729 on Build GRCh38
rs1031740619 Chr.3: 109300771-109300771 on Build GRCh38
rs869090260 Chr.3: 109300842-109300842 on Build GRCh38
rs1022928709 Chr.3: 109300448-109300448 on Build GRCh38
rs971414753 Chr.3: 109300491-109300491 on Build GRCh38
rs9865540 Chr.3: 109300588-109300588 on Build GRCh38
rs754401561 Chr.3: 109300734-109300735 on Build GRCh38
rs751483506 Chr.3: 109300440-109300440 on Build GRCh38
rs62274254 Chr.3: 109300653-109300653 on Build GRCh38
rs768113129 Chr.3: 109300657-109300657 on Build GRCh38
rs773575302 Chr.3: 109300480-109300480 on Build GRCh38
rs750797945 Chr.3: 109300679-109300679 on Build GRCh38
rs763036997 Chr.3: 109300534-109300534 on Build GRCh38
rs953919917 Chr.3: 109300728-109300728 on Build GRCh38
rs976127425 Chr.3: 109300834-109300834 on Build GRCh38
rs72034182 Chr.3: 109300735-109300738 on Build GRCh38
rs531020504 Chr.3: 109300689-109300689 on Build GRCh38
rs4855589 Chr.3: 109300822-109300822 on Build GRCh38
rs534796010 Chr.3: 109300770-109300770 on Build GRCh38
rs768893998 Chr.3: 109300457-109300457 on Build GRCh38
rs1022897515 Chr.3: 109300715-109300715 on Build GRCh38
rs10575889 Chr.3: 109300738-109300741 on Build GRCh38
rs552775835 Chr.3: 109300686-109300686 on Build GRCh38
rs201146409 Chr.3: 109300593-109300593 on Build GRCh38
rs764572643 Chr.3: 109300559-109300559 on Build GRCh38
rs924753670 Chr.3: 109300508-109300508 on Build GRCh38
rs778670416 Chr.3: 109300713-109300713 on Build GRCh38
rs550514936 Chr.3: 109300592-109300592 on Build GRCh38

View Full Product Details