Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 111567406-111567947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD96
Entrez Gene ID
10225
Gene Name
CD96 molecule
Gene Aliases
TACTILE
UniGene
Hs.142023
Gene Chromosome Location
Chr.3: 111542079-111665996 on Build GRCh38
Transcripts
NM_001318889.1 NM_005816.4 NM_198196.2 NR_134917.1 XM_005247063.3 XM_006713469.3 XM_006713470.3 XM_017005521.1 XM_017005522.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6192733 c.423A>T p.T141T Chr.3: 111567527-111567527 on Build GRCh38
4900904 c.515C>T p.S172L Chr.3: 111567619-111567619 on Build GRCh38
4999529 c.424G>C p.A142P Chr.3: 111567528-111567528 on Build GRCh38
3845781 c.460G>A p.E154K Chr.3: 111567564-111567564 on Build GRCh38
3682920 c.424G>T p.A142S Chr.3: 111567528-111567528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890313310 Chr.3: 111567893-111567893 on Build GRCh38
rs139639575 Chr.3: 111567635-111567635 on Build GRCh38
rs532537422 Chr.3: 111567604-111567604 on Build GRCh38
rs758588487 Chr.3: 111567708-111567708 on Build GRCh38
rs921359814 Chr.3: 111567797-111567797 on Build GRCh38
rs1014584510 Chr.3: 111567634-111567634 on Build GRCh38
rs146909370 Chr.3: 111567753-111567753 on Build GRCh38
rs7629206 Chr.3: 111567912-111567912 on Build GRCh38
rs200635065 Chr.3: 111567682-111567682 on Build GRCh38
rs775536421 Chr.3: 111567497-111567497 on Build GRCh38
rs956860029 Chr.3: 111567793-111567793 on Build GRCh38
rs775765829 Chr.3: 111567652-111567652 on Build GRCh38
rs147832650 Chr.3: 111567556-111567556 on Build GRCh38
rs753189773 Chr.3: 111567541-111567541 on Build GRCh38
rs374066536 Chr.3: 111567630-111567630 on Build GRCh38
rs947864774 Chr.3: 111567869-111567869 on Build GRCh38
rs777881366 Chr.3: 111567727-111567727 on Build GRCh38
rs760810384 Chr.3: 111567513-111567513 on Build GRCh38
rs368955050 Chr.3: 111567668-111567668 on Build GRCh38
rs113905243 Chr.3: 111567845-111567845 on Build GRCh38
rs745602337 Chr.3: 111567486-111567486 on Build GRCh38
rs755948670 Chr.3: 111567585-111567585 on Build GRCh38
rs1043814319 Chr.3: 111567542-111567542 on Build GRCh38
rs1043757321 Chr.3: 111567788-111567788 on Build GRCh38
rs537299593 Chr.3: 111567813-111567813 on Build GRCh38
rs568154032 Chr.3: 111567778-111567778 on Build GRCh38
rs780670315 Chr.3: 111567557-111567557 on Build GRCh38
rs989399121 Chr.3: 111567809-111567809 on Build GRCh38
rs756688474 Chr.3: 111567686-111567686 on Build GRCh38
rs371659001 Chr.3: 111567588-111567588 on Build GRCh38
rs765581924 Chr.3: 111567644-111567644 on Build GRCh38
rs369084693 Chr.3: 111567554-111567554 on Build GRCh38
rs765642865 Chr.3: 111567532-111567532 on Build GRCh38
rs932767458 Chr.3: 111567816-111567816 on Build GRCh38
rs376355617 Chr.3: 111567656-111567656 on Build GRCh38
rs774839745 Chr.3: 111567699-111567699 on Build GRCh38
rs780911928 Chr.3: 111567482-111567482 on Build GRCh38
rs530355737 Chr.3: 111567433-111567433 on Build GRCh38
rs758233817 Chr.3: 111567577-111567577 on Build GRCh38
rs563996981 Chr.3: 111567591-111567591 on Build GRCh38
rs190624304 Chr.3: 111567710-111567710 on Build GRCh38
rs367821277 Chr.3: 111567489-111567489 on Build GRCh38
rs1050260919 Chr.3: 111567839-111567839 on Build GRCh38
rs939311107 Chr.3: 111567432-111567432 on Build GRCh38
rs995037220 Chr.3: 111567658-111567658 on Build GRCh38
rs770423662 Chr.3: 111567475-111567475 on Build GRCh38
rs759813586 Chr.3: 111567638-111567638 on Build GRCh38
rs767100930 Chr.3: 111567698-111567698 on Build GRCh38
rs745553922 Chr.3: 111567566-111567566 on Build GRCh38
rs2276872 Chr.3: 111567528-111567528 on Build GRCh38
rs566103646 Chr.3: 111567741-111567741 on Build GRCh38
rs976676936 Chr.3: 111567455-111567455 on Build GRCh38
rs868102725 Chr.3: 111567589-111567589 on Build GRCh38
rs772265929 Chr.3: 111567510-111567510 on Build GRCh38
rs142624479 Chr.3: 111567637-111567637 on Build GRCh38
rs759864939 Chr.3: 111567529-111567529 on Build GRCh38
rs994567443 Chr.3: 111567640-111567640 on Build GRCh38
rs749228868 Chr.3: 111567590-111567590 on Build GRCh38
rs758971440 Chr.3: 111567551-111567551 on Build GRCh38
rs1036296616 Chr.3: 111567922-111567922 on Build GRCh38
rs773759995 Chr.3: 111567641-111567641 on Build GRCh38
rs576288492 Chr.3: 111567762-111567762 on Build GRCh38
rs764463486 Chr.3: 111567681-111567681 on Build GRCh38
rs766450137 Chr.3: 111567522-111567522 on Build GRCh38
rs149798144 Chr.3: 111567553-111567553 on Build GRCh38
rs774358172 Chr.3: 111567602-111567602 on Build GRCh38
rs772402974 Chr.3: 111567577-111567578 on Build GRCh38
rs904336516 Chr.3: 111567549-111567549 on Build GRCh38
rs749286076 Chr.3: 111567509-111567509 on Build GRCh38
rs200256046 Chr.3: 111567511-111567511 on Build GRCh38

View Full Product Details