Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 111593320-111593814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD96
Entrez Gene ID
10225
Gene Name
CD96 molecule
Gene Aliases
TACTILE
UniGene
Hs.142023
Gene Chromosome Location
Chr.3: 111542079-111665996 on Build GRCh38
Transcripts
NM_001318889.1 NM_005816.4 NM_198196.2 NR_134917.1 XM_005247063.3 XM_006713469.3 XM_006713470.3 XM_017005521.1 XM_017005522.1 NM_024508.4

Gene Symbol
ZBED2
Entrez Gene ID
79413
Gene Name
zinc finger BED-type containing 2
Gene Aliases
-
UniGene
Hs.136912
Gene Chromosome Location
Chr.3: 111592900-111595335 on Build GRCh38
Transcripts
NM_001318889.1 NM_005816.4 NM_198196.2 NR_134917.1 XM_005247063.3 XM_006713469.3 XM_006713470.3 XM_017005521.1 XM_017005522.1 NM_024508.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4991819 c.436G>A p.V146M Chr.3: 111593766-111593766 on Build GRCh38
324352 c.548A>T p.E183V Chr.3: 111593654-111593654 on Build GRCh38
4991818 c.477G>A p.E159E Chr.3: 111593725-111593725 on Build GRCh38
3585202 c.646C>T p.H216Y Chr.3: 111593556-111593556 on Build GRCh38
5059193 c.497C>T p.A166V Chr.3: 111593705-111593705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200890861 Chr.3: 111593706-111593706 on Build GRCh38
rs987115682 Chr.3: 111593385-111593385 on Build GRCh38
rs376107359 Chr.3: 111593694-111593694 on Build GRCh38
rs148715222 Chr.3: 111593793-111593793 on Build GRCh38
rs549953191 Chr.3: 111593497-111593497 on Build GRCh38
rs757959677 Chr.3: 111593530-111593530 on Build GRCh38
rs946025078 Chr.3: 111593615-111593615 on Build GRCh38
rs537798430 Chr.3: 111593418-111593418 on Build GRCh38
rs1034005819 Chr.3: 111593709-111593709 on Build GRCh38
rs7612391 Chr.3: 111593458-111593458 on Build GRCh38
rs760913690 Chr.3: 111593514-111593514 on Build GRCh38
rs779956120 Chr.3: 111593727-111593727 on Build GRCh38
rs149370111 Chr.3: 111593642-111593642 on Build GRCh38
rs746148851 Chr.3: 111593773-111593773 on Build GRCh38
rs181400868 Chr.3: 111593592-111593592 on Build GRCh38
rs953954487 Chr.3: 111593346-111593346 on Build GRCh38
rs1010388414 Chr.3: 111593499-111593499 on Build GRCh38
rs1021351057 Chr.3: 111593506-111593506 on Build GRCh38
rs747063283 Chr.3: 111593677-111593677 on Build GRCh38
rs200140607 Chr.3: 111593595-111593595 on Build GRCh38
rs918717508 Chr.3: 111593389-111593389 on Build GRCh38
rs145332036 Chr.3: 111593565-111593565 on Build GRCh38
rs892645438 Chr.3: 111593669-111593669 on Build GRCh38
rs1052645902 Chr.3: 111593668-111593668 on Build GRCh38
rs561192127 Chr.3: 111593591-111593591 on Build GRCh38
rs576772492 Chr.3: 111593755-111593755 on Build GRCh38
rs778354829 Chr.3: 111593457-111593457 on Build GRCh38
rs769800049 Chr.3: 111593577-111593577 on Build GRCh38
rs199706568 Chr.3: 111593647-111593647 on Build GRCh38
rs375236307 Chr.3: 111593764-111593764 on Build GRCh38
rs923016429 Chr.3: 111593651-111593651 on Build GRCh38
rs775860464 Chr.3: 111593779-111593779 on Build GRCh38
rs1039947115 Chr.3: 111593456-111593456 on Build GRCh38
rs773511256 Chr.3: 111593513-111593513 on Build GRCh38
rs996673849 Chr.3: 111593644-111593644 on Build GRCh38
rs760677985 Chr.3: 111593511-111593511 on Build GRCh38
rs763244553 Chr.3: 111593784-111593784 on Build GRCh38
rs980254189 Chr.3: 111593780-111593780 on Build GRCh38
rs867531661 Chr.3: 111593561-111593561 on Build GRCh38
rs569589964 Chr.3: 111593523-111593523 on Build GRCh38
rs755544182 Chr.3: 111593527-111593527 on Build GRCh38
rs189388435 Chr.3: 111593363-111593363 on Build GRCh38
rs138334089 Chr.3: 111593666-111593666 on Build GRCh38
rs1031585606 Chr.3: 111593472-111593472 on Build GRCh38
rs565366191 Chr.3: 111593599-111593599 on Build GRCh38
rs780059462 Chr.3: 111593516-111593516 on Build GRCh38
rs767235258 Chr.3: 111593714-111593714 on Build GRCh38
rs779821922 Chr.3: 111593656-111593656 on Build GRCh38
rs374773886 Chr.3: 111593700-111593700 on Build GRCh38
rs764516611 Chr.3: 111593703-111593703 on Build GRCh38
rs554406437 Chr.3: 111593643-111593643 on Build GRCh38
rs754329237 Chr.3: 111593526-111593526 on Build GRCh38
rs766274839 Chr.3: 111593547-111593547 on Build GRCh38
rs202000750 Chr.3: 111593528-111593528 on Build GRCh38
rs142166892 Chr.3: 111593635-111593635 on Build GRCh38
rs927165006 Chr.3: 111593382-111593382 on Build GRCh38
rs372487455 Chr.3: 111593733-111593733 on Build GRCh38
rs1006825348 Chr.3: 111593439-111593439 on Build GRCh38
rs752101088 Chr.3: 111593529-111593529 on Build GRCh38
rs1011045972 Chr.3: 111593672-111593672 on Build GRCh38
rs568155531 Chr.3: 111593667-111593667 on Build GRCh38
rs867575294 Chr.3: 111593397-111593397 on Build GRCh38
rs888431765 Chr.3: 111593422-111593422 on Build GRCh38
rs756906761 Chr.3: 111593641-111593641 on Build GRCh38
rs202151657 Chr.3: 111593708-111593708 on Build GRCh38
rs774508182 Chr.3: 111593494-111593494 on Build GRCh38
rs972405605 Chr.3: 111593435-111593435 on Build GRCh38
rs959941052 Chr.3: 111593584-111593584 on Build GRCh38
rs748504170 Chr.3: 111593409-111593409 on Build GRCh38
rs34129684 Chr.3: 111593598-111593598 on Build GRCh38
rs777260703 Chr.3: 111593531-111593531 on Build GRCh38
rs1057326925 Chr.3: 111593388-111593388 on Build GRCh38
rs144259563 Chr.3: 111593717-111593717 on Build GRCh38
rs561129460 Chr.3: 111593417-111593417 on Build GRCh38
rs763721227 Chr.3: 111593612-111593612 on Build GRCh38
rs776869952 Chr.3: 111593689-111593689 on Build GRCh38
rs768726793 Chr.3: 111593596-111593596 on Build GRCh38
rs374062388 Chr.3: 111593580-111593580 on Build GRCh38
rs972352331 Chr.3: 111593744-111593744 on Build GRCh38
rs762349211 Chr.3: 111593600-111593600 on Build GRCh38
rs149628986 Chr.3: 111593693-111593693 on Build GRCh38
rs770053365 Chr.3: 111593775-111593775 on Build GRCh38
rs756992935 Chr.3: 111593558-111593558 on Build GRCh38
rs955849752 Chr.3: 111593490-111593490 on Build GRCh38
rs927085631 Chr.3: 111593782-111593782 on Build GRCh38
rs373021069 Chr.3: 111593726-111593726 on Build GRCh38
rs146394022 Chr.3: 111593734-111593734 on Build GRCh38
rs930041384 Chr.3: 111593410-111593410 on Build GRCh38
rs754877087 Chr.3: 111593742-111593742 on Build GRCh38
rs747052026 Chr.3: 111593756-111593756 on Build GRCh38
rs775587783 Chr.3: 111593578-111593578 on Build GRCh38
rs746764792 Chr.3: 111593762-111593762 on Build GRCh38
rs781203792 Chr.3: 111593564-111593564 on Build GRCh38
rs998367842 Chr.3: 111593462-111593463 on Build GRCh38
rs767583888 Chr.3: 111593508-111593508 on Build GRCh38
rs761434494 Chr.3: 111593707-111593707 on Build GRCh38
rs749397403 Chr.3: 111593585-111593585 on Build GRCh38
rs771083137 Chr.3: 111593686-111593686 on Build GRCh38

View Full Product Details