Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112086245-112086731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf52
Entrez Gene ID
79669
Gene Name
chromosome 3 open reading frame 52
Gene Aliases
TTMP
UniGene
Hs.434247
Gene Chromosome Location
Chr.3: 112086335-112131095 on Build GRCh38
Transcripts
NM_001171747.1 NM_024616.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6385072 c.32A>G p.D11G Chr.3: 112086439-112086439 on Build GRCh38
5552169 c.116A>G p.D39G Chr.3: 112086523-112086523 on Build GRCh38
5552167 c.116A>G p.D39G Chr.3: 112086523-112086523 on Build GRCh38
1484434 c.118G>A p.E40K Chr.3: 112086525-112086525 on Build GRCh38
7244403 c.92C>T p.A31V Chr.3: 112086499-112086499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187434834 Chr.3: 112086348-112086348 on Build GRCh38
rs376310649 Chr.3: 112086403-112086403 on Build GRCh38
rs1994732 Chr.3: 112086374-112086374 on Build GRCh38
rs771216189 Chr.3: 112086584-112086584 on Build GRCh38
rs762553554 Chr.3: 112086496-112086496 on Build GRCh38
rs397713567 Chr.3: 112086592-112086593 on Build GRCh38
rs991729066 Chr.3: 112086539-112086539 on Build GRCh38
rs935642703 Chr.3: 112086613-112086613 on Build GRCh38
rs45497696 Chr.3: 112086675-112086675 on Build GRCh38
rs771449239 Chr.3: 112086409-112086409 on Build GRCh38
rs575810951 Chr.3: 112086308-112086308 on Build GRCh38
rs748223327 Chr.3: 112086541-112086541 on Build GRCh38
rs755185953 Chr.3: 112086417-112086418 on Build GRCh38
rs1019420929 Chr.3: 112086292-112086292 on Build GRCh38
rs751337492 Chr.3: 112086500-112086500 on Build GRCh38
rs531934309 Chr.3: 112086532-112086532 on Build GRCh38
rs774980584 Chr.3: 112086376-112086376 on Build GRCh38
rs539325342 Chr.3: 112086590-112086591 on Build GRCh38
rs1041472898 Chr.3: 112086607-112086607 on Build GRCh38
rs749362347 Chr.3: 112086411-112086411 on Build GRCh38
rs764787339 Chr.3: 112086441-112086441 on Build GRCh38
rs370456268 Chr.3: 112086428-112086429 on Build GRCh38
rs200879475 Chr.3: 112086565-112086567 on Build GRCh38
rs949921519 Chr.3: 112086466-112086466 on Build GRCh38
rs1047414404 Chr.3: 112086280-112086280 on Build GRCh38
rs909761219 Chr.3: 112086530-112086530 on Build GRCh38
rs563113090 Chr.3: 112086525-112086525 on Build GRCh38
rs1009853487 Chr.3: 112086370-112086370 on Build GRCh38
rs747403614 Chr.3: 112086564-112086564 on Build GRCh38
rs972174974 Chr.3: 112086472-112086472 on Build GRCh38
rs926581094 Chr.3: 112086415-112086415 on Build GRCh38
rs996531231 Chr.3: 112086302-112086302 on Build GRCh38
rs768792720 Chr.3: 112086361-112086361 on Build GRCh38
rs9880242 Chr.3: 112086273-112086273 on Build GRCh38
rs369406802 Chr.3: 112086506-112086506 on Build GRCh38
rs891331393 Chr.3: 112086585-112086585 on Build GRCh38
rs1029811423 Chr.3: 112086331-112086331 on Build GRCh38
rs3833596 Chr.3: 112086591-112086592 on Build GRCh38
rs565444400 Chr.3: 112086630-112086630 on Build GRCh38
rs373309847 Chr.3: 112086300-112086300 on Build GRCh38
rs1019112520 Chr.3: 112086440-112086440 on Build GRCh38
rs979848676 Chr.3: 112086412-112086412 on Build GRCh38
rs536563065 Chr.3: 112086678-112086678 on Build GRCh38
rs75888213 Chr.3: 112086562-112086562 on Build GRCh38
rs554515845 Chr.3: 112086465-112086465 on Build GRCh38
rs984066585 Chr.3: 112086330-112086330 on Build GRCh38
rs1054060769 Chr.3: 112086617-112086617 on Build GRCh38
rs527943414 Chr.3: 112086633-112086633 on Build GRCh38
rs895517014 Chr.3: 112086357-112086357 on Build GRCh38
rs950021128 Chr.3: 112086278-112086278 on Build GRCh38
rs1055644919 Chr.3: 112086353-112086353 on Build GRCh38
rs536714128 Chr.3: 112086281-112086281 on Build GRCh38
rs756105032 Chr.3: 112086533-112086533 on Build GRCh38
rs554526687 Chr.3: 112086709-112086709 on Build GRCh38
rs1033129283 Chr.3: 112086271-112086271 on Build GRCh38
rs917125221 Chr.3: 112086561-112086561 on Build GRCh38
rs919296351 Chr.3: 112086301-112086301 on Build GRCh38
rs752248651 Chr.3: 112086493-112086493 on Build GRCh38
rs966160416 Chr.3: 112086299-112086299 on Build GRCh38
rs778923035 Chr.3: 112086538-112086538 on Build GRCh38
rs1005634363 Chr.3: 112086285-112086285 on Build GRCh38
rs1010151022 Chr.3: 112086592-112086592 on Build GRCh38
rs200081719 Chr.3: 112086590-112086590 on Build GRCh38
rs755171890 Chr.3: 112086643-112086643 on Build GRCh38
rs1038020555 Chr.3: 112086514-112086514 on Build GRCh38
rs986337090 Chr.3: 112086282-112086282 on Build GRCh38
rs767869321 Chr.3: 112086532-112086533 on Build GRCh38
rs547488340 Chr.3: 112086655-112086655 on Build GRCh38
rs777082126 Chr.3: 112086419-112086419 on Build GRCh38
rs924383965 Chr.3: 112086609-112086609 on Build GRCh38
rs772330937 Chr.3: 112086384-112086384 on Build GRCh38
rs748373464 Chr.3: 112086378-112086378 on Build GRCh38
rs140695813 Chr.3: 112086408-112086408 on Build GRCh38
rs937147696 Chr.3: 112086352-112086352 on Build GRCh38
rs567566854 Chr.3: 112086371-112086371 on Build GRCh38
rs1000387689 Chr.3: 112086401-112086401 on Build GRCh38
rs949870284 Chr.3: 112086360-112086360 on Build GRCh38
rs377543671 Chr.3: 112086499-112086499 on Build GRCh38
rs762728493 Chr.3: 112086424-112086424 on Build GRCh38

View Full Product Details