Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112538004-112538421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG3
Entrez Gene ID
64422
Gene Name
autophagy related 3
Gene Aliases
APG3, APG3-LIKE, APG3L, PC3-96
UniGene
Hs.477126
Gene Chromosome Location
Chr.3: 112532507-112561963 on Build GRCh38
Transcripts
NM_001278712.1 NM_022488.4 XM_011513074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7233667 c.476-1G>A p.? Chr.3: 112538181-112538181 on Build GRCh38
4768973 c.481G>T p.E161* Chr.3: 112538175-112538175 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371194916 Chr.3: 112538193-112538195 on Build GRCh38
rs951852094 Chr.3: 112538250-112538250 on Build GRCh38
rs769783390 Chr.3: 112538152-112538152 on Build GRCh38
rs116705008 Chr.3: 112538385-112538385 on Build GRCh38
rs775236991 Chr.3: 112538157-112538157 on Build GRCh38
rs770545607 Chr.3: 112538118-112538118 on Build GRCh38
rs1009919331 Chr.3: 112538049-112538059 on Build GRCh38
rs754341323 Chr.3: 112538223-112538224 on Build GRCh38
rs891918656 Chr.3: 112538047-112538047 on Build GRCh38
rs1025342108 Chr.3: 112538231-112538231 on Build GRCh38
rs992103987 Chr.3: 112538311-112538311 on Build GRCh38
rs959307525 Chr.3: 112538299-112538299 on Build GRCh38
rs772009623 Chr.3: 112538195-112538195 on Build GRCh38
rs377353456 Chr.3: 112538071-112538071 on Build GRCh38
rs748161283 Chr.3: 112538186-112538187 on Build GRCh38
rs749256001 Chr.3: 112538280-112538280 on Build GRCh38
rs1001807577 Chr.3: 112538158-112538158 on Build GRCh38
rs557735471 Chr.3: 112538308-112538308 on Build GRCh38
rs1026072399 Chr.3: 112538243-112538243 on Build GRCh38
rs1007161082 Chr.3: 112538068-112538068 on Build GRCh38
rs984661899 Chr.3: 112538260-112538260 on Build GRCh38
rs922552604 Chr.3: 112538360-112538360 on Build GRCh38
rs1054575112 Chr.3: 112538393-112538393 on Build GRCh38
rs761578432 Chr.3: 112538374-112538374 on Build GRCh38
rs781002268 Chr.3: 112538123-112538123 on Build GRCh38
rs768275158 Chr.3: 112538328-112538328 on Build GRCh38
rs761960437 Chr.3: 112538185-112538185 on Build GRCh38
rs543916696 Chr.3: 112538215-112538215 on Build GRCh38
rs766678587 Chr.3: 112538205-112538205 on Build GRCh38
rs775619749 Chr.3: 112538199-112538200 on Build GRCh38
rs772294046 Chr.3: 112538196-112538199 on Build GRCh38
rs917216957 Chr.3: 112538322-112538322 on Build GRCh38
rs372735416 Chr.3: 112538198-112538200 on Build GRCh38
rs374344422 Chr.3: 112538045-112538045 on Build GRCh38
rs760856527 Chr.3: 112538203-112538203 on Build GRCh38
rs535521064 Chr.3: 112538090-112538090 on Build GRCh38
rs769857068 Chr.3: 112538188-112538188 on Build GRCh38
rs758060999 Chr.3: 112538074-112538076 on Build GRCh38
rs185552569 Chr.3: 112538095-112538095 on Build GRCh38
rs141939095 Chr.3: 112538353-112538353 on Build GRCh38
rs760790207 Chr.3: 112538209-112538214 on Build GRCh38
rs745656685 Chr.3: 112538143-112538143 on Build GRCh38
rs889404841 Chr.3: 112538057-112538057 on Build GRCh38
rs575167082 Chr.3: 112538200-112538200 on Build GRCh38
rs779869970 Chr.3: 112538312-112538315 on Build GRCh38
rs112263043 Chr.3: 112538044-112538044 on Build GRCh38
rs776739966 Chr.3: 112538195-112538195 on Build GRCh38
rs964268054 Chr.3: 112538317-112538317 on Build GRCh38
rs577422291 Chr.3: 112538333-112538333 on Build GRCh38
rs764219134 Chr.3: 112538210-112538210 on Build GRCh38

View Full Product Details