Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112609844-112610387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC80
Entrez Gene ID
151887
Gene Name
coiled-coil domain containing 80
Gene Aliases
DRO1, SSG1, URB, okuribin
UniGene
Hs.477128
Gene Chromosome Location
Chr.3: 112604386-112641143 on Build GRCh38
Transcripts
NM_199511.2 NM_199512.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
727229 c.2405G>A p.S802N Chr.3: 112609998-112609998 on Build GRCh38
4760194 c.2376G>A p.W792* Chr.3: 112610027-112610027 on Build GRCh38
3118442 c.2362A>G p.N788D Chr.3: 112610041-112610041 on Build GRCh38
5778027 c.2352C>G p.I784M Chr.3: 112610051-112610051 on Build GRCh38
6511369 c.2334G>T p.R778S Chr.3: 112610069-112610069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11305336 Chr.3: 112610094-112610094 on Build GRCh38
rs1044534141 Chr.3: 112610199-112610199 on Build GRCh38
rs771845042 Chr.3: 112610126-112610126 on Build GRCh38
rs762029547 Chr.3: 112610179-112610179 on Build GRCh38
rs374757202 Chr.3: 112610113-112610113 on Build GRCh38
rs761143986 Chr.3: 112610085-112610086 on Build GRCh38
rs796125846 Chr.3: 112610197-112610197 on Build GRCh38
rs778463679 Chr.3: 112610103-112610103 on Build GRCh38
rs61732242 Chr.3: 112610009-112610009 on Build GRCh38
rs763580261 Chr.3: 112610039-112610039 on Build GRCh38
rs780433649 Chr.3: 112610214-112610214 on Build GRCh38
rs372191896 Chr.3: 112610099-112610099 on Build GRCh38
rs540598306 Chr.3: 112610036-112610036 on Build GRCh38
rs759138615 Chr.3: 112609978-112609978 on Build GRCh38
rs754350343 Chr.3: 112610223-112610223 on Build GRCh38
rs771011068 Chr.3: 112610140-112610140 on Build GRCh38
rs773125885 Chr.3: 112610024-112610024 on Build GRCh38
rs867954025 Chr.3: 112610049-112610049 on Build GRCh38
rs116306624 Chr.3: 112610329-112610329 on Build GRCh38
rs375085037 Chr.3: 112610050-112610050 on Build GRCh38
rs376369360 Chr.3: 112610102-112610102 on Build GRCh38
rs770547172 Chr.3: 112609896-112609896 on Build GRCh38
rs926379663 Chr.3: 112610217-112610217 on Build GRCh38
rs757432596 Chr.3: 112610318-112610318 on Build GRCh38
rs773834085 Chr.3: 112610004-112610004 on Build GRCh38
rs56683778 Chr.3: 112609989-112609989 on Build GRCh38
rs979530508 Chr.3: 112610203-112610203 on Build GRCh38
rs758649316 Chr.3: 112609932-112609932 on Build GRCh38
rs759499743 Chr.3: 112610166-112610166 on Build GRCh38
rs796121999 Chr.3: 112610086-112610086 on Build GRCh38
rs939991734 Chr.3: 112610115-112610115 on Build GRCh38
rs200219122 Chr.3: 112610076-112610076 on Build GRCh38
rs778352820 Chr.3: 112610258-112610258 on Build GRCh38
rs761208387 Chr.3: 112610170-112610170 on Build GRCh38
rs368585913 Chr.3: 112609937-112609937 on Build GRCh38
rs747418513 Chr.3: 112609942-112609942 on Build GRCh38
rs772039423 Chr.3: 112610011-112610011 on Build GRCh38
rs760758784 Chr.3: 112610026-112610026 on Build GRCh38
rs78427541 Chr.3: 112610187-112610187 on Build GRCh38
rs764349419 Chr.3: 112610088-112610088 on Build GRCh38
rs997006407 Chr.3: 112610142-112610142 on Build GRCh38
rs774986920 Chr.3: 112609988-112609988 on Build GRCh38
rs926424067 Chr.3: 112609907-112609907 on Build GRCh38
rs921853548 Chr.3: 112610046-112610046 on Build GRCh38
rs756431889 Chr.3: 112610210-112610210 on Build GRCh38
rs755497188 Chr.3: 112610244-112610244 on Build GRCh38
rs542774199 Chr.3: 112610171-112610171 on Build GRCh38
rs988553792 Chr.3: 112609931-112609931 on Build GRCh38
rs1056255983 Chr.3: 112609966-112609966 on Build GRCh38
rs79343253 Chr.3: 112610196-112610197 on Build GRCh38
rs750810900 Chr.3: 112610196-112610196 on Build GRCh38
rs111693458 Chr.3: 112610182-112610182 on Build GRCh38
rs770284120 Chr.3: 112609983-112609983 on Build GRCh38
rs762888869 Chr.3: 112610048-112610048 on Build GRCh38
rs757490983 Chr.3: 112610075-112610075 on Build GRCh38
rs769387223 Chr.3: 112610253-112610253 on Build GRCh38
rs750720921 Chr.3: 112610077-112610077 on Build GRCh38
rs760486897 Chr.3: 112610138-112610138 on Build GRCh38
rs560386080 Chr.3: 112610097-112610097 on Build GRCh38
rs529462099 Chr.3: 112610151-112610151 on Build GRCh38
rs762817694 Chr.3: 112610187-112610188 on Build GRCh38
rs776387154 Chr.3: 112610105-112610106 on Build GRCh38
rs370702391 Chr.3: 112609945-112609945 on Build GRCh38
rs768156493 Chr.3: 112610094-112610096 on Build GRCh38
rs781518857 Chr.3: 112609955-112609955 on Build GRCh38
rs773142186 Chr.3: 112610132-112610132 on Build GRCh38
rs768091141 Chr.3: 112610100-112610100 on Build GRCh38
rs75709265 Chr.3: 112610094-112610094 on Build GRCh38
rs11305337 Chr.3: 112610187-112610187 on Build GRCh38
rs768378775 Chr.3: 112609990-112609990 on Build GRCh38
rs562647798 Chr.3: 112610211-112610211 on Build GRCh38
rs751781820 Chr.3: 112610074-112610074 on Build GRCh38
rs772714489 Chr.3: 112609969-112609969 on Build GRCh38
rs955702253 Chr.3: 112609902-112609902 on Build GRCh38
rs75114163 Chr.3: 112610095-112610095 on Build GRCh38
rs545083923 Chr.3: 112609892-112609892 on Build GRCh38
rs767670032 Chr.3: 112610191-112610191 on Build GRCh38
rs764868321 Chr.3: 112610186-112610187 on Build GRCh38
rs746434996 Chr.3: 112609971-112609971 on Build GRCh38

View Full Product Details