Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113003361-113003870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEPRO
Entrez Gene ID
25871
Gene Name
nucleolus and neural progenitor protein
Gene Aliases
C3orf17, NET17
UniGene
Hs.591288
Gene Chromosome Location
Chr.3: 113002444-113019733 on Build GRCh38
Transcripts
NM_001319109.1 NM_001319110.1 NM_001319111.1 NM_001319112.1 NM_001319114.1 NM_001319115.1 NM_015412.3 NR_134969.1 XM_017006102.1

SNP Detail

SNP ID Chromosome Location
rs71299332 Chr.3: 113003721-113003722 on Build GRCh38
rs143676889 Chr.3: 113003551-113003551 on Build GRCh38
rs775934019 Chr.3: 113003745-113003745 on Build GRCh38
rs1002356962 Chr.3: 113003776-113003776 on Build GRCh38
rs746242341 Chr.3: 113003688-113003688 on Build GRCh38
rs72036388 Chr.3: 113003463-113003467 on Build GRCh38
rs1010680007 Chr.3: 113003535-113003535 on Build GRCh38
rs928225012 Chr.3: 113003461-113003461 on Build GRCh38
rs899584127 Chr.3: 113003813-113003813 on Build GRCh38
rs896227010 Chr.3: 113003795-113003795 on Build GRCh38
rs16860319 Chr.3: 113003416-113003416 on Build GRCh38
rs767086505 Chr.3: 113003828-113003830 on Build GRCh38
rs562214823 Chr.3: 113003830-113003830 on Build GRCh38
rs914114917 Chr.3: 113003737-113003737 on Build GRCh38
rs796812338 Chr.3: 113003411-113003411 on Build GRCh38
rs576457404 Chr.3: 113003447-113003447 on Build GRCh38
rs368217291 Chr.3: 113003496-113003496 on Build GRCh38
rs922577065 Chr.3: 113003439-113003440 on Build GRCh38
rs928193872 Chr.3: 113003710-113003710 on Build GRCh38
rs771410036 Chr.3: 113003674-113003674 on Build GRCh38
rs577966414 Chr.3: 113003713-113003714 on Build GRCh38
rs973546298 Chr.3: 113003669-113003669 on Build GRCh38
rs142146233 Chr.3: 113003832-113003832 on Build GRCh38
rs1043449872 Chr.3: 113003796-113003796 on Build GRCh38
rs73857319 Chr.3: 113003676-113003676 on Build GRCh38
rs574504262 Chr.3: 113003538-113003538 on Build GRCh38
rs866115790 Chr.3: 113003600-113003600 on Build GRCh38
rs892710639 Chr.3: 113003469-113003469 on Build GRCh38
rs59137833 Chr.3: 113003725-113003725 on Build GRCh38
rs758479717 Chr.3: 113003596-113003596 on Build GRCh38
rs564446857 Chr.3: 113003700-113003700 on Build GRCh38
rs191353898 Chr.3: 113003613-113003613 on Build GRCh38
rs939636466 Chr.3: 113003716-113003716 on Build GRCh38
rs184160918 Chr.3: 113003636-113003636 on Build GRCh38
rs899531050 Chr.3: 113003541-113003541 on Build GRCh38
rs146824265 Chr.3: 113003668-113003668 on Build GRCh38
rs1004034580 Chr.3: 113003615-113003615 on Build GRCh38
rs1056684898 Chr.3: 113003786-113003786 on Build GRCh38
rs910391226 Chr.3: 113003405-113003405 on Build GRCh38
rs1015458825 Chr.3: 113003629-113003629 on Build GRCh38
rs960469955 Chr.3: 113003538-113003539 on Build GRCh38
rs1052566514 Chr.3: 113003504-113003504 on Build GRCh38
rs7644174 Chr.3: 113003479-113003479 on Build GRCh38
rs1011584276 Chr.3: 113003473-113003473 on Build GRCh38
rs1001926601 Chr.3: 113003505-113003505 on Build GRCh38
rs745983345 Chr.3: 113003682-113003690 on Build GRCh38
rs746545532 Chr.3: 113003602-113003602 on Build GRCh38
rs905473719 Chr.3: 113003760-113003760 on Build GRCh38
rs905380597 Chr.3: 113003489-113003489 on Build GRCh38
rs948294648 Chr.3: 113003432-113003432 on Build GRCh38
rs962433579 Chr.3: 113003633-113003633 on Build GRCh38
rs151197066 Chr.3: 113003846-113003846 on Build GRCh38
rs973791165 Chr.3: 113003418-113003418 on Build GRCh38
rs549964824 Chr.3: 113003782-113003782 on Build GRCh38
rs556235247 Chr.3: 113003522-113003522 on Build GRCh38
rs920643295 Chr.3: 113003421-113003421 on Build GRCh38
rs556619832 Chr.3: 113003692-113003692 on Build GRCh38
rs892199116 Chr.3: 113003532-113003532 on Build GRCh38
rs951961690 Chr.3: 113003568-113003568 on Build GRCh38

View Full Product Details