Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113003705-113004232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEPRO
Entrez Gene ID
25871
Gene Name
nucleolus and neural progenitor protein
Gene Aliases
C3orf17, NET17
UniGene
Hs.591288
Gene Chromosome Location
Chr.3: 113002444-113019733 on Build GRCh38
Transcripts
NM_001319109.1 NM_001319110.1 NM_001319111.1 NM_001319112.1 NM_001319114.1 NM_001319115.1 NM_015412.3 NR_134969.1 XM_017006102.1

SNP Detail

SNP ID Chromosome Location
rs757538876 Chr.3: 113004075-113004075 on Build GRCh38
rs775934019 Chr.3: 113003745-113003745 on Build GRCh38
rs1002356962 Chr.3: 113003776-113003776 on Build GRCh38
rs762032899 Chr.3: 113003980-113003980 on Build GRCh38
rs1030129439 Chr.3: 113003917-113003917 on Build GRCh38
rs1027717641 Chr.3: 113003984-113003984 on Build GRCh38
rs137890929 Chr.3: 113004098-113004098 on Build GRCh38
rs752569455 Chr.3: 113004136-113004136 on Build GRCh38
rs988651035 Chr.3: 113003928-113003928 on Build GRCh38
rs968201807 Chr.3: 113004146-113004146 on Build GRCh38
rs899584127 Chr.3: 113003813-113003813 on Build GRCh38
rs932413020 Chr.3: 113004150-113004150 on Build GRCh38
rs774522420 Chr.3: 113003974-113003974 on Build GRCh38
rs896227010 Chr.3: 113003795-113003795 on Build GRCh38
rs767086505 Chr.3: 113003828-113003830 on Build GRCh38
rs939673754 Chr.3: 113004163-113004163 on Build GRCh38
rs150380884 Chr.3: 113003919-113003919 on Build GRCh38
rs764876109 Chr.3: 113004099-113004099 on Build GRCh38
rs562214823 Chr.3: 113003830-113003830 on Build GRCh38
rs979422741 Chr.3: 113004148-113004148 on Build GRCh38
rs975880642 Chr.3: 113003914-113003914 on Build GRCh38
rs914114917 Chr.3: 113003737-113003737 on Build GRCh38
rs896648282 Chr.3: 113004018-113004018 on Build GRCh38
rs946866916 Chr.3: 113004124-113004124 on Build GRCh38
rs1035009612 Chr.3: 113004025-113004025 on Build GRCh38
rs754957742 Chr.3: 113004040-113004040 on Build GRCh38
rs914148979 Chr.3: 113004119-113004119 on Build GRCh38
rs142146233 Chr.3: 113003832-113003832 on Build GRCh38
rs1043449872 Chr.3: 113003796-113003796 on Build GRCh38
rs570314363 Chr.3: 113004158-113004158 on Build GRCh38
rs140375684 Chr.3: 113003871-113003871 on Build GRCh38
rs964621674 Chr.3: 113003913-113003913 on Build GRCh38
rs753783947 Chr.3: 113003993-113003993 on Build GRCh38
rs1047946952 Chr.3: 113004052-113004052 on Build GRCh38
rs921008020 Chr.3: 113004142-113004142 on Build GRCh38
rs537852235 Chr.3: 113004160-113004160 on Build GRCh38
rs929045342 Chr.3: 113004048-113004049 on Build GRCh38
rs1021440514 Chr.3: 113004139-113004139 on Build GRCh38
rs1017461654 Chr.3: 113003899-113003899 on Build GRCh38
rs898870940 Chr.3: 113003882-113003890 on Build GRCh38
rs913876306 Chr.3: 113003952-113003952 on Build GRCh38
rs1056684898 Chr.3: 113003786-113003786 on Build GRCh38
rs918062587 Chr.3: 113004170-113004170 on Build GRCh38
rs1005824553 Chr.3: 113004084-113004084 on Build GRCh38
rs373247207 Chr.3: 113004143-113004143 on Build GRCh38
rs1004170214 Chr.3: 113003891-113003891 on Build GRCh38
rs376002638 Chr.3: 113004064-113004064 on Build GRCh38
rs764438325 Chr.3: 113003948-113003948 on Build GRCh38
rs905473719 Chr.3: 113003760-113003760 on Build GRCh38
rs1017262917 Chr.3: 113004093-113004093 on Build GRCh38
rs955446268 Chr.3: 113003924-113003924 on Build GRCh38
rs151197066 Chr.3: 113003846-113003846 on Build GRCh38
rs1030098623 Chr.3: 113004108-113004108 on Build GRCh38
rs549964824 Chr.3: 113003782-113003782 on Build GRCh38
rs777454058 Chr.3: 113004201-113004201 on Build GRCh38
rs980727631 Chr.3: 113004009-113004009 on Build GRCh38
rs766385297 Chr.3: 113003981-113003981 on Build GRCh38
rs938276004 Chr.3: 113003996-113003996 on Build GRCh38

View Full Product Details