Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 114131040-114131541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD3
Entrez Gene ID
1814
Gene Name
dopamine receptor D3
Gene Aliases
D3DR, ETM1, FET1
UniGene
Hs.121478
Gene Chromosome Location
Chr.3: 114127623-114199407 on Build GRCh38
Transcripts
NM_000796.5 NM_001282563.2 NM_001290809.1 NM_033663.5 XM_017005829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7290897 c.968G>A p.R323Q Chr.3: 114131156-114131156 on Build GRCh38
6428595 c.1004T>G p.L335R Chr.3: 114131120-114131120 on Build GRCh38
5603428 c.955G>A p.G319R Chr.3: 114131169-114131169 on Build GRCh38
4513563 c.942C>T p.P314P Chr.3: 114131182-114131182 on Build GRCh38
201806 c.895C>T p.R299* Chr.3: 114131229-114131229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377277597 Chr.3: 114131068-114131072 on Build GRCh38
rs775904984 Chr.3: 114131254-114131254 on Build GRCh38
rs750067173 Chr.3: 114131300-114131300 on Build GRCh38
rs201716308 Chr.3: 114131215-114131215 on Build GRCh38
rs201951194 Chr.3: 114131383-114131383 on Build GRCh38
rs771118247 Chr.3: 114131088-114131088 on Build GRCh38
rs747247775 Chr.3: 114131084-114131084 on Build GRCh38
rs886756536 Chr.3: 114131334-114131334 on Build GRCh38
rs145473585 Chr.3: 114131471-114131471 on Build GRCh38
rs1046746860 Chr.3: 114131322-114131322 on Build GRCh38
rs546870579 Chr.3: 114131513-114131513 on Build GRCh38
rs867863807 Chr.3: 114131337-114131337 on Build GRCh38
rs201888918 Chr.3: 114131124-114131124 on Build GRCh38
rs746864271 Chr.3: 114131478-114131478 on Build GRCh38
rs780097954 Chr.3: 114131377-114131377 on Build GRCh38
rs144611669 Chr.3: 114131276-114131276 on Build GRCh38
rs139991171 Chr.3: 114131282-114131282 on Build GRCh38
rs767144510 Chr.3: 114131180-114131180 on Build GRCh38
rs765754670 Chr.3: 114131148-114131148 on Build GRCh38
rs149736958 Chr.3: 114131364-114131364 on Build GRCh38
rs758147383 Chr.3: 114131359-114131359 on Build GRCh38
rs61735073 Chr.3: 114131137-114131137 on Build GRCh38
rs368783016 Chr.3: 114131141-114131141 on Build GRCh38
rs200825904 Chr.3: 114131303-114131303 on Build GRCh38
rs772355095 Chr.3: 114131420-114131420 on Build GRCh38
rs752515716 Chr.3: 114131355-114131355 on Build GRCh38
rs937150085 Chr.3: 114131245-114131245 on Build GRCh38
rs73232569 Chr.3: 114131485-114131485 on Build GRCh38
rs201118680 Chr.3: 114131251-114131251 on Build GRCh38
rs201759000 Chr.3: 114131171-114131171 on Build GRCh38
rs979088112 Chr.3: 114131500-114131500 on Build GRCh38
rs1036572865 Chr.3: 114131277-114131277 on Build GRCh38
rs147915170 Chr.3: 114131340-114131340 on Build GRCh38
rs201431633 Chr.3: 114131209-114131209 on Build GRCh38
rs889058830 Chr.3: 114131341-114131341 on Build GRCh38
rs554514985 Chr.3: 114131097-114131097 on Build GRCh38
rs763468762 Chr.3: 114131255-114131255 on Build GRCh38
rs370774588 Chr.3: 114131156-114131156 on Build GRCh38
rs762190683 Chr.3: 114131419-114131419 on Build GRCh38
rs774890470 Chr.3: 114131384-114131384 on Build GRCh38
rs760230309 Chr.3: 114131440-114131440 on Build GRCh38
rs140725146 Chr.3: 114131183-114131183 on Build GRCh38
rs773528617 Chr.3: 114131083-114131083 on Build GRCh38
rs374601195 Chr.3: 114131270-114131270 on Build GRCh38
rs201876283 Chr.3: 114131252-114131252 on Build GRCh38
rs201125521 Chr.3: 114131203-114131203 on Build GRCh38
rs748022163 Chr.3: 114131229-114131229 on Build GRCh38
rs764646377 Chr.3: 114131261-114131261 on Build GRCh38
rs200227297 Chr.3: 114131068-114131068 on Build GRCh38
rs751158863 Chr.3: 114131170-114131170 on Build GRCh38
rs201430965 Chr.3: 114131380-114131380 on Build GRCh38
rs534251008 Chr.3: 114131390-114131390 on Build GRCh38
rs769792721 Chr.3: 114131253-114131253 on Build GRCh38
rs967369330 Chr.3: 114131497-114131497 on Build GRCh38
rs781432566 Chr.3: 114131363-114131363 on Build GRCh38
rs749068681 Chr.3: 114131075-114131075 on Build GRCh38
rs147733622 Chr.3: 114131160-114131160 on Build GRCh38
rs77109748 Chr.3: 114131074-114131074 on Build GRCh38
rs769849791 Chr.3: 114131365-114131365 on Build GRCh38
rs774696564 Chr.3: 114131264-114131264 on Build GRCh38
rs1025519858 Chr.3: 114131439-114131439 on Build GRCh38
rs199724096 Chr.3: 114131082-114131082 on Build GRCh38
rs758150318 Chr.3: 114131328-114131328 on Build GRCh38
rs781029932 Chr.3: 114131486-114131486 on Build GRCh38
rs200871535 Chr.3: 114131100-114131100 on Build GRCh38
rs528105486 Chr.3: 114131496-114131496 on Build GRCh38
rs1016484744 Chr.3: 114131356-114131356 on Build GRCh38
rs544517200 Chr.3: 114131326-114131326 on Build GRCh38
rs922016075 Chr.3: 114131182-114131182 on Build GRCh38
rs200135822 Chr.3: 114131184-114131184 on Build GRCh38
rs779523642 Chr.3: 114131284-114131286 on Build GRCh38
rs958347845 Chr.3: 114131205-114131205 on Build GRCh38
rs200551833 Chr.3: 114131353-114131353 on Build GRCh38
rs373127350 Chr.3: 114131096-114131096 on Build GRCh38
rs1029059769 Chr.3: 114131468-114131468 on Build GRCh38
rs376131620 Chr.3: 114131427-114131427 on Build GRCh38
rs565082082 Chr.3: 114131517-114131517 on Build GRCh38
rs200082316 Chr.3: 114131228-114131228 on Build GRCh38

View Full Product Details