Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 114171640-114172090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD3
Entrez Gene ID
1814
Gene Name
dopamine receptor D3
Gene Aliases
D3DR, ETM1, FET1
UniGene
Hs.121478
Gene Chromosome Location
Chr.3: 114127623-114199407 on Build GRCh38
Transcripts
NM_000796.5 NM_001282563.2 NM_001290809.1 NM_033663.5 XM_017005829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3585620 c.255G>C p.W85C Chr.3: 114171738-114171738 on Build GRCh38
1693219 c.170A>T p.E57V Chr.3: 114171823-114171823 on Build GRCh38
3585621 c.216T>C p.A72A Chr.3: 114171777-114171777 on Build GRCh38
1721845 c.126C>T p.A42A Chr.3: 114171867-114171867 on Build GRCh38
6442160 c.71G>A p.S24N Chr.3: 114171922-114171922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149488384 Chr.3: 114171764-114171764 on Build GRCh38
rs943787790 Chr.3: 114171910-114171910 on Build GRCh38
rs777035682 Chr.3: 114171713-114171713 on Build GRCh38
rs760223628 Chr.3: 114171989-114171989 on Build GRCh38
rs200245212 Chr.3: 114171889-114171889 on Build GRCh38
rs777136888 Chr.3: 114171817-114171817 on Build GRCh38
rs753115718 Chr.3: 114171727-114171727 on Build GRCh38
rs755918251 Chr.3: 114171771-114171771 on Build GRCh38
rs756743581 Chr.3: 114171757-114171757 on Build GRCh38
rs752602616 Chr.3: 114171939-114171939 on Build GRCh38
rs781522239 Chr.3: 114171943-114171943 on Build GRCh38
rs766372564 Chr.3: 114171917-114171917 on Build GRCh38
rs772481537 Chr.3: 114171698-114171698 on Build GRCh38
rs76256558 Chr.3: 114171738-114171738 on Build GRCh38
rs780181006 Chr.3: 114171852-114171852 on Build GRCh38
rs759386053 Chr.3: 114172017-114172017 on Build GRCh38
rs202132481 Chr.3: 114171880-114171880 on Build GRCh38
rs199834258 Chr.3: 114171688-114171688 on Build GRCh38
rs143935709 Chr.3: 114171824-114171824 on Build GRCh38
rs368727669 Chr.3: 114171828-114171828 on Build GRCh38
rs201146268 Chr.3: 114171858-114171858 on Build GRCh38
rs747070751 Chr.3: 114171814-114171814 on Build GRCh38
rs200653531 Chr.3: 114171707-114171707 on Build GRCh38
rs75091114 Chr.3: 114171709-114171709 on Build GRCh38
rs745800410 Chr.3: 114171765-114171765 on Build GRCh38
rs559792277 Chr.3: 114172040-114172040 on Build GRCh38
rs903414431 Chr.3: 114171897-114171897 on Build GRCh38
rs138000689 Chr.3: 114171726-114171726 on Build GRCh38
rs199685326 Chr.3: 114171916-114171916 on Build GRCh38
rs866207080 Chr.3: 114171786-114171786 on Build GRCh38
rs201744394 Chr.3: 114171682-114171682 on Build GRCh38
rs749923013 Chr.3: 114171914-114171914 on Build GRCh38
rs376795590 Chr.3: 114171821-114171821 on Build GRCh38
rs201960958 Chr.3: 114172044-114172044 on Build GRCh38
rs190430646 Chr.3: 114171965-114171965 on Build GRCh38
rs200654863 Chr.3: 114172016-114172016 on Build GRCh38
rs770920173 Chr.3: 114171816-114171816 on Build GRCh38
rs147452721 Chr.3: 114171966-114171966 on Build GRCh38
rs201156548 Chr.3: 114171882-114171882 on Build GRCh38
rs756117429 Chr.3: 114171849-114171849 on Build GRCh38
rs777983208 Chr.3: 114171863-114171863 on Build GRCh38
rs763684802 Chr.3: 114171837-114171837 on Build GRCh38
rs772329638 Chr.3: 114172038-114172039 on Build GRCh38
rs773355773 Chr.3: 114171808-114171808 on Build GRCh38
rs765255501 Chr.3: 114172020-114172020 on Build GRCh38
rs1012941151 Chr.3: 114172000-114172000 on Build GRCh38
rs1033532111 Chr.3: 114171669-114171669 on Build GRCh38
rs1035670361 Chr.3: 114171982-114171982 on Build GRCh38
rs749163990 Chr.3: 114171777-114171777 on Build GRCh38
rs769889675 Chr.3: 114171946-114171946 on Build GRCh38
rs947234012 Chr.3: 114171930-114171930 on Build GRCh38
rs763772611 Chr.3: 114171741-114171741 on Build GRCh38
rs200972336 Chr.3: 114171926-114171926 on Build GRCh38
rs6280 Chr.3: 114171968-114171968 on Build GRCh38
rs754759927 Chr.3: 114171927-114171927 on Build GRCh38
rs775683765 Chr.3: 114172042-114172045 on Build GRCh38
rs768909028 Chr.3: 114171903-114171903 on Build GRCh38
rs77498054 Chr.3: 114171879-114171879 on Build GRCh38
rs540966161 Chr.3: 114172009-114172009 on Build GRCh38
rs759875594 Chr.3: 114171722-114171722 on Build GRCh38
rs761362051 Chr.3: 114171907-114171907 on Build GRCh38
rs960115346 Chr.3: 114171761-114171761 on Build GRCh38
rs375257153 Chr.3: 114171703-114171703 on Build GRCh38
rs199793869 Chr.3: 114171887-114171887 on Build GRCh38
rs373701434 Chr.3: 114171944-114171944 on Build GRCh38
rs200587715 Chr.3: 114171954-114171954 on Build GRCh38
rs201554805 Chr.3: 114171913-114171913 on Build GRCh38
rs199935593 Chr.3: 114172066-114172066 on Build GRCh38
rs1021992722 Chr.3: 114171714-114171714 on Build GRCh38
rs77164329 Chr.3: 114171980-114171980 on Build GRCh38
rs138354054 Chr.3: 114171881-114171881 on Build GRCh38
rs367858277 Chr.3: 114171967-114171967 on Build GRCh38
rs3732783 Chr.3: 114171942-114171942 on Build GRCh38
rs765998815 Chr.3: 114171995-114171995 on Build GRCh38
rs766954152 Chr.3: 114171840-114171840 on Build GRCh38
rs758039676 Chr.3: 114171732-114171732 on Build GRCh38
rs200530143 Chr.3: 114171676-114171676 on Build GRCh38
rs751054980 Chr.3: 114171754-114171754 on Build GRCh38
rs755372382 Chr.3: 114171747-114171747 on Build GRCh38
rs778559103 Chr.3: 114171931-114171931 on Build GRCh38
rs144644130 Chr.3: 114171952-114171952 on Build GRCh38
rs750011387 Chr.3: 114171843-114171843 on Build GRCh38
rs974013500 Chr.3: 114172046-114172046 on Build GRCh38
rs200629672 Chr.3: 114171975-114171975 on Build GRCh38
rs748398354 Chr.3: 114171686-114171686 on Build GRCh38
rs371939561 Chr.3: 114171791-114171791 on Build GRCh38
rs199854203 Chr.3: 114171857-114171857 on Build GRCh38
rs774001173 Chr.3: 114171904-114171904 on Build GRCh38
rs143953030 Chr.3: 114171864-114171864 on Build GRCh38
rs201882973 Chr.3: 114171838-114171838 on Build GRCh38
rs201102020 Chr.3: 114171820-114171820 on Build GRCh38
rs149122773 Chr.3: 114171876-114171876 on Build GRCh38
rs760709416 Chr.3: 114171708-114171708 on Build GRCh38

View Full Product Details