Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 114178963-114179473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD3
Entrez Gene ID
1814
Gene Name
dopamine receptor D3
Gene Aliases
D3DR, ETM1, FET1
UniGene
Hs.121478
Gene Chromosome Location
Chr.3: 114127623-114199407 on Build GRCh38
Transcripts
NM_000796.5 NM_001282563.2 NM_033663.5 XM_017005829.1

SNP Detail

SNP ID Chromosome Location
rs149324973 Chr.3: 114179084-114179084 on Build GRCh38
rs1028579544 Chr.3: 114179090-114179090 on Build GRCh38
rs371511310 Chr.3: 114179250-114179250 on Build GRCh38
rs944957563 Chr.3: 114179057-114179057 on Build GRCh38
rs936213275 Chr.3: 114178995-114178995 on Build GRCh38
rs970043752 Chr.3: 114179213-114179213 on Build GRCh38
rs908021342 Chr.3: 114179339-114179339 on Build GRCh38
rs1004196461 Chr.3: 114179076-114179076 on Build GRCh38
rs114607171 Chr.3: 114179436-114179436 on Build GRCh38
rs186899365 Chr.3: 114179044-114179044 on Build GRCh38
rs565137834 Chr.3: 114179402-114179402 on Build GRCh38
rs1041053492 Chr.3: 114179058-114179058 on Build GRCh38
rs199526290 Chr.3: 114179041-114179041 on Build GRCh38
rs200452733 Chr.3: 114179012-114179012 on Build GRCh38
rs1027710532 Chr.3: 114179263-114179263 on Build GRCh38
rs200225823 Chr.3: 114178993-114178993 on Build GRCh38
rs977725509 Chr.3: 114179188-114179188 on Build GRCh38
rs922188545 Chr.3: 114179383-114179383 on Build GRCh38
rs146999634 Chr.3: 114179197-114179197 on Build GRCh38
rs544470555 Chr.3: 114179381-114179381 on Build GRCh38
rs772128353 Chr.3: 114179138-114179138 on Build GRCh38
rs200331615 Chr.3: 114179056-114179056 on Build GRCh38
rs530460761 Chr.3: 114179106-114179106 on Build GRCh38
rs1008407723 Chr.3: 114179104-114179104 on Build GRCh38
rs752070197 Chr.3: 114179069-114179069 on Build GRCh38
rs1019347976 Chr.3: 114179170-114179170 on Build GRCh38
rs920679921 Chr.3: 114178986-114178986 on Build GRCh38
rs966373113 Chr.3: 114179186-114179186 on Build GRCh38
rs907224635 Chr.3: 114179065-114179065 on Build GRCh38
rs778061095 Chr.3: 114179382-114179382 on Build GRCh38
rs201572508 Chr.3: 114179061-114179061 on Build GRCh38
rs201273386 Chr.3: 114179038-114179038 on Build GRCh38
rs956097193 Chr.3: 114179169-114179169 on Build GRCh38
rs202129893 Chr.3: 114179006-114179006 on Build GRCh38
rs201471739 Chr.3: 114179102-114179102 on Build GRCh38
rs990393572 Chr.3: 114179244-114179244 on Build GRCh38
rs957620101 Chr.3: 114179231-114179231 on Build GRCh38
rs1024681235 Chr.3: 114179204-114179204 on Build GRCh38
rs201385820 Chr.3: 114179004-114179004 on Build GRCh38
rs199792325 Chr.3: 114179082-114179082 on Build GRCh38
rs983557487 Chr.3: 114179318-114179318 on Build GRCh38
rs948834038 Chr.3: 114179018-114179018 on Build GRCh38
rs952123534 Chr.3: 114179314-114179314 on Build GRCh38
rs867313686 Chr.3: 114179401-114179401 on Build GRCh38
rs1031485129 Chr.3: 114179156-114179156 on Build GRCh38
rs1049437547 Chr.3: 114179093-114179093 on Build GRCh38
rs886057791 Chr.3: 114179009-114179009 on Build GRCh38
rs575294415 Chr.3: 114179081-114179081 on Build GRCh38
rs572695090 Chr.3: 114179320-114179320 on Build GRCh38

View Full Product Details