Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119675062-119675573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX17
Entrez Gene ID
10063
Gene Name
COX17, cytochrome c oxidase copper chaperone
Gene Aliases
-
UniGene
Hs.534383
Gene Chromosome Location
Chr.3: 119659975-119677441 on Build GRCh38
Transcripts
NM_005694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
479260 c.191G>T p.*64L Chr.3: 119675150-119675150 on Build GRCh38
5397865 c.135_136delTG p.C45fs*5 Chr.3: 119675205-119675206 on Build GRCh38
1036775 c.115G>A p.E39K Chr.3: 119675226-119675226 on Build GRCh38
6634324 c.117G>T p.E39D Chr.3: 119675224-119675224 on Build GRCh38
306948 c.153C>G p.A51A Chr.3: 119675188-119675188 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372254420 Chr.3: 119675129-119675129 on Build GRCh38
rs746738765 Chr.3: 119675265-119675268 on Build GRCh38
rs756405847 Chr.3: 119675278-119675280 on Build GRCh38
rs570849811 Chr.3: 119675358-119675358 on Build GRCh38
rs1051379978 Chr.3: 119675232-119675232 on Build GRCh38
rs560680593 Chr.3: 119675306-119675306 on Build GRCh38
rs761746207 Chr.3: 119675164-119675164 on Build GRCh38
rs909215282 Chr.3: 119675547-119675547 on Build GRCh38
rs768798403 Chr.3: 119675151-119675151 on Build GRCh38
rs758226702 Chr.3: 119675227-119675229 on Build GRCh38
rs747650021 Chr.3: 119675122-119675122 on Build GRCh38
rs1056058126 Chr.3: 119675107-119675107 on Build GRCh38
rs750418357 Chr.3: 119675102-119675102 on Build GRCh38
rs777804769 Chr.3: 119675128-119675128 on Build GRCh38
rs932014786 Chr.3: 119675127-119675127 on Build GRCh38
rs1051452984 Chr.3: 119675495-119675495 on Build GRCh38
rs568087707 Chr.3: 119675454-119675454 on Build GRCh38
rs753469233 Chr.3: 119675113-119675113 on Build GRCh38
rs530394082 Chr.3: 119675269-119675269 on Build GRCh38
rs879719668 Chr.3: 119675468-119675468 on Build GRCh38
rs946084616 Chr.3: 119675095-119675095 on Build GRCh38
rs888810493 Chr.3: 119675515-119675515 on Build GRCh38
rs967465645 Chr.3: 119675356-119675356 on Build GRCh38
rs553609859 Chr.3: 119675527-119675527 on Build GRCh38
rs754225586 Chr.3: 119675195-119675195 on Build GRCh38
rs576035241 Chr.3: 119675169-119675169 on Build GRCh38
rs1009830994 Chr.3: 119675332-119675332 on Build GRCh38
rs776349148 Chr.3: 119675134-119675134 on Build GRCh38
rs548868790 Chr.3: 119675304-119675304 on Build GRCh38
rs1020755657 Chr.3: 119675337-119675337 on Build GRCh38
rs549886919 Chr.3: 119675447-119675447 on Build GRCh38
rs750429987 Chr.3: 119675182-119675182 on Build GRCh38
rs764847539 Chr.3: 119675214-119675214 on Build GRCh38
rs1007465192 Chr.3: 119675289-119675289 on Build GRCh38
rs79456079 Chr.3: 119675469-119675469 on Build GRCh38
rs991770108 Chr.3: 119675417-119675417 on Build GRCh38
rs2293235 Chr.3: 119675321-119675321 on Build GRCh38
rs965256695 Chr.3: 119675440-119675440 on Build GRCh38
rs928661271 Chr.3: 119675391-119675391 on Build GRCh38
rs116800046 Chr.3: 119675088-119675088 on Build GRCh38
rs988079116 Chr.3: 119675428-119675428 on Build GRCh38
rs143622776 Chr.3: 119675227-119675227 on Build GRCh38
rs754629509 Chr.3: 119675210-119675210 on Build GRCh38
rs377673780 Chr.3: 119675098-119675098 on Build GRCh38
rs754642772 Chr.3: 119675118-119675118 on Build GRCh38
rs749518706 Chr.3: 119675135-119675135 on Build GRCh38
rs563978726 Chr.3: 119675126-119675126 on Build GRCh38
rs904841658 Chr.3: 119675330-119675330 on Build GRCh38
rs1021183990 Chr.3: 119675439-119675440 on Build GRCh38
rs766731819 Chr.3: 119675104-119675104 on Build GRCh38
rs777438789 Chr.3: 119675249-119675249 on Build GRCh38
rs909435806 Chr.3: 119675444-119675444 on Build GRCh38
rs779919330 Chr.3: 119675263-119675264 on Build GRCh38
rs568481033 Chr.3: 119675541-119675544 on Build GRCh38
rs761187154 Chr.3: 119675176-119675176 on Build GRCh38
rs774464411 Chr.3: 119675155-119675155 on Build GRCh38
rs942302289 Chr.3: 119675226-119675226 on Build GRCh38
rs72963142 Chr.3: 119675534-119675534 on Build GRCh38
rs958689513 Chr.3: 119675411-119675411 on Build GRCh38
rs766719676 Chr.3: 119675189-119675189 on Build GRCh38
rs148059056 Chr.3: 119675237-119675237 on Build GRCh38
rs770792629 Chr.3: 119675131-119675131 on Build GRCh38
rs1036952043 Chr.3: 119675451-119675451 on Build GRCh38
rs576821136 Chr.3: 119675322-119675322 on Build GRCh38
rs190798972 Chr.3: 119675120-119675120 on Build GRCh38
rs559931 Chr.3: 119675508-119675508 on Build GRCh38
rs746928690 Chr.3: 119675262-119675262 on Build GRCh38
rs141748315 Chr.3: 119675365-119675365 on Build GRCh38
rs757328643 Chr.3: 119675264-119675264 on Build GRCh38

View Full Product Details