Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122103761-122104270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD86
Entrez Gene ID
942
Gene Name
CD86 molecule
Gene Aliases
B7-2, B7.2, B70, CD28LG2, LAB72
UniGene
Hs.171182
Gene Chromosome Location
Chr.3: 122055362-122121143 on Build GRCh38
Transcripts
NM_001206924.1 NM_001206925.1 NM_006889.4 NM_175862.4 NM_176892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7112675 c.378G>T p.M126I Chr.3: 122103825-122103825 on Build GRCh38
7360319 c.355G>A p.G119R Chr.3: 122103802-122103802 on Build GRCh38
7100298 c.335T>C p.I112T Chr.3: 122103782-122103782 on Build GRCh38
6511434 c.350C>A p.P117H Chr.3: 122103797-122103797 on Build GRCh38
4945700 c.387A>G p.E129E Chr.3: 122103834-122103834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938607416 Chr.3: 122104221-122104221 on Build GRCh38
rs541446705 Chr.3: 122104195-122104195 on Build GRCh38
rs987246454 Chr.3: 122104035-122104035 on Build GRCh38
rs368197998 Chr.3: 122103968-122103968 on Build GRCh38
rs761299621 Chr.3: 122103911-122103911 on Build GRCh38
rs760318309 Chr.3: 122103862-122103862 on Build GRCh38
rs749851519 Chr.3: 122103815-122103815 on Build GRCh38
rs763707192 Chr.3: 122103789-122103789 on Build GRCh38
rs770052627 Chr.3: 122103811-122103811 on Build GRCh38
rs200704712 Chr.3: 122103846-122103846 on Build GRCh38
rs982155281 Chr.3: 122104218-122104218 on Build GRCh38
rs898918635 Chr.3: 122104105-122104105 on Build GRCh38
rs969820708 Chr.3: 122104214-122104214 on Build GRCh38
rs927927775 Chr.3: 122104219-122104219 on Build GRCh38
rs375186745 Chr.3: 122103871-122103871 on Build GRCh38
rs968035284 Chr.3: 122103980-122103980 on Build GRCh38
rs148848334 Chr.3: 122103897-122103897 on Build GRCh38
rs938576038 Chr.3: 122103882-122103882 on Build GRCh38
rs554894676 Chr.3: 122103858-122103858 on Build GRCh38
rs561256779 Chr.3: 122104208-122104208 on Build GRCh38
rs775704428 Chr.3: 122103812-122103812 on Build GRCh38
rs983290584 Chr.3: 122104229-122104229 on Build GRCh38
rs577148727 Chr.3: 122104226-122104226 on Build GRCh38
rs1031024494 Chr.3: 122104034-122104034 on Build GRCh38
rs975391280 Chr.3: 122103984-122103984 on Build GRCh38
rs1037408495 Chr.3: 122104012-122104012 on Build GRCh38
rs956994664 Chr.3: 122103946-122103946 on Build GRCh38
rs1036072838 Chr.3: 122104090-122104090 on Build GRCh38
rs767481719 Chr.3: 122103918-122103918 on Build GRCh38
rs969643514 Chr.3: 122103841-122103841 on Build GRCh38
rs890730571 Chr.3: 122104055-122104055 on Build GRCh38
rs762268459 Chr.3: 122103855-122103855 on Build GRCh38
rs553921677 Chr.3: 122104028-122104028 on Build GRCh38
rs769261211 Chr.3: 122103837-122103837 on Build GRCh38
rs916624078 Chr.3: 122103843-122103843 on Build GRCh38
rs911757841 Chr.3: 122104050-122104050 on Build GRCh38
rs367934963 Chr.3: 122103791-122103791 on Build GRCh38
rs759852794 Chr.3: 122103856-122103856 on Build GRCh38
rs988275288 Chr.3: 122103952-122103952 on Build GRCh38
rs1023708774 Chr.3: 122104188-122104188 on Build GRCh38
rs989598194 Chr.3: 122104141-122104141 on Build GRCh38
rs1003691070 Chr.3: 122103861-122103861 on Build GRCh38
rs1031178973 Chr.3: 122104093-122104093 on Build GRCh38
rs900199048 Chr.3: 122104019-122104019 on Build GRCh38
rs941785712 Chr.3: 122104001-122104001 on Build GRCh38
rs772975462 Chr.3: 122103859-122103859 on Build GRCh38
rs371492354 Chr.3: 122103869-122103869 on Build GRCh38
rs754437585 Chr.3: 122103891-122103891 on Build GRCh38
rs753555067 Chr.3: 122103816-122103816 on Build GRCh38
rs907750871 Chr.3: 122104234-122104234 on Build GRCh38
rs1044772200 Chr.3: 122104131-122104131 on Build GRCh38
rs999748773 Chr.3: 122104063-122104063 on Build GRCh38
rs34972450 Chr.3: 122103920-122103921 on Build GRCh38
rs905428049 Chr.3: 122104140-122104140 on Build GRCh38
rs186266609 Chr.3: 122104125-122104125 on Build GRCh38

View Full Product Details