Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122281991-122282487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASR
Entrez Gene ID
846
Gene Name
calcium sensing receptor
Gene Aliases
CAR, EIG8, FHH, FIH, GPRC2A, HHC, HHC1, HYPOC1, NSHPT, PCAR1
UniGene
Hs.435615
Gene Chromosome Location
Chr.3: 122183683-122286503 on Build GRCh38
Transcripts
NM_000388.3 NM_001178065.1 XM_005247837.2 XM_006713789.3 XM_017007324.1 XM_017007325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3586257 c.1705C>T p.P569S Chr.3: 122282209-122282209 on Build GRCh38
4669416 c.1691A>G p.E564G Chr.3: 122282195-122282195 on Build GRCh38
3586258 c.1706C>T p.P569L Chr.3: 122282210-122282210 on Build GRCh38
3914712 c.1713G>A p.G571G Chr.3: 122282217-122282217 on Build GRCh38
3586259 c.1723G>A p.D575N Chr.3: 122282227-122282227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749467949 Chr.3: 122282182-122282182 on Build GRCh38
rs768606456 Chr.3: 122282083-122282083 on Build GRCh38
rs1028245246 Chr.3: 122282457-122282457 on Build GRCh38
rs758253880 Chr.3: 122282036-122282036 on Build GRCh38
rs201225985 Chr.3: 122282278-122282278 on Build GRCh38
rs199503752 Chr.3: 122282063-122282063 on Build GRCh38
rs764894871 Chr.3: 122282277-122282277 on Build GRCh38
rs946945741 Chr.3: 122282220-122282220 on Build GRCh38
rs4678174 Chr.3: 122282024-122282024 on Build GRCh38
rs746754045 Chr.3: 122282064-122282064 on Build GRCh38
rs200534176 Chr.3: 122282109-122282109 on Build GRCh38
rs4678175 Chr.3: 122282086-122282086 on Build GRCh38
rs796166535 Chr.3: 122282446-122282446 on Build GRCh38
rs987515726 Chr.3: 122282070-122282070 on Build GRCh38
rs754420513 Chr.3: 122282347-122282347 on Build GRCh38
rs201202700 Chr.3: 122282126-122282126 on Build GRCh38
rs1032023222 Chr.3: 122282247-122282247 on Build GRCh38
rs761084315 Chr.3: 122282111-122282111 on Build GRCh38
rs759084805 Chr.3: 122282268-122282268 on Build GRCh38
rs780842974 Chr.3: 122282106-122282106 on Build GRCh38
rs1034039364 Chr.3: 122282071-122282071 on Build GRCh38
rs766140790 Chr.3: 122282121-122282121 on Build GRCh38
rs2270916 Chr.3: 122282252-122282252 on Build GRCh38
rs886041637 Chr.3: 122282134-122282134 on Build GRCh38
rs944294489 Chr.3: 122282331-122282331 on Build GRCh38
rs867038730 Chr.3: 122282166-122282166 on Build GRCh38
rs772740738 Chr.3: 122282239-122282239 on Build GRCh38
rs200878919 Chr.3: 122282217-122282217 on Build GRCh38
rs202141993 Chr.3: 122282282-122282282 on Build GRCh38
rs202124167 Chr.3: 122282264-122282264 on Build GRCh38
rs193922427 Chr.3: 122282189-122282190 on Build GRCh38
rs201859314 Chr.3: 122282246-122282246 on Build GRCh38
rs370857956 Chr.3: 122282286-122282286 on Build GRCh38
rs560880978 Chr.3: 122282388-122282388 on Build GRCh38
rs565142014 Chr.3: 122282412-122282412 on Build GRCh38
rs868017076 Chr.3: 122282175-122282175 on Build GRCh38
rs775620853 Chr.3: 122282082-122282082 on Build GRCh38
rs104893719 Chr.3: 122282161-122282161 on Build GRCh38
rs773737428 Chr.3: 122282102-122282102 on Build GRCh38
rs868390788 Chr.3: 122282203-122282203 on Build GRCh38
rs193922426 Chr.3: 122282189-122282189 on Build GRCh38
rs777646067 Chr.3: 122282167-122282167 on Build GRCh38
rs527382802 Chr.3: 122282326-122282326 on Build GRCh38
rs201955278 Chr.3: 122282169-122282169 on Build GRCh38
rs770521098 Chr.3: 122282384-122282384 on Build GRCh38
rs931442885 Chr.3: 122282400-122282400 on Build GRCh38
rs904853547 Chr.3: 122282262-122282262 on Build GRCh38
rs569159127 Chr.3: 122282068-122282068 on Build GRCh38
rs387907399 Chr.3: 122282133-122282133 on Build GRCh38
rs762199941 Chr.3: 122282090-122282090 on Build GRCh38
rs193922428 Chr.3: 122282190-122282190 on Build GRCh38
rs553224162 Chr.3: 122282298-122282298 on Build GRCh38
rs748213079 Chr.3: 122282229-122282229 on Build GRCh38
rs201033132 Chr.3: 122282253-122282253 on Build GRCh38
rs905931863 Chr.3: 122282434-122282434 on Build GRCh38
rs962610583 Chr.3: 122282421-122282421 on Build GRCh38
rs2270917 Chr.3: 122282399-122282399 on Build GRCh38
rs115230894 Chr.3: 122282135-122282135 on Build GRCh38
rs768047341 Chr.3: 122282092-122282092 on Build GRCh38
rs762381427 Chr.3: 122282281-122282281 on Build GRCh38
rs200118008 Chr.3: 122282269-122282269 on Build GRCh38
rs186279271 Chr.3: 122282172-122282172 on Build GRCh38
rs143047343 Chr.3: 122282142-122282142 on Build GRCh38
rs768820665 Chr.3: 122282183-122282183 on Build GRCh38
rs200647941 Chr.3: 122282146-122282146 on Build GRCh38
rs745306131 Chr.3: 122282407-122282407 on Build GRCh38
rs190262119 Chr.3: 122282337-122282337 on Build GRCh38
rs193922425 Chr.3: 122282180-122282180 on Build GRCh38
rs1057518409 Chr.3: 122282123-122282123 on Build GRCh38
rs776383129 Chr.3: 122282261-122282261 on Build GRCh38
rs575199288 Chr.3: 122282430-122282430 on Build GRCh38
rs552802416 Chr.3: 122282061-122282061 on Build GRCh38

View Full Product Details