Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 123915212-123915718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC14
Entrez Gene ID
64770
Gene Name
coiled-coil domain containing 14
Gene Aliases
-
UniGene
Hs.645028
Gene Chromosome Location
Chr.3: 123887383-123961412 on Build GRCh38
Transcripts
NM_001308317.1 NM_022757.4 XM_005247710.4 XM_005247711.4 XM_005247713.2 XM_005247714.2 XM_005247715.4 XM_005247716.4 XM_006713731.3 XM_006713733.3 XM_011513081.2 XM_017007056.1 XM_017007057.1 XM_017007058.1 XM_017007059.1 XM_017007060.1 XM_017007061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3057295 c.1916C>T p.P639L Chr.3: 123915245-123915245 on Build GRCh38
5570051 c.1973C>T p.T658I Chr.3: 123915545-123915545 on Build GRCh38
5001596 c.1546G>A p.D516N Chr.3: 123915615-123915615 on Build GRCh38
5620427 c.2007A>C p.K669N Chr.3: 123915634-123915634 on Build GRCh38
4478388 c.2236C>T p.P746S Chr.3: 123915282-123915282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747997561 Chr.3: 123915688-123915688 on Build GRCh38
rs779133904 Chr.3: 123915387-123915387 on Build GRCh38
rs774375265 Chr.3: 123915482-123915482 on Build GRCh38
rs751078758 Chr.3: 123915341-123915341 on Build GRCh38
rs780955739 Chr.3: 123915235-123915235 on Build GRCh38
rs141054502 Chr.3: 123915260-123915260 on Build GRCh38
rs200496498 Chr.3: 123915438-123915438 on Build GRCh38
rs762878962 Chr.3: 123915470-123915470 on Build GRCh38
rs761832514 Chr.3: 123915607-123915607 on Build GRCh38
rs768023084 Chr.3: 123915277-123915281 on Build GRCh38
rs772096114 Chr.3: 123915613-123915613 on Build GRCh38
rs762784036 Chr.3: 123915317-123915317 on Build GRCh38
rs759816096 Chr.3: 123915291-123915291 on Build GRCh38
rs779343412 Chr.3: 123915246-123915246 on Build GRCh38
rs751997140 Chr.3: 123915526-123915526 on Build GRCh38
rs778431888 Chr.3: 123915494-123915496 on Build GRCh38
rs780280104 Chr.3: 123915568-123915568 on Build GRCh38
rs13099684 Chr.3: 123915644-123915644 on Build GRCh38
rs955253913 Chr.3: 123915314-123915314 on Build GRCh38
rs753037412 Chr.3: 123915312-123915313 on Build GRCh38
rs139035263 Chr.3: 123915399-123915399 on Build GRCh38
rs558180745 Chr.3: 123915288-123915288 on Build GRCh38
rs755866458 Chr.3: 123915384-123915384 on Build GRCh38
rs758711919 Chr.3: 123915510-123915510 on Build GRCh38
rs543895784 Chr.3: 123915563-123915563 on Build GRCh38
rs746004869 Chr.3: 123915561-123915561 on Build GRCh38
rs768501219 Chr.3: 123915588-123915588 on Build GRCh38
rs777307907 Chr.3: 123915432-123915433 on Build GRCh38
rs770940140 Chr.3: 123915437-123915437 on Build GRCh38
rs778829947 Chr.3: 123915687-123915687 on Build GRCh38
rs369940399 Chr.3: 123915411-123915411 on Build GRCh38
rs778123014 Chr.3: 123915520-123915520 on Build GRCh38
rs140706654 Chr.3: 123915425-123915425 on Build GRCh38
rs111934942 Chr.3: 123915676-123915676 on Build GRCh38
rs138141215 Chr.3: 123915497-123915497 on Build GRCh38
rs775120782 Chr.3: 123915462-123915462 on Build GRCh38
rs756895289 Chr.3: 123915473-123915474 on Build GRCh38
rs201965890 Chr.3: 123915685-123915685 on Build GRCh38
rs764437658 Chr.3: 123915635-123915635 on Build GRCh38
rs747536708 Chr.3: 123915435-123915435 on Build GRCh38
rs199658457 Chr.3: 123915489-123915489 on Build GRCh38
rs376539891 Chr.3: 123915573-123915573 on Build GRCh38
rs1038054164 Chr.3: 123915630-123915630 on Build GRCh38
rs376040674 Chr.3: 123915647-123915647 on Build GRCh38
rs761851006 Chr.3: 123915483-123915483 on Build GRCh38
rs772566477 Chr.3: 123915617-123915617 on Build GRCh38
rs758288417 Chr.3: 123915594-123915598 on Build GRCh38
rs543324055 Chr.3: 123915660-123915660 on Build GRCh38
rs759191567 Chr.3: 123915631-123915631 on Build GRCh38
rs781115664 Chr.3: 123915557-123915557 on Build GRCh38
rs1031586848 Chr.3: 123915365-123915365 on Build GRCh38
rs149552137 Chr.3: 123915553-123915553 on Build GRCh38
rs11551802 Chr.3: 123915233-123915233 on Build GRCh38
rs111692567 Chr.3: 123915432-123915432 on Build GRCh38
rs750009322 Chr.3: 123915490-123915490 on Build GRCh38
rs753466359 Chr.3: 123915507-123915507 on Build GRCh38
rs773637830 Chr.3: 123915658-123915658 on Build GRCh38
rs563912586 Chr.3: 123915627-123915627 on Build GRCh38
rs200971033 Chr.3: 123915279-123915279 on Build GRCh38
rs375111506 Chr.3: 123915311-123915311 on Build GRCh38
rs755312806 Chr.3: 123915686-123915686 on Build GRCh38
rs755242665 Chr.3: 123915244-123915244 on Build GRCh38
rs780005647 Chr.3: 123915681-123915681 on Build GRCh38
rs751977146 Chr.3: 123915643-123915643 on Build GRCh38
rs151156664 Chr.3: 123915258-123915258 on Build GRCh38
rs756438758 Chr.3: 123915390-123915390 on Build GRCh38
rs953430141 Chr.3: 123915618-123915618 on Build GRCh38
rs888054701 Chr.3: 123915576-123915576 on Build GRCh38
rs780000987 Chr.3: 123915606-123915608 on Build GRCh38
rs774358831 Chr.3: 123915599-123915599 on Build GRCh38
rs867199884 Chr.3: 123915684-123915684 on Build GRCh38
rs767153571 Chr.3: 123915364-123915364 on Build GRCh38
rs753543438 Chr.3: 123915471-123915472 on Build GRCh38
rs749979822 Chr.3: 123915375-123915375 on Build GRCh38
rs148318048 Chr.3: 123915628-123915628 on Build GRCh38
rs756782020 Chr.3: 123915350-123915350 on Build GRCh38
rs745561683 Chr.3: 123915241-123915241 on Build GRCh38
rs372947520 Chr.3: 123915473-123915473 on Build GRCh38
rs939047139 Chr.3: 123915386-123915386 on Build GRCh38
rs745357130 Chr.3: 123915537-123915538 on Build GRCh38
rs771029869 Chr.3: 123915282-123915282 on Build GRCh38
rs374105030 Chr.3: 123915663-123915663 on Build GRCh38
rs373182696 Chr.3: 123915300-123915300 on Build GRCh38
rs904726391 Chr.3: 123915517-123915517 on Build GRCh38
rs145664821 Chr.3: 123915447-123915447 on Build GRCh38
rs762149010 Chr.3: 123915339-123915339 on Build GRCh38
rs760330126 Chr.3: 123915422-123915422 on Build GRCh38
rs200972510 Chr.3: 123915428-123915428 on Build GRCh38
rs746071963 Chr.3: 123915443-123915443 on Build GRCh38
rs142824937 Chr.3: 123915662-123915662 on Build GRCh38
rs774656371 Chr.3: 123915263-123915263 on Build GRCh38

View Full Product Details