Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 123944771-123945284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC14
Entrez Gene ID
64770
Gene Name
coiled-coil domain containing 14
Gene Aliases
-
UniGene
Hs.645028
Gene Chromosome Location
Chr.3: 123887383-123961412 on Build GRCh38
Transcripts
NM_001308317.1 NM_022757.4 XM_005247710.4 XM_005247711.4 XM_005247713.2 XM_005247714.2 XM_005247715.4 XM_005247716.4 XM_006713731.3 XM_006713733.3 XM_011513081.2 XM_017007056.1 XM_017007057.1 XM_017007058.1 XM_017007059.1 XM_017007060.1 XM_017007061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3586539 c.1234A>G p.I412V Chr.3: 123944979-123944979 on Build GRCh38
5863361 c.923C>T p.P308L Chr.3: 123944933-123944933 on Build GRCh38
1484525 c.1269A>G p.I423M Chr.3: 123944944-123944944 on Build GRCh38
7289242 c.1372A>G p.T458A Chr.3: 123944964-123944964 on Build GRCh38
5803822 c.883A>T p.R295W Chr.3: 123944973-123944973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952663852 Chr.3: 123945198-123945198 on Build GRCh38
rs760892803 Chr.3: 123944853-123944853 on Build GRCh38
rs891877040 Chr.3: 123944885-123944885 on Build GRCh38
rs774594651 Chr.3: 123945132-123945132 on Build GRCh38
rs961148662 Chr.3: 123945179-123945179 on Build GRCh38
rs72964825 Chr.3: 123944852-123944852 on Build GRCh38
rs772166357 Chr.3: 123944846-123944846 on Build GRCh38
rs535663464 Chr.3: 123945230-123945230 on Build GRCh38
rs1047538168 Chr.3: 123945153-123945153 on Build GRCh38
rs752711885 Chr.3: 123944971-123944971 on Build GRCh38
rs185358934 Chr.3: 123945103-123945103 on Build GRCh38
rs762503101 Chr.3: 123944972-123944972 on Build GRCh38
rs991710066 Chr.3: 123944831-123944831 on Build GRCh38
rs774386321 Chr.3: 123944809-123944809 on Build GRCh38
rs529055088 Chr.3: 123944889-123944889 on Build GRCh38
rs140742273 Chr.3: 123944919-123944919 on Build GRCh38
rs770629793 Chr.3: 123944870-123944870 on Build GRCh38
rs1023188675 Chr.3: 123945247-123945247 on Build GRCh38
rs369505518 Chr.3: 123944949-123944949 on Build GRCh38
rs1034614409 Chr.3: 123945016-123945016 on Build GRCh38
rs943996237 Chr.3: 123945239-123945239 on Build GRCh38
rs113517175 Chr.3: 123944936-123944936 on Build GRCh38
rs34246651 Chr.3: 123945258-123945258 on Build GRCh38
rs35163128 Chr.3: 123945030-123945030 on Build GRCh38
rs751186914 Chr.3: 123945022-123945022 on Build GRCh38
rs777777888 Chr.3: 123944882-123944882 on Build GRCh38
rs767983999 Chr.3: 123945056-123945056 on Build GRCh38
rs146927723 Chr.3: 123944994-123944994 on Build GRCh38
rs111414811 Chr.3: 123945214-123945214 on Build GRCh38
rs774597677 Chr.3: 123944839-123944839 on Build GRCh38
rs749417360 Chr.3: 123945073-123945073 on Build GRCh38
rs779347968 Chr.3: 123945038-123945038 on Build GRCh38
rs765259203 Chr.3: 123944930-123944930 on Build GRCh38
rs200650151 Chr.3: 123944833-123944833 on Build GRCh38
rs768440366 Chr.3: 123945235-123945235 on Build GRCh38
rs576611905 Chr.3: 123945210-123945210 on Build GRCh38
rs2682242 Chr.3: 123945188-123945188 on Build GRCh38
rs1011846838 Chr.3: 123945246-123945247 on Build GRCh38
rs776549385 Chr.3: 123944891-123944891 on Build GRCh38
rs756998108 Chr.3: 123945024-123945024 on Build GRCh38
rs997787764 Chr.3: 123945136-123945136 on Build GRCh38
rs199975294 Chr.3: 123944998-123944998 on Build GRCh38
rs754272304 Chr.3: 123945029-123945029 on Build GRCh38
rs771034813 Chr.3: 123945087-123945087 on Build GRCh38
rs1010771094 Chr.3: 123944909-123944909 on Build GRCh38
rs1050124274 Chr.3: 123944808-123944808 on Build GRCh38
rs780265619 Chr.3: 123945027-123945027 on Build GRCh38
rs901732379 Chr.3: 123945088-123945088 on Build GRCh38
rs3736532 Chr.3: 123945192-123945192 on Build GRCh38
rs199713333 Chr.3: 123945037-123945037 on Build GRCh38
rs1013240808 Chr.3: 123945063-123945063 on Build GRCh38
rs556816393 Chr.3: 123945206-123945206 on Build GRCh38
rs777995139 Chr.3: 123944802-123944802 on Build GRCh38
rs759622705 Chr.3: 123944914-123944915 on Build GRCh38
rs904692779 Chr.3: 123944944-123944944 on Build GRCh38

View Full Product Details