Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126394759-126395243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC37-AS1
Entrez Gene ID
100506907
Gene Name
CCDC37 antisense RNA 1 (head to head)
Gene Aliases
-
UniGene
Hs.369473
Gene Chromosome Location
Chr.3: 126393032-126394798 on Build GRCh38
Transcripts
NR_103787.1 NR_103788.1 NR_103789.1 NM_182628.2 XM_006713623.2 XM_017006320.1 XM_017006321.1 XM_017006322.1 XM_017006323.1 XM_017006324.1 XM_017006325.1 XM_017006326.1 XM_017006327.1

Gene Symbol
CFAP100
Entrez Gene ID
348807
Gene Name
cilia and flagella associated protein 100
Gene Aliases
CCDC37, MIA1
UniGene
Hs.591305
Gene Chromosome Location
Chr.3: 126394893-126436556 on Build GRCh38
Transcripts
NR_103787.1 NR_103788.1 NR_103789.1 NM_182628.2 XM_006713623.2 XM_017006320.1 XM_017006321.1 XM_017006322.1 XM_017006323.1 XM_017006324.1 XM_017006325.1 XM_017006326.1 XM_017006327.1

SNP Detail

SNP ID Chromosome Location
rs953784652 Chr.3: 126394805-126394805 on Build GRCh38
rs547055393 Chr.3: 126395127-126395127 on Build GRCh38
rs1015596956 Chr.3: 126394917-126394917 on Build GRCh38
rs914436425 Chr.3: 126394851-126394851 on Build GRCh38
rs940158820 Chr.3: 126395050-126395050 on Build GRCh38
rs980037289 Chr.3: 126395034-126395034 on Build GRCh38
rs1015072919 Chr.3: 126394794-126394794 on Build GRCh38
rs370589779 Chr.3: 126394799-126394799 on Build GRCh38
rs988194370 Chr.3: 126395030-126395030 on Build GRCh38
rs898527046 Chr.3: 126395076-126395076 on Build GRCh38
rs1056973412 Chr.3: 126395040-126395040 on Build GRCh38
rs995485749 Chr.3: 126395111-126395111 on Build GRCh38
rs1019821386 Chr.3: 126395155-126395155 on Build GRCh38
rs898747849 Chr.3: 126394935-126394935 on Build GRCh38
rs775442320 Chr.3: 126394813-126394813 on Build GRCh38
rs552385404 Chr.3: 126394962-126394962 on Build GRCh38
rs890046886 Chr.3: 126395151-126395151 on Build GRCh38
rs896582022 Chr.3: 126394913-126394913 on Build GRCh38
rs187715298 Chr.3: 126395198-126395198 on Build GRCh38
rs562689481 Chr.3: 126395010-126395010 on Build GRCh38
rs927210794 Chr.3: 126395036-126395036 on Build GRCh38
rs897966142 Chr.3: 126394997-126394997 on Build GRCh38
rs970079027 Chr.3: 126395021-126395021 on Build GRCh38
rs1037431709 Chr.3: 126395070-126395070 on Build GRCh38
rs889963181 Chr.3: 126395222-126395222 on Build GRCh38
rs928063844 Chr.3: 126394871-126394871 on Build GRCh38
rs764160136 Chr.3: 126394944-126394944 on Build GRCh38
rs1019750586 Chr.3: 126394978-126394978 on Build GRCh38
rs773423703 Chr.3: 126395136-126395136 on Build GRCh38
rs1035454841 Chr.3: 126395024-126395024 on Build GRCh38
rs988881563 Chr.3: 126394998-126394998 on Build GRCh38
rs939699606 Chr.3: 126395124-126395124 on Build GRCh38
rs780566930 Chr.3: 126394779-126394779 on Build GRCh38
rs1029372254 Chr.3: 126394778-126394778 on Build GRCh38
rs1036701852 Chr.3: 126395143-126395143 on Build GRCh38
rs182306584 Chr.3: 126394928-126394928 on Build GRCh38
rs1027904892 Chr.3: 126395019-126395020 on Build GRCh38
rs1008775879 Chr.3: 126395152-126395152 on Build GRCh38
rs934294563 Chr.3: 126394815-126394815 on Build GRCh38
rs1028431243 Chr.3: 126394950-126394950 on Build GRCh38
rs78199006 Chr.3: 126394792-126394792 on Build GRCh38
rs574499106 Chr.3: 126394859-126394859 on Build GRCh38
rs7639164 Chr.3: 126395039-126395039 on Build GRCh38
rs1056927051 Chr.3: 126394901-126394901 on Build GRCh38
rs932586768 Chr.3: 126395086-126395086 on Build GRCh38
rs960756599 Chr.3: 126395029-126395029 on Build GRCh38
rs1028486903 Chr.3: 126394802-126394802 on Build GRCh38
rs533433454 Chr.3: 126394900-126394900 on Build GRCh38
rs980783772 Chr.3: 126395023-126395023 on Build GRCh38
rs781394465 Chr.3: 126394972-126394972 on Build GRCh38
rs938057065 Chr.3: 126394895-126394895 on Build GRCh38
rs531506696 Chr.3: 126395012-126395012 on Build GRCh38
rs986537931 Chr.3: 126394808-126394808 on Build GRCh38
rs988505928 Chr.3: 126394849-126394849 on Build GRCh38
rs555338875 Chr.3: 126395042-126395042 on Build GRCh38
rs963000732 Chr.3: 126394796-126394796 on Build GRCh38
rs905722342 Chr.3: 126394861-126394861 on Build GRCh38
rs1043629277 Chr.3: 126394943-126394943 on Build GRCh38
rs1027871763 Chr.3: 126394856-126394856 on Build GRCh38
rs955820795 Chr.3: 126394993-126394993 on Build GRCh38
rs921181352 Chr.3: 126395084-126395084 on Build GRCh38
rs560446797 Chr.3: 126394916-126394916 on Build GRCh38
rs1051041821 Chr.3: 126395103-126395103 on Build GRCh38
rs769911729 Chr.3: 126395123-126395123 on Build GRCh38
rs946492147 Chr.3: 126395037-126395037 on Build GRCh38
rs56136817 Chr.3: 126394860-126394860 on Build GRCh38
rs913713064 Chr.3: 126394930-126394930 on Build GRCh38
rs898291465 Chr.3: 126395147-126395147 on Build GRCh38
rs533829996 Chr.3: 126395125-126395125 on Build GRCh38
rs375846743 Chr.3: 126394958-126394958 on Build GRCh38
rs981220489 Chr.3: 126394863-126394863 on Build GRCh38

View Full Product Details