Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126524149-126524638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf22
Entrez Gene ID
152065
Gene Name
chromosome 3 open reading frame 22
Gene Aliases
-
UniGene
Hs.178210
Gene Chromosome Location
Chr.3: 126524439-126558965 on Build GRCh38
Transcripts
XM_017005753.1 NM_152889.2

Gene Symbol
CHST13
Entrez Gene ID
166012
Gene Name
carbohydrate sulfotransferase 13
Gene Aliases
C4ST3
UniGene
Hs.292375
Gene Chromosome Location
Chr.3: 126524288-126543291 on Build GRCh38
Transcripts
XM_017005753.1 NM_152889.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5687926 c.39C>A p.A13A Chr.3: 126524371-126524371 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1009131288 Chr.3: 126524262-126524262 on Build GRCh38
rs896119322 Chr.3: 126524425-126524425 on Build GRCh38
rs892604083 Chr.3: 126524269-126524269 on Build GRCh38
rs960644800 Chr.3: 126524187-126524187 on Build GRCh38
rs1010233344 Chr.3: 126524586-126524586 on Build GRCh38
rs539056394 Chr.3: 126524432-126524432 on Build GRCh38
rs1004538852 Chr.3: 126524427-126524427 on Build GRCh38
rs1004359845 Chr.3: 126524239-126524239 on Build GRCh38
rs968571424 Chr.3: 126524449-126524449 on Build GRCh38
rs986115949 Chr.3: 126524340-126524340 on Build GRCh38
rs1022032391 Chr.3: 126524284-126524284 on Build GRCh38
rs970714379 Chr.3: 126524382-126524382 on Build GRCh38
rs568725588 Chr.3: 126524390-126524390 on Build GRCh38
rs986222462 Chr.3: 126524558-126524558 on Build GRCh38
rs940805214 Chr.3: 126524228-126524228 on Build GRCh38
rs1035694091 Chr.3: 126524542-126524542 on Build GRCh38
rs892648490 Chr.3: 126524386-126524386 on Build GRCh38
rs1050648501 Chr.3: 126524251-126524251 on Build GRCh38
rs933218691 Chr.3: 126524458-126524458 on Build GRCh38
rs538139917 Chr.3: 126524266-126524266 on Build GRCh38
rs1023392624 Chr.3: 126524274-126524274 on Build GRCh38
rs973475811 Chr.3: 126524325-126524325 on Build GRCh38
rs911892115 Chr.3: 126524355-126524355 on Build GRCh38
rs914186788 Chr.3: 126524600-126524600 on Build GRCh38
rs886216471 Chr.3: 126524342-126524342 on Build GRCh38
rs931959296 Chr.3: 126524237-126524237 on Build GRCh38
rs770077592 Chr.3: 126524589-126524589 on Build GRCh38
rs1034674324 Chr.3: 126524288-126524288 on Build GRCh38
rs1010665570 Chr.3: 126524276-126524276 on Build GRCh38
rs547475646 Chr.3: 126524374-126524374 on Build GRCh38
rs933038903 Chr.3: 126524330-126524330 on Build GRCh38
rs1037382172 Chr.3: 126524497-126524497 on Build GRCh38
rs550478618 Chr.3: 126524389-126524389 on Build GRCh38
rs549594683 Chr.3: 126524247-126524247 on Build GRCh38
rs908369302 Chr.3: 126524203-126524203 on Build GRCh38
rs914415750 Chr.3: 126524420-126524420 on Build GRCh38
rs932054009 Chr.3: 126524329-126524329 on Build GRCh38
rs940449234 Chr.3: 126524494-126524494 on Build GRCh38
rs758922253 Chr.3: 126524407-126524407 on Build GRCh38
rs960947228 Chr.3: 126524293-126524293 on Build GRCh38
rs1043905908 Chr.3: 126524405-126524405 on Build GRCh38
rs113217824 Chr.3: 126524253-126524253 on Build GRCh38
rs995729282 Chr.3: 126524517-126524517 on Build GRCh38
rs1037715422 Chr.3: 126524230-126524230 on Build GRCh38
rs914384548 Chr.3: 126524311-126524311 on Build GRCh38
rs983045567 Chr.3: 126524201-126524201 on Build GRCh38
rs527838611 Chr.3: 126524236-126524236 on Build GRCh38
rs7613347 Chr.3: 126524576-126524576 on Build GRCh38
rs920649326 Chr.3: 126524327-126524327 on Build GRCh38
rs1015735454 Chr.3: 126524348-126524348 on Build GRCh38
rs946934863 Chr.3: 126524603-126524603 on Build GRCh38
rs921893823 Chr.3: 126524456-126524456 on Build GRCh38
rs1001900321 Chr.3: 126524286-126524286 on Build GRCh38
rs962479224 Chr.3: 126524321-126524321 on Build GRCh38
rs907526888 Chr.3: 126524460-126524460 on Build GRCh38
rs962530279 Chr.3: 126524491-126524491 on Build GRCh38
rs1015702936 Chr.3: 126524243-126524243 on Build GRCh38
rs1001951225 Chr.3: 126524408-126524408 on Build GRCh38
rs1015850826 Chr.3: 126524309-126524309 on Build GRCh38
rs920598336 Chr.3: 126524233-126524233 on Build GRCh38
rs934715254 Chr.3: 126524302-126524302 on Build GRCh38
rs1015487151 Chr.3: 126524447-126524447 on Build GRCh38
rs898631597 Chr.3: 126524361-126524361 on Build GRCh38
rs1051846658 Chr.3: 126524338-126524338 on Build GRCh38
rs1004561414 Chr.3: 126524343-126524343 on Build GRCh38
rs988854264 Chr.3: 126524415-126524415 on Build GRCh38
rs973905793 Chr.3: 126524506-126524506 on Build GRCh38
rs1035133528 Chr.3: 126524414-126524414 on Build GRCh38
rs1039128334 Chr.3: 126524223-126524223 on Build GRCh38
rs943376418 Chr.3: 126524382-126524402 on Build GRCh38
rs982712737 Chr.3: 126524296-126524296 on Build GRCh38
rs557385385 Chr.3: 126524519-126524519 on Build GRCh38
rs748914588 Chr.3: 126524221-126524221 on Build GRCh38
rs953500220 Chr.3: 126524529-126524529 on Build GRCh38
rs1051454680 Chr.3: 126524459-126524459 on Build GRCh38
rs974275187 Chr.3: 126524268-126524268 on Build GRCh38
rs996171882 Chr.3: 126524372-126524372 on Build GRCh38
rs974976774 Chr.3: 126524452-126524452 on Build GRCh38
rs1052664231 Chr.3: 126524383-126524383 on Build GRCh38
rs969057585 Chr.3: 126524285-126524285 on Build GRCh38
rs944702565 Chr.3: 126524578-126524578 on Build GRCh38
rs988655628 Chr.3: 126524308-126524310 on Build GRCh38
rs1038917531 Chr.3: 126524324-126524324 on Build GRCh38
rs1003512169 Chr.3: 126524522-126524522 on Build GRCh38

View Full Product Details