Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126542234-126542767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf22
Entrez Gene ID
152065
Gene Name
chromosome 3 open reading frame 22
Gene Aliases
-
UniGene
Hs.178210
Gene Chromosome Location
Chr.3: 126524439-126558965 on Build GRCh38
Transcripts
NR_130715.1 XM_017005753.1 NM_152889.2

Gene Symbol
CHST13
Entrez Gene ID
166012
Gene Name
carbohydrate sulfotransferase 13
Gene Aliases
C4ST3
UniGene
Hs.292375
Gene Chromosome Location
Chr.3: 126524288-126543291 on Build GRCh38
Transcripts
NR_130715.1 XM_017005753.1 NM_152889.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672328 c.783C>T p.Y261Y Chr.3: 126542335-126542335 on Build GRCh38
4672329 c.961G>A p.D321N Chr.3: 126542513-126542513 on Build GRCh38
3586943 c.960C>T p.F320F Chr.3: 126542512-126542512 on Build GRCh38
4672327 c.746G>A p.R249H Chr.3: 126542298-126542298 on Build GRCh38
7328911 c.736C>T p.H246Y Chr.3: 126542288-126542288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777225443 Chr.3: 126542530-126542530 on Build GRCh38
rs765728786 Chr.3: 126542300-126542300 on Build GRCh38
rs762369449 Chr.3: 126542274-126542274 on Build GRCh38
rs1056523 Chr.3: 126542364-126542364 on Build GRCh38
rs980884408 Chr.3: 126542725-126542725 on Build GRCh38
rs755955147 Chr.3: 126542436-126542436 on Build GRCh38
rs766798602 Chr.3: 126542559-126542559 on Build GRCh38
rs781150127 Chr.3: 126542334-126542334 on Build GRCh38
rs774980258 Chr.3: 126542584-126542584 on Build GRCh38
rs772828007 Chr.3: 126542479-126542479 on Build GRCh38
rs766098235 Chr.3: 126542513-126542513 on Build GRCh38
rs996648340 Chr.3: 126542544-126542544 on Build GRCh38
rs775724154 Chr.3: 126542551-126542551 on Build GRCh38
rs769922797 Chr.3: 126542343-126542343 on Build GRCh38
rs1011167370 Chr.3: 126542311-126542311 on Build GRCh38
rs368566139 Chr.3: 126542398-126542398 on Build GRCh38
rs761141102 Chr.3: 126542659-126542659 on Build GRCh38
rs778895015 Chr.3: 126542517-126542517 on Build GRCh38
rs775073093 Chr.3: 126542266-126542266 on Build GRCh38
rs948216756 Chr.3: 126542428-126542428 on Build GRCh38
rs746686745 Chr.3: 126542534-126542534 on Build GRCh38
rs116054348 Chr.3: 126542470-126542470 on Build GRCh38
rs948228162 Chr.3: 126542720-126542720 on Build GRCh38
rs775876794 Chr.3: 126542484-126542484 on Build GRCh38
rs374548681 Chr.3: 126542353-126542353 on Build GRCh38
rs962690436 Chr.3: 126542698-126542698 on Build GRCh38
rs534446670 Chr.3: 126542327-126542327 on Build GRCh38
rs767904337 Chr.3: 126542501-126542501 on Build GRCh38
rs1043520265 Chr.3: 126542631-126542631 on Build GRCh38
rs781562956 Chr.3: 126542429-126542429 on Build GRCh38
rs770086435 Chr.3: 126542480-126542480 on Build GRCh38
rs896037096 Chr.3: 126542679-126542679 on Build GRCh38
rs774224959 Chr.3: 126542515-126542515 on Build GRCh38
rs201021354 Chr.3: 126542579-126542579 on Build GRCh38
rs1056524 Chr.3: 126542359-126542359 on Build GRCh38
rs906756509 Chr.3: 126542435-126542435 on Build GRCh38
rs761411451 Chr.3: 126542289-126542289 on Build GRCh38
rs747651308 Chr.3: 126542477-126542477 on Build GRCh38
rs376899694 Chr.3: 126542263-126542263 on Build GRCh38
rs1034632198 Chr.3: 126542642-126542642 on Build GRCh38
rs370535687 Chr.3: 126542319-126542319 on Build GRCh38
rs540754806 Chr.3: 126542656-126542656 on Build GRCh38
rs763384644 Chr.3: 126542485-126542485 on Build GRCh38
rs1027799898 Chr.3: 126542730-126542730 on Build GRCh38
rs745595048 Chr.3: 126542339-126542339 on Build GRCh38
rs765821914 Chr.3: 126542394-126542394 on Build GRCh38
rs1001936454 Chr.3: 126542634-126542634 on Build GRCh38
rs755479275 Chr.3: 126542294-126542294 on Build GRCh38
rs747705774 Chr.3: 126542522-126542522 on Build GRCh38
rs779854156 Chr.3: 126542466-126542466 on Build GRCh38
rs758783079 Chr.3: 126542549-126542550 on Build GRCh38
rs754142853 Chr.3: 126542292-126542292 on Build GRCh38
rs763869348 Chr.3: 126542285-126542285 on Build GRCh38
rs374084728 Chr.3: 126542341-126542341 on Build GRCh38
rs546173379 Chr.3: 126542538-126542538 on Build GRCh38
rs768079076 Chr.3: 126542354-126542354 on Build GRCh38
rs533127456 Chr.3: 126542699-126542699 on Build GRCh38
rs373737256 Chr.3: 126542404-126542404 on Build GRCh38
rs751763090 Chr.3: 126542331-126542331 on Build GRCh38
rs764597932 Chr.3: 126542488-126542488 on Build GRCh38
rs759879260 Chr.3: 126542388-126542388 on Build GRCh38
rs111645062 Chr.3: 126542383-126542383 on Build GRCh38
rs564359850 Chr.3: 126542552-126542552 on Build GRCh38
rs763477922 Chr.3: 126542405-126542405 on Build GRCh38
rs569521301 Chr.3: 126542746-126542746 on Build GRCh38
rs769798290 Chr.3: 126542265-126542265 on Build GRCh38
rs973207685 Chr.3: 126542718-126542718 on Build GRCh38
rs764105342 Chr.3: 126542408-126542408 on Build GRCh38
rs148180435 Chr.3: 126542489-126542489 on Build GRCh38
rs751601431 Chr.3: 126542413-126542413 on Build GRCh38
rs757872487 Chr.3: 126542524-126542524 on Build GRCh38
rs769105010 Chr.3: 126542581-126542581 on Build GRCh38
rs867554675 Chr.3: 126542493-126542493 on Build GRCh38
rs528566206 Chr.3: 126542621-126542621 on Build GRCh38
rs1022156174 Chr.3: 126542324-126542324 on Build GRCh38
rs771769757 Chr.3: 126542478-126542478 on Build GRCh38
rs767723826 Chr.3: 126542609-126542609 on Build GRCh38
rs570511427 Chr.3: 126542290-126542290 on Build GRCh38
rs776529195 Chr.3: 126542260-126542260 on Build GRCh38
rs751908356 Chr.3: 126542624-126542624 on Build GRCh38
rs756306340 Chr.3: 126542509-126542509 on Build GRCh38
rs1038511151 Chr.3: 126542419-126542419 on Build GRCh38
rs778097791 Chr.3: 126542329-126542329 on Build GRCh38
rs533489820 Chr.3: 126542747-126542747 on Build GRCh38
rs562222002 Chr.3: 126542683-126542683 on Build GRCh38
rs778289276 Chr.3: 126542473-126542473 on Build GRCh38
rs773365084 Chr.3: 126542615-126542615 on Build GRCh38
rs762028792 Chr.3: 126542590-126542590 on Build GRCh38
rs1056522 Chr.3: 126542502-126542502 on Build GRCh38
rs891885280 Chr.3: 126542591-126542591 on Build GRCh38
rs772359081 Chr.3: 126542284-126542284 on Build GRCh38
rs904545218 Chr.3: 126542633-126542633 on Build GRCh38
rs750988620 Chr.3: 126542269-126542270 on Build GRCh38
rs750305343 Chr.3: 126542430-126542430 on Build GRCh38
rs768577396 Chr.3: 126542471-126542471 on Build GRCh38
rs757560097 Chr.3: 126542423-126542423 on Build GRCh38
rs184400732 Chr.3: 126542702-126542702 on Build GRCh38

View Full Product Details