Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 127674239-127674718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABTB1
Entrez Gene ID
80325
Gene Name
ankyrin repeat and BTB domain containing 1
Gene Aliases
BPOZ, BTB3, BTBD21, EF1ABP, PP2259
UniGene
Hs.107812
Gene Chromosome Location
Chr.3: 127672928-127680926 on Build GRCh38
Transcripts
NM_032548.3 NM_172027.2 NR_033429.1 XM_006713769.3 XM_011513213.1 XM_017007285.1 XM_017007286.1 XM_017007287.1 XM_017007288.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5736743 c.124T>C p.Y42H Chr.3: 127674549-127674549 on Build GRCh38
4974622 c.142C>A p.H48N Chr.3: 127674567-127674567 on Build GRCh38
238652 c.106G>A p.D36N Chr.3: 127674440-127674440 on Build GRCh38
6314759 c.154G>T p.V52L Chr.3: 127674579-127674579 on Build GRCh38
1616853 c.113delC p.L40fs*38 Chr.3: 127674447-127674447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989858882 Chr.3: 127674566-127674566 on Build GRCh38
rs377204168 Chr.3: 127674655-127674655 on Build GRCh38
rs763634953 Chr.3: 127674557-127674557 on Build GRCh38
rs773359049 Chr.3: 127674534-127674537 on Build GRCh38
rs535145806 Chr.3: 127674286-127674286 on Build GRCh38
rs747285611 Chr.3: 127674624-127674624 on Build GRCh38
rs369258777 Chr.3: 127674618-127674618 on Build GRCh38
rs201970604 Chr.3: 127674625-127674625 on Build GRCh38
rs897811313 Chr.3: 127674425-127674425 on Build GRCh38
rs938928341 Chr.3: 127674663-127674663 on Build GRCh38
rs772221285 Chr.3: 127674408-127674408 on Build GRCh38
rs543860148 Chr.3: 127674622-127674622 on Build GRCh38
rs772420559 Chr.3: 127674480-127674480 on Build GRCh38
rs976205964 Chr.3: 127674296-127674296 on Build GRCh38
rs932047599 Chr.3: 127674320-127674320 on Build GRCh38
rs867640759 Chr.3: 127674630-127674630 on Build GRCh38
rs746324247 Chr.3: 127674375-127674375 on Build GRCh38
rs142908756 Chr.3: 127674432-127674432 on Build GRCh38
rs769776751 Chr.3: 127674527-127674530 on Build GRCh38
rs543867724 Chr.3: 127674675-127674675 on Build GRCh38
rs763590394 Chr.3: 127674438-127674438 on Build GRCh38
rs199986435 Chr.3: 127674582-127674582 on Build GRCh38
rs940958662 Chr.3: 127674335-127674335 on Build GRCh38
rs753860555 Chr.3: 127674417-127674417 on Build GRCh38
rs977381045 Chr.3: 127674611-127674611 on Build GRCh38
rs149015195 Chr.3: 127674670-127674670 on Build GRCh38
rs746062317 Chr.3: 127674649-127674649 on Build GRCh38
rs1042010080 Chr.3: 127674282-127674282 on Build GRCh38
rs374504962 Chr.3: 127674345-127674345 on Build GRCh38
rs781335293 Chr.3: 127674464-127674465 on Build GRCh38
rs373134391 Chr.3: 127674674-127674674 on Build GRCh38
rs776782696 Chr.3: 127674632-127674632 on Build GRCh38
rs1056184886 Chr.3: 127674667-127674667 on Build GRCh38
rs1049256066 Chr.3: 127674321-127674321 on Build GRCh38
rs375716769 Chr.3: 127674673-127674673 on Build GRCh38
rs751025720 Chr.3: 127674445-127674445 on Build GRCh38
rs113834781 Chr.3: 127674659-127674660 on Build GRCh38
rs942116773 Chr.3: 127674639-127674639 on Build GRCh38
rs748316565 Chr.3: 127674478-127674478 on Build GRCh38
rs762801220 Chr.3: 127674537-127674537 on Build GRCh38
rs62264122 Chr.3: 127674677-127674677 on Build GRCh38
rs371938330 Chr.3: 127674584-127674584 on Build GRCh38
rs928910889 Chr.3: 127674658-127674658 on Build GRCh38
rs749841270 Chr.3: 127674626-127674627 on Build GRCh38
rs371432584 Chr.3: 127674671-127674671 on Build GRCh38
rs769712238 Chr.3: 127674637-127674637 on Build GRCh38
rs766212718 Chr.3: 127674415-127674415 on Build GRCh38
rs115405284 Chr.3: 127674633-127674633 on Build GRCh38
rs1037776087 Chr.3: 127674640-127674640 on Build GRCh38
rs374859435 Chr.3: 127674588-127674588 on Build GRCh38
rs774974114 Chr.3: 127674633-127674654 on Build GRCh38
rs372256421 Chr.3: 127674462-127674462 on Build GRCh38
rs775461361 Chr.3: 127674651-127674651 on Build GRCh38
rs760876224 Chr.3: 127674413-127674413 on Build GRCh38
rs762149389 Chr.3: 127674407-127674407 on Build GRCh38
rs759488569 Chr.3: 127674495-127674495 on Build GRCh38
rs770202809 Chr.3: 127674376-127674376 on Build GRCh38
rs747036426 Chr.3: 127674486-127674486 on Build GRCh38
rs779434009 Chr.3: 127674475-127674475 on Build GRCh38
rs368950072 Chr.3: 127674494-127674494 on Build GRCh38
rs1036983325 Chr.3: 127674336-127674336 on Build GRCh38
rs555066978 Chr.3: 127674653-127674653 on Build GRCh38
rs748386833 Chr.3: 127674470-127674470 on Build GRCh38
rs1008102184 Chr.3: 127674519-127674519 on Build GRCh38
rs1031305879 Chr.3: 127674535-127674535 on Build GRCh38
rs780672598 Chr.3: 127674386-127674386 on Build GRCh38
rs755546763 Chr.3: 127674603-127674603 on Build GRCh38
rs907064896 Chr.3: 127674284-127674284 on Build GRCh38
rs757538620 Chr.3: 127674297-127674297 on Build GRCh38
rs145860347 Chr.3: 127674652-127674652 on Build GRCh38
rs375767771 Chr.3: 127674356-127674356 on Build GRCh38
rs775265477 Chr.3: 127674499-127674499 on Build GRCh38
rs757860241 Chr.3: 127674355-127674355 on Build GRCh38
rs755349408 Chr.3: 127674473-127674473 on Build GRCh38
rs148235279 Chr.3: 127674429-127674429 on Build GRCh38
rs13080584 Chr.3: 127674614-127674614 on Build GRCh38
rs771511963 Chr.3: 127674632-127674633 on Build GRCh38
rs527462760 Chr.3: 127674323-127674323 on Build GRCh38
rs923965993 Chr.3: 127674278-127674278 on Build GRCh38
rs748510920 Chr.3: 127674617-127674617 on Build GRCh38
rs756842905 Chr.3: 127674447-127674447 on Build GRCh38
rs771082365 Chr.3: 127674493-127674493 on Build GRCh38
rs146456729 Chr.3: 127674406-127674406 on Build GRCh38
rs143132342 Chr.3: 127674428-127674428 on Build GRCh38
rs780396289 Chr.3: 127674460-127674460 on Build GRCh38
rs938486205 Chr.3: 127674292-127674292 on Build GRCh38
rs773171364 Chr.3: 127674412-127674412 on Build GRCh38
rs571792168 Chr.3: 127674689-127674689 on Build GRCh38
rs773540062 Chr.3: 127674484-127674484 on Build GRCh38
rs779632265 Chr.3: 127674616-127674616 on Build GRCh38
rs749598564 Chr.3: 127674391-127674391 on Build GRCh38
rs13080152 Chr.3: 127674591-127674591 on Build GRCh38
rs749364568 Chr.3: 127674266-127674266 on Build GRCh38
rs369456649 Chr.3: 127674498-127674498 on Build GRCh38
rs751267762 Chr.3: 127674570-127674570 on Build GRCh38
rs909375470 Chr.3: 127674334-127674334 on Build GRCh38
rs1012001688 Chr.3: 127674440-127674440 on Build GRCh38
rs769185428 Chr.3: 127674403-127674403 on Build GRCh38
rs946026229 Chr.3: 127674269-127674269 on Build GRCh38

View Full Product Details